Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1916613-1917155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML6
Entrez Gene ID
163688
Gene Name
calmodulin like 6
Gene Aliases
CAGLP
UniGene
Hs.85902
Gene Chromosome Location
Chr.1: 1913751-1917294 on Build GRCh38
Transcripts
NM_138705.2 XM_005244729.3 XM_011540841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5732327 c.387G>C p.W129C Chr.1: 1916885-1916885 on Build GRCh38
6123466 c.399G>T p.K133N Chr.1: 1916974-1916974 on Build GRCh38
6247490 c.411G>T p.M137I Chr.1: 1916986-1916986 on Build GRCh38
6387423 c.443C>A p.A148E Chr.1: 1917018-1917018 on Build GRCh38
6619121 c.444G>A p.A148A Chr.1: 1917019-1917019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201807862 Chr.1: 1917008-1917008 on Build GRCh38
rs533293378 Chr.1: 1916823-1916823 on Build GRCh38
rs768053464 Chr.1: 1916635-1916635 on Build GRCh38
rs1048142154 Chr.1: 1916719-1916719 on Build GRCh38
rs755058878 Chr.1: 1916650-1916650 on Build GRCh38
rs534957043 Chr.1: 1916681-1916681 on Build GRCh38
rs758558534 Chr.1: 1917075-1917075 on Build GRCh38
rs775702187 Chr.1: 1916964-1916964 on Build GRCh38
rs377222826 Chr.1: 1917078-1917078 on Build GRCh38
rs778639993 Chr.1: 1916772-1916772 on Build GRCh38
rs767141727 Chr.1: 1917040-1917040 on Build GRCh38
rs766538533 Chr.1: 1916645-1916645 on Build GRCh38
rs200800474 Chr.1: 1916792-1916792 on Build GRCh38
rs938793486 Chr.1: 1916861-1916861 on Build GRCh38
rs200415259 Chr.1: 1916767-1916767 on Build GRCh38
rs747586485 Chr.1: 1916690-1916690 on Build GRCh38
rs749236965 Chr.1: 1917101-1917101 on Build GRCh38
rs755285228 Chr.1: 1916988-1916988 on Build GRCh38
rs781748394 Chr.1: 1916927-1916927 on Build GRCh38
rs372840731 Chr.1: 1916978-1916978 on Build GRCh38
rs953793471 Chr.1: 1916780-1916780 on Build GRCh38
rs779623102 Chr.1: 1916717-1916717 on Build GRCh38
rs771480524 Chr.1: 1916868-1916868 on Build GRCh38
rs749908265 Chr.1: 1917041-1917041 on Build GRCh38
rs891091428 Chr.1: 1917121-1917121 on Build GRCh38
rs886479299 Chr.1: 1916877-1916877 on Build GRCh38
rs551736613 Chr.1: 1916949-1916949 on Build GRCh38
rs1016515809 Chr.1: 1916731-1916731 on Build GRCh38
rs771081881 Chr.1: 1916720-1916721 on Build GRCh38
rs74773083 Chr.1: 1916758-1916758 on Build GRCh38
rs780474840 Chr.1: 1916983-1916983 on Build GRCh38
rs769995760 Chr.1: 1916917-1916917 on Build GRCh38
rs549785941 Chr.1: 1916905-1916905 on Build GRCh38
rs996406457 Chr.1: 1916944-1916944 on Build GRCh38
rs148911334 Chr.1: 1917005-1917005 on Build GRCh38
rs755857395 Chr.1: 1916704-1916704 on Build GRCh38
rs981627708 Chr.1: 1916903-1916903 on Build GRCh38
rs771115467 Chr.1: 1917100-1917100 on Build GRCh38
rs147764999 Chr.1: 1916989-1916989 on Build GRCh38
rs775733294 Chr.1: 1916878-1916878 on Build GRCh38
rs200257908 Chr.1: 1917039-1917039 on Build GRCh38
rs764129043 Chr.1: 1916828-1916828 on Build GRCh38
rs777038741 Chr.1: 1916796-1916796 on Build GRCh38
rs28634421 Chr.1: 1916721-1916721 on Build GRCh38
rs200019110 Chr.1: 1916655-1916655 on Build GRCh38
rs899135551 Chr.1: 1917073-1917073 on Build GRCh38
rs367726631 Chr.1: 1916771-1916771 on Build GRCh38
rs35971167 Chr.1: 1916786-1916786 on Build GRCh38
rs144769611 Chr.1: 1917056-1917056 on Build GRCh38
rs758893678 Chr.1: 1916649-1916650 on Build GRCh38
rs369396860 Chr.1: 1917077-1917077 on Build GRCh38
rs764059788 Chr.1: 1917025-1917027 on Build GRCh38
rs568745793 Chr.1: 1916711-1916735 on Build GRCh38
rs746047212 Chr.1: 1916910-1916911 on Build GRCh38
rs375029035 Chr.1: 1917047-1917047 on Build GRCh38
rs28529346 Chr.1: 1916678-1916678 on Build GRCh38
rs201233346 Chr.1: 1916822-1916822 on Build GRCh38
rs374263236 Chr.1: 1916855-1916855 on Build GRCh38
rs75157634 Chr.1: 1916804-1916804 on Build GRCh38
rs761053917 Chr.1: 1916639-1916639 on Build GRCh38
rs28472657 Chr.1: 1916682-1916682 on Build GRCh38
rs2803296 Chr.1: 1916670-1916670 on Build GRCh38
rs776875261 Chr.1: 1916757-1916757 on Build GRCh38
rs1037724089 Chr.1: 1917052-1917052 on Build GRCh38
rs774429318 Chr.1: 1916898-1916898 on Build GRCh38
rs774030209 Chr.1: 1917112-1917112 on Build GRCh38
rs767467758 Chr.1: 1916753-1916753 on Build GRCh38
rs751081773 Chr.1: 1916920-1916920 on Build GRCh38
rs372098198 Chr.1: 1916961-1916961 on Build GRCh38
rs749685769 Chr.1: 1916986-1916986 on Build GRCh38
rs747603210 Chr.1: 1916728-1916728 on Build GRCh38
rs768781392 Chr.1: 1916892-1916892 on Build GRCh38
rs553441708 Chr.1: 1916992-1916992 on Build GRCh38
rs367960952 Chr.1: 1916928-1916928 on Build GRCh38
rs947321510 Chr.1: 1917000-1917000 on Build GRCh38
rs774123848 Chr.1: 1917034-1917034 on Build GRCh38
rs771659057 Chr.1: 1916781-1916781 on Build GRCh38
rs753671207 Chr.1: 1916840-1916840 on Build GRCh38
rs781515161 Chr.1: 1916870-1916870 on Build GRCh38
rs79330154 Chr.1: 1916689-1916689 on Build GRCh38
rs752782636 Chr.1: 1916748-1916748 on Build GRCh38
rs201706077 Chr.1: 1916848-1916848 on Build GRCh38
rs770088866 Chr.1: 1916813-1916813 on Build GRCh38
rs747525246 Chr.1: 1916865-1916865 on Build GRCh38
rs377673391 Chr.1: 1916916-1916916 on Build GRCh38
rs773899751 Chr.1: 1916723-1916723 on Build GRCh38
rs746269034 Chr.1: 1916872-1916872 on Build GRCh38
rs116120710 Chr.1: 1916970-1916970 on Build GRCh38
rs753017450 Chr.1: 1917060-1917060 on Build GRCh38
rs569744216 Chr.1: 1916634-1916634 on Build GRCh38
rs763767936 Chr.1: 1917023-1917023 on Build GRCh38
rs772259710 Chr.1: 1917001-1917001 on Build GRCh38
rs781045096 Chr.1: 1916658-1916658 on Build GRCh38
rs199694729 Chr.1: 1916959-1916959 on Build GRCh38
rs759795676 Chr.1: 1916743-1916743 on Build GRCh38
rs567005335 Chr.1: 1916832-1916832 on Build GRCh38
rs201925985 Chr.1: 1916656-1916656 on Build GRCh38
rs375451264 Chr.1: 1916864-1916864 on Build GRCh38
rs368951637 Chr.1: 1917018-1917018 on Build GRCh38
rs772710272 Chr.1: 1916730-1916730 on Build GRCh38
rs563286750 Chr.1: 1916906-1916906 on Build GRCh38
rs770036839 Chr.1: 1916875-1916875 on Build GRCh38
rs766891693 Chr.1: 1917048-1917048 on Build GRCh38
rs775998951 Chr.1: 1916933-1916933 on Build GRCh38
rs761141748 Chr.1: 1916734-1916734 on Build GRCh38
rs28670453 Chr.1: 1916741-1916741 on Build GRCh38
rs747114527 Chr.1: 1917098-1917098 on Build GRCh38
rs112025436 Chr.1: 1917107-1917107 on Build GRCh38
rs977617115 Chr.1: 1916820-1916820 on Build GRCh38
rs774032426 Chr.1: 1917031-1917033 on Build GRCh38
rs909073911 Chr.1: 1917070-1917070 on Build GRCh38
rs766741105 Chr.1: 1916739-1916739 on Build GRCh38
rs768558665 Chr.1: 1917105-1917105 on Build GRCh38
rs756542049 Chr.1: 1916931-1916931 on Build GRCh38
rs755375170 Chr.1: 1917055-1917055 on Build GRCh38
rs1002892798 Chr.1: 1916687-1916687 on Build GRCh38
rs758437010 Chr.1: 1916751-1916751 on Build GRCh38
rs556966372 Chr.1: 1916733-1916733 on Build GRCh38
rs371311970 Chr.1: 1917026-1917026 on Build GRCh38
rs150985474 Chr.1: 1917019-1917019 on Build GRCh38
rs772022088 Chr.1: 1916943-1916943 on Build GRCh38
rs758018193 Chr.1: 1916857-1916857 on Build GRCh38
rs765287544 Chr.1: 1916744-1916744 on Build GRCh38
rs370135296 Chr.1: 1916829-1916829 on Build GRCh38
rs754041454 Chr.1: 1916647-1916647 on Build GRCh38
rs749599512 Chr.1: 1916934-1916934 on Build GRCh38
rs748940761 Chr.1: 1916720-1916720 on Build GRCh38
rs201343661 Chr.1: 1916909-1916909 on Build GRCh38
rs776669864 Chr.1: 1916953-1916953 on Build GRCh38
rs763158102 Chr.1: 1916890-1916890 on Build GRCh38
rs868476250 Chr.1: 1916841-1916841 on Build GRCh38
rs756676094 Chr.1: 1916923-1916923 on Build GRCh38
rs914499129 Chr.1: 1916831-1916831 on Build GRCh38
rs770043470 Chr.1: 1916913-1916923 on Build GRCh38
rs773725031 Chr.1: 1916638-1916638 on Build GRCh38
rs373605319 Chr.1: 1916649-1916649 on Build GRCh38
rs771655850 Chr.1: 1917128-1917128 on Build GRCh38
rs767103412 Chr.1: 1917093-1917094 on Build GRCh38
rs375295754 Chr.1: 1917007-1917007 on Build GRCh38
rs539887260 Chr.1: 1916791-1916791 on Build GRCh38
rs748287400 Chr.1: 1916994-1916994 on Build GRCh38
rs375098098 Chr.1: 1916968-1916968 on Build GRCh38
rs983583494 Chr.1: 1917067-1917067 on Build GRCh38
rs758105957 Chr.1: 1916918-1916918 on Build GRCh38
rs940731431 Chr.1: 1917051-1917051 on Build GRCh38
rs1038964317 Chr.1: 1917086-1917086 on Build GRCh38
rs147577765 Chr.1: 1916977-1916977 on Build GRCh38
rs763054598 Chr.1: 1916998-1916998 on Build GRCh38
rs756836406 Chr.1: 1916969-1916969 on Build GRCh38
rs201599990 Chr.1: 1916960-1916960 on Build GRCh38
rs777506687 Chr.1: 1916919-1916919 on Build GRCh38
rs566097950 Chr.1: 1916935-1916935 on Build GRCh38
rs760650110 Chr.1: 1916839-1916839 on Build GRCh38
rs754318898 Chr.1: 1917050-1917050 on Build GRCh38
rs778064441 Chr.1: 1916650-1916650 on Build GRCh38
rs778729991 Chr.1: 1916852-1916852 on Build GRCh38
rs996457067 Chr.1: 1917113-1917113 on Build GRCh38
rs200453629 Chr.1: 1916941-1916941 on Build GRCh38
rs761617038 Chr.1: 1917037-1917037 on Build GRCh38
rs747484520 Chr.1: 1916788-1916788 on Build GRCh38
rs745697944 Chr.1: 1916662-1916662 on Build GRCh38

View Full Product Details