Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134276289-134276777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
-
UniGene
Hs.721382
Gene Chromosome Location
Chr.11: 134275447-134321364 on Build GRCh38
Transcripts
NM_001080407.2 XM_017017155.1 XM_017017156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418649 c.16C>T p.L6F Chr.11: 134276756-134276756 on Build GRCh38
6418648 c.16C>T p.L6F Chr.11: 134276756-134276756 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938114375 Chr.11: 134276574-134276574 on Build GRCh38
rs546559037 Chr.11: 134276487-134276487 on Build GRCh38
rs936849073 Chr.11: 134276653-134276653 on Build GRCh38
rs902650220 Chr.11: 134276508-134276508 on Build GRCh38
rs995848475 Chr.11: 134276720-134276720 on Build GRCh38
rs1019504013 Chr.11: 134276623-134276623 on Build GRCh38
rs768754898 Chr.11: 134276465-134276465 on Build GRCh38
rs3132809 Chr.11: 134276512-134276512 on Build GRCh38
rs900682307 Chr.11: 134276716-134276716 on Build GRCh38
rs865797612 Chr.11: 134276650-134276650 on Build GRCh38
rs564929421 Chr.11: 134276510-134276510 on Build GRCh38
rs145395751 Chr.11: 134276586-134276586 on Build GRCh38
rs964331470 Chr.11: 134276377-134276377 on Build GRCh38
rs1014037725 Chr.11: 134276357-134276357 on Build GRCh38
rs530949285 Chr.11: 134276651-134276651 on Build GRCh38
rs941872480 Chr.11: 134276462-134276462 on Build GRCh38
rs907666441 Chr.11: 134276456-134276456 on Build GRCh38
rs983359923 Chr.11: 134276534-134276534 on Build GRCh38
rs900296222 Chr.11: 134276518-134276518 on Build GRCh38
rs973673246 Chr.11: 134276546-134276546 on Build GRCh38
rs967196734 Chr.11: 134276369-134276369 on Build GRCh38
rs890105114 Chr.11: 134276748-134276748 on Build GRCh38
rs918309593 Chr.11: 134276464-134276464 on Build GRCh38
rs759972510 Chr.11: 134276698-134276698 on Build GRCh38
rs967280235 Chr.11: 134276314-134276314 on Build GRCh38
rs954666672 Chr.11: 134276419-134276419 on Build GRCh38
rs943717986 Chr.11: 134276434-134276435 on Build GRCh38
rs1029914903 Chr.11: 134276526-134276526 on Build GRCh38
rs912519214 Chr.11: 134276417-134276417 on Build GRCh38
rs937980224 Chr.11: 134276476-134276476 on Build GRCh38
rs1001374942 Chr.11: 134276568-134276568 on Build GRCh38
rs569000240 Chr.11: 134276480-134276480 on Build GRCh38
rs918267890 Chr.11: 134276669-134276669 on Build GRCh38
rs181828569 Chr.11: 134276589-134276589 on Build GRCh38
rs768069662 Chr.11: 134276752-134276752 on Build GRCh38
rs920000804 Chr.11: 134276559-134276559 on Build GRCh38
rs535197411 Chr.11: 134276538-134276538 on Build GRCh38
rs949426022 Chr.11: 134276625-134276625 on Build GRCh38
rs1033915310 Chr.11: 134276329-134276329 on Build GRCh38
rs563422070 Chr.11: 134276649-134276649 on Build GRCh38
rs11223742 Chr.11: 134276339-134276339 on Build GRCh38
rs1042628276 Chr.11: 134276643-134276643 on Build GRCh38
rs949621234 Chr.11: 134276484-134276484 on Build GRCh38
rs1030052488 Chr.11: 134276722-134276722 on Build GRCh38
rs974411004 Chr.11: 134276385-134276385 on Build GRCh38
rs963223731 Chr.11: 134276542-134276542 on Build GRCh38
rs954387877 Chr.11: 134276533-134276533 on Build GRCh38
rs749498724 Chr.11: 134276416-134276416 on Build GRCh38
rs1032527403 Chr.11: 134276346-134276346 on Build GRCh38
rs920031995 Chr.11: 134276469-134276469 on Build GRCh38
rs974161522 Chr.11: 134276467-134276467 on Build GRCh38
rs1030199750 Chr.11: 134276390-134276390 on Build GRCh38
rs983776871 Chr.11: 134276438-134276438 on Build GRCh38
rs1053922111 Chr.11: 134276665-134276665 on Build GRCh38
rs985331263 Chr.11: 134276608-134276608 on Build GRCh38
rs1021035427 Chr.11: 134276360-134276360 on Build GRCh38
rs953919187 Chr.11: 134276409-134276409 on Build GRCh38
rs112981277 Chr.11: 134276524-134276524 on Build GRCh38
rs925119597 Chr.11: 134276383-134276383 on Build GRCh38
rs553290128 Chr.11: 134276529-134276529 on Build GRCh38
rs1055163571 Chr.11: 134276479-134276479 on Build GRCh38
rs539628005 Chr.11: 134276499-134276499 on Build GRCh38

View Full Product Details