Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134292069-134292575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
-
UniGene
Hs.721382
Gene Chromosome Location
Chr.11: 134275447-134321364 on Build GRCh38
Transcripts
NM_001080407.2 XM_011542566.2 XM_011542567.2 XM_011542568.2 XM_011542569.2 XM_011542570.2 XM_011542571.2 XM_011542572.1 XM_011542574.2 XM_017017155.1 XM_017017156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506921 c.733C>T p.L245L Chr.11: 134292135-134292135 on Build GRCh38
4190124 c.769T>A p.S257T Chr.11: 134292171-134292171 on Build GRCh38
381873 c.747G>T p.G249G Chr.11: 134292149-134292149 on Build GRCh38
7369517 c.760C>T p.L254F Chr.11: 134292162-134292162 on Build GRCh38
1297869 c.778G>A p.E260K Chr.11: 134292180-134292180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780802672 Chr.11: 134292214-134292214 on Build GRCh38
rs770686641 Chr.11: 134292222-134292222 on Build GRCh38
rs530517120 Chr.11: 134292425-134292425 on Build GRCh38
rs574967968 Chr.11: 134292333-134292333 on Build GRCh38
rs750731694 Chr.11: 134292167-134292167 on Build GRCh38
rs748052188 Chr.11: 134292216-134292216 on Build GRCh38
rs758676879 Chr.11: 134292180-134292180 on Build GRCh38
rs948321302 Chr.11: 134292349-134292349 on Build GRCh38
rs967595129 Chr.11: 134292544-134292544 on Build GRCh38
rs760843951 Chr.11: 134292255-134292255 on Build GRCh38
rs769709721 Chr.11: 134292218-134292218 on Build GRCh38
rs563872990 Chr.11: 134292407-134292407 on Build GRCh38
rs993386143 Chr.11: 134292521-134292521 on Build GRCh38
rs200669668 Chr.11: 134292263-134292263 on Build GRCh38
rs1052221816 Chr.11: 134292405-134292405 on Build GRCh38
rs576929800 Chr.11: 134292134-134292134 on Build GRCh38
rs546420 Chr.11: 134292543-134292543 on Build GRCh38
rs546780846 Chr.11: 134292483-134292483 on Build GRCh38
rs889015540 Chr.11: 134292426-134292426 on Build GRCh38
rs377199205 Chr.11: 134292467-134292467 on Build GRCh38
rs779902368 Chr.11: 134292185-134292185 on Build GRCh38
rs1050527485 Chr.11: 134292414-134292414 on Build GRCh38
rs79406811 Chr.11: 134292469-134292469 on Build GRCh38
rs917517005 Chr.11: 134292356-134292356 on Build GRCh38
rs553547890 Chr.11: 134292265-134292265 on Build GRCh38
rs777570906 Chr.11: 134292219-134292219 on Build GRCh38
rs946324599 Chr.11: 134292358-134292358 on Build GRCh38
rs528214192 Chr.11: 134292475-134292475 on Build GRCh38
rs927442830 Chr.11: 134292322-134292322 on Build GRCh38
rs1018721450 Chr.11: 134292452-134292452 on Build GRCh38
rs371897409 Chr.11: 134292367-134292367 on Build GRCh38
rs755312946 Chr.11: 134292498-134292498 on Build GRCh38
rs375420532 Chr.11: 134292120-134292120 on Build GRCh38
rs1144225 Chr.11: 134292518-134292518 on Build GRCh38
rs372495089 Chr.11: 134292187-134292187 on Build GRCh38
rs1028817883 Chr.11: 134292127-134292127 on Build GRCh38
rs928144611 Chr.11: 134292385-134292385 on Build GRCh38
rs780443869 Chr.11: 134292142-134292143 on Build GRCh38
rs58651308 Chr.11: 134292105-134292105 on Build GRCh38
rs961888604 Chr.11: 134292468-134292468 on Build GRCh38
rs1027318775 Chr.11: 134292475-134292476 on Build GRCh38
rs775672234 Chr.11: 134292129-134292129 on Build GRCh38
rs1046580760 Chr.11: 134292382-134292382 on Build GRCh38
rs757732787 Chr.11: 134292166-134292166 on Build GRCh38
rs1009160699 Chr.11: 134292435-134292435 on Build GRCh38
rs545821707 Chr.11: 134292440-134292440 on Build GRCh38
rs542488192 Chr.11: 134292359-134292359 on Build GRCh38
rs764125105 Chr.11: 134292144-134292144 on Build GRCh38
rs796702495 Chr.11: 134292113-134292113 on Build GRCh38
rs984112960 Chr.11: 134292492-134292492 on Build GRCh38
rs529102536 Chr.11: 134292522-134292522 on Build GRCh38
rs775727137 Chr.11: 134292236-134292236 on Build GRCh38
rs977683032 Chr.11: 134292545-134292545 on Build GRCh38
rs980967168 Chr.11: 134292288-134292288 on Build GRCh38
rs747553351 Chr.11: 134292187-134292188 on Build GRCh38
rs951765753 Chr.11: 134292280-134292280 on Build GRCh38
rs147751931 Chr.11: 134292200-134292200 on Build GRCh38
rs758639790 Chr.11: 134292141-134292142 on Build GRCh38
rs1033857681 Chr.11: 134292507-134292507 on Build GRCh38
rs893549108 Chr.11: 134292406-134292406 on Build GRCh38
rs935059066 Chr.11: 134292391-134292391 on Build GRCh38
rs145649248 Chr.11: 134292119-134292119 on Build GRCh38
rs367948889 Chr.11: 134292151-134292151 on Build GRCh38

View Full Product Details