Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 460157-460651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
-
UniGene
Hs.504416
Gene Chromosome Location
Chr.12: 460377-561892 on Build GRCh38
Transcripts
NM_173593.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6387225 c.19G>A p.A7T Chr.12: 460395-460395 on Build GRCh38
6609394 c.71G>T p.R24L Chr.12: 460447-460447 on Build GRCh38
1361665 c.144G>A p.Q48Q Chr.12: 460520-460520 on Build GRCh38
6359865 c.29C>A p.P10Q Chr.12: 460405-460405 on Build GRCh38
4410117 c.83C>G p.A28G Chr.12: 460459-460459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376271214 Chr.12: 460459-460459 on Build GRCh38
rs968962741 Chr.12: 460236-460236 on Build GRCh38
rs775018931 Chr.12: 460513-460513 on Build GRCh38
rs909461205 Chr.12: 460557-460557 on Build GRCh38
rs931235770 Chr.12: 460375-460375 on Build GRCh38
rs775917957 Chr.12: 460553-460553 on Build GRCh38
rs961112042 Chr.12: 460630-460630 on Build GRCh38
rs552875897 Chr.12: 460351-460351 on Build GRCh38
rs956137092 Chr.12: 460184-460184 on Build GRCh38
rs12307182 Chr.12: 460559-460559 on Build GRCh38
rs898124885 Chr.12: 460368-460368 on Build GRCh38
rs766979963 Chr.12: 460381-460381 on Build GRCh38
rs770051823 Chr.12: 460484-460484 on Build GRCh38
rs111479236 Chr.12: 460314-460314 on Build GRCh38
rs1044261664 Chr.12: 460400-460401 on Build GRCh38
rs752305464 Chr.12: 460437-460437 on Build GRCh38
rs968677460 Chr.12: 460479-460479 on Build GRCh38
rs987637478 Chr.12: 460297-460297 on Build GRCh38
rs763776420 Chr.12: 460568-460568 on Build GRCh38
rs11609193 Chr.12: 460520-460520 on Build GRCh38
rs748328155 Chr.12: 460480-460480 on Build GRCh38
rs904963098 Chr.12: 460606-460606 on Build GRCh38
rs201909336 Chr.12: 460486-460486 on Build GRCh38
rs771608042 Chr.12: 460511-460511 on Build GRCh38
rs758215549 Chr.12: 460464-460464 on Build GRCh38
rs763574754 Chr.12: 460446-460446 on Build GRCh38
rs1024274552 Chr.12: 460614-460614 on Build GRCh38
rs988847568 Chr.12: 460188-460189 on Build GRCh38
rs763437734 Chr.12: 460343-460343 on Build GRCh38
rs933462293 Chr.12: 460293-460293 on Build GRCh38
rs994168179 Chr.12: 460582-460582 on Build GRCh38
rs370603270 Chr.12: 460466-460466 on Build GRCh38
rs760061774 Chr.12: 460524-460524 on Build GRCh38
rs772569056 Chr.12: 460533-460533 on Build GRCh38
rs1021643354 Chr.12: 460472-460472 on Build GRCh38
rs1017173634 Chr.12: 460196-460196 on Build GRCh38
rs755724776 Chr.12: 460442-460442 on Build GRCh38
rs760299350 Chr.12: 460563-460563 on Build GRCh38
rs777775023 Chr.12: 460503-460503 on Build GRCh38
rs750323363 Chr.12: 460463-460463 on Build GRCh38
rs1038025004 Chr.12: 460605-460605 on Build GRCh38
rs534300230 Chr.12: 460263-460263 on Build GRCh38
rs762322490 Chr.12: 460538-460538 on Build GRCh38
rs1035522519 Chr.12: 460440-460440 on Build GRCh38
rs144955552 Chr.12: 460460-460460 on Build GRCh38
rs974957785 Chr.12: 460248-460248 on Build GRCh38
rs767943173 Chr.12: 460435-460435 on Build GRCh38
rs1036695135 Chr.12: 460356-460356 on Build GRCh38
rs759949676 Chr.12: 460420-460420 on Build GRCh38
rs772879961 Chr.12: 460491-460492 on Build GRCh38
rs774906715 Chr.12: 460419-460419 on Build GRCh38
rs975011802 Chr.12: 460538-460538 on Build GRCh38
rs1029683352 Chr.12: 460559-460560 on Build GRCh38
rs753325506 Chr.12: 460575-460575 on Build GRCh38
rs905712850 Chr.12: 460401-460401 on Build GRCh38
rs765893277 Chr.12: 460583-460584 on Build GRCh38
rs568259087 Chr.12: 460451-460451 on Build GRCh38
rs535679791 Chr.12: 460452-460452 on Build GRCh38
rs912328386 Chr.12: 460317-460317 on Build GRCh38

View Full Product Details