Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 547927-548423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
-
UniGene
Hs.504416
Gene Chromosome Location
Chr.12: 460377-561892 on Build GRCh38
Transcripts
NM_173593.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6137924 c.786+1G>T p.? Chr.12: 548103-548103 on Build GRCh38
3464264 c.784G>A p.A262T Chr.12: 548100-548100 on Build GRCh38
468779 c.750G>A p.K250K Chr.12: 548066-548066 on Build GRCh38
7288981 c.815T>C p.F272S Chr.12: 548259-548259 on Build GRCh38
694768 c.834C>A p.L278L Chr.12: 548278-548278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780455486 Chr.12: 548234-548234 on Build GRCh38
rs960733369 Chr.12: 548061-548061 on Build GRCh38
rs369928928 Chr.12: 548084-548084 on Build GRCh38
rs370319947 Chr.12: 548131-548131 on Build GRCh38
rs946286762 Chr.12: 548171-548171 on Build GRCh38
rs1043291362 Chr.12: 548207-548207 on Build GRCh38
rs770423770 Chr.12: 548252-548252 on Build GRCh38
rs1047048752 Chr.12: 547968-547968 on Build GRCh38
rs747337703 Chr.12: 548305-548313 on Build GRCh38
rs768969436 Chr.12: 548102-548102 on Build GRCh38
rs759475069 Chr.12: 548268-548268 on Build GRCh38
rs896474978 Chr.12: 548381-548381 on Build GRCh38
rs370189632 Chr.12: 548342-548342 on Build GRCh38
rs533014786 Chr.12: 547951-547951 on Build GRCh38
rs749975541 Chr.12: 547955-547955 on Build GRCh38
rs74056264 Chr.12: 547970-547970 on Build GRCh38
rs181878287 Chr.12: 547967-547967 on Build GRCh38
rs758817709 Chr.12: 548085-548085 on Build GRCh38
rs748660370 Chr.12: 548346-548347 on Build GRCh38
rs11063529 Chr.12: 548238-548238 on Build GRCh38
rs200361013 Chr.12: 548123-548123 on Build GRCh38
rs777394109 Chr.12: 548247-548247 on Build GRCh38
rs764602909 Chr.12: 548010-548010 on Build GRCh38
rs199512986 Chr.12: 548337-548337 on Build GRCh38
rs769310020 Chr.12: 548317-548317 on Build GRCh38
rs141804572 Chr.12: 548090-548090 on Build GRCh38
rs143095519 Chr.12: 548352-548352 on Build GRCh38
rs141391152 Chr.12: 548249-548249 on Build GRCh38
rs762736807 Chr.12: 548019-548019 on Build GRCh38
rs913519756 Chr.12: 548163-548163 on Build GRCh38
rs746721409 Chr.12: 547988-547988 on Build GRCh38
rs765414420 Chr.12: 548208-548208 on Build GRCh38
rs146986730 Chr.12: 548288-548288 on Build GRCh38
rs369168214 Chr.12: 547980-547980 on Build GRCh38
rs746137676 Chr.12: 548008-548008 on Build GRCh38
rs1000623266 Chr.12: 547964-547964 on Build GRCh38
rs762966410 Chr.12: 548333-548333 on Build GRCh38
rs1002125293 Chr.12: 548037-548037 on Build GRCh38
rs960390557 Chr.12: 548401-548401 on Build GRCh38
rs776156666 Chr.12: 547996-547996 on Build GRCh38
rs545124312 Chr.12: 547953-547953 on Build GRCh38
rs148446452 Chr.12: 548284-548284 on Build GRCh38
rs754113768 Chr.12: 548194-548194 on Build GRCh38
rs756456841 Chr.12: 548041-548041 on Build GRCh38
rs373046927 Chr.12: 548122-548122 on Build GRCh38
rs755473496 Chr.12: 548348-548348 on Build GRCh38
rs201697293 Chr.12: 548018-548018 on Build GRCh38
rs763009118 Chr.12: 548132-548132 on Build GRCh38
rs77079055 Chr.12: 548078-548078 on Build GRCh38
rs766268825 Chr.12: 548021-548021 on Build GRCh38
rs774440618 Chr.12: 548253-548253 on Build GRCh38
rs780847078 Chr.12: 548111-548111 on Build GRCh38
rs376621948 Chr.12: 548243-548243 on Build GRCh38
rs199845960 Chr.12: 548040-548040 on Build GRCh38
rs770754675 Chr.12: 548133-548133 on Build GRCh38
rs768273664 Chr.12: 547994-547994 on Build GRCh38
rs762118729 Chr.12: 548196-548196 on Build GRCh38
rs758639022 Chr.12: 548105-548106 on Build GRCh38
rs764439974 Chr.12: 548052-548052 on Build GRCh38
rs765344595 Chr.12: 548289-548289 on Build GRCh38
rs750447539 Chr.12: 548215-548215 on Build GRCh38
rs777054458 Chr.12: 548319-548342 on Build GRCh38
rs781469775 Chr.12: 548242-548242 on Build GRCh38
rs1005822557 Chr.12: 547972-547972 on Build GRCh38
rs186232793 Chr.12: 548192-548192 on Build GRCh38
rs919770545 Chr.12: 548087-548087 on Build GRCh38
rs927437894 Chr.12: 548400-548400 on Build GRCh38
rs779740750 Chr.12: 548187-548187 on Build GRCh38
rs370374991 Chr.12: 548287-548287 on Build GRCh38
rs774331919 Chr.12: 548182-548182 on Build GRCh38
rs747740942 Chr.12: 548114-548114 on Build GRCh38
rs1009544293 Chr.12: 548387-548387 on Build GRCh38
rs780575353 Chr.12: 548120-548120 on Build GRCh38
rs149065903 Chr.12: 548344-548344 on Build GRCh38
rs922475777 Chr.12: 547952-547952 on Build GRCh38
rs374755766 Chr.12: 548237-548237 on Build GRCh38
rs138017455 Chr.12: 548316-548316 on Build GRCh38
rs994049688 Chr.12: 548353-548353 on Build GRCh38
rs757422478 Chr.12: 548073-548073 on Build GRCh38
rs12820465 Chr.12: 548384-548384 on Build GRCh38
rs754062616 Chr.12: 548060-548060 on Build GRCh38
rs377452956 Chr.12: 548309-548309 on Build GRCh38
rs775712053 Chr.12: 547975-547975 on Build GRCh38
rs556823085 Chr.12: 548367-548374 on Build GRCh38
rs761346509 Chr.12: 548006-548006 on Build GRCh38
rs1038618272 Chr.12: 548315-548315 on Build GRCh38
rs747342036 Chr.12: 548091-548091 on Build GRCh38

View Full Product Details