Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2800300-2800796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP4
Entrez Gene ID
2288
Gene Name
FK506 binding protein 4
Gene Aliases
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
UniGene
Hs.524183
Gene Chromosome Location
Chr.12: 2794942-2805423 on Build GRCh38
Transcripts
NM_002014.3 XM_011520929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6014785 c.901G>A p.E301K Chr.12: 2800446-2800446 on Build GRCh38
7099445 c.936G>C p.Q312H Chr.12: 2800481-2800481 on Build GRCh38
3871453 c.920C>T p.S307F Chr.12: 2800465-2800465 on Build GRCh38
4456377 c.1007C>T p.S336F Chr.12: 2800552-2800552 on Build GRCh38
3986858 c.867T>G p.A289A Chr.12: 2800412-2800412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964211802 Chr.12: 2800568-2800568 on Build GRCh38
rs755186050 Chr.12: 2800550-2800550 on Build GRCh38
rs755968529 Chr.12: 2800614-2800614 on Build GRCh38
rs922306492 Chr.12: 2800488-2800488 on Build GRCh38
rs933658225 Chr.12: 2800339-2800339 on Build GRCh38
rs762538300 Chr.12: 2800371-2800371 on Build GRCh38
rs948184371 Chr.12: 2800668-2800668 on Build GRCh38
rs144966634 Chr.12: 2800427-2800427 on Build GRCh38
rs766468008 Chr.12: 2800591-2800592 on Build GRCh38
rs906586483 Chr.12: 2800713-2800713 on Build GRCh38
rs377098730 Chr.12: 2800702-2800702 on Build GRCh38
rs1030994725 Chr.12: 2800733-2800733 on Build GRCh38
rs375816184 Chr.12: 2800541-2800541 on Build GRCh38
rs770400725 Chr.12: 2800470-2800470 on Build GRCh38
rs759231013 Chr.12: 2800375-2800375 on Build GRCh38
rs539277079 Chr.12: 2800561-2800561 on Build GRCh38
rs1045155574 Chr.12: 2800681-2800681 on Build GRCh38
rs746107167 Chr.12: 2800667-2800667 on Build GRCh38
rs752385363 Chr.12: 2800584-2800584 on Build GRCh38
rs758569908 Chr.12: 2800425-2800425 on Build GRCh38
rs762218575 Chr.12: 2800392-2800392 on Build GRCh38
rs749304428 Chr.12: 2800651-2800651 on Build GRCh38
rs56081209 Chr.12: 2800769-2800769 on Build GRCh38
rs1020219262 Chr.12: 2800526-2800526 on Build GRCh38
rs573131910 Chr.12: 2800759-2800759 on Build GRCh38
rs1000218110 Chr.12: 2800551-2800551 on Build GRCh38
rs747981676 Chr.12: 2800445-2800445 on Build GRCh38
rs566903026 Chr.12: 2800368-2800368 on Build GRCh38
rs764770209 Chr.12: 2800567-2800567 on Build GRCh38
rs762817746 Chr.12: 2800412-2800412 on Build GRCh38
rs772532017 Chr.12: 2800344-2800344 on Build GRCh38
rs773902439 Chr.12: 2800474-2800474 on Build GRCh38
rs764423146 Chr.12: 2800381-2800381 on Build GRCh38
rs940930043 Chr.12: 2800636-2800636 on Build GRCh38
rs769561303 Chr.12: 2800447-2800447 on Build GRCh38
rs1003558660 Chr.12: 2800722-2800722 on Build GRCh38
rs899329427 Chr.12: 2800656-2800656 on Build GRCh38
rs765776735 Chr.12: 2800400-2800400 on Build GRCh38
rs776791993 Chr.12: 2800479-2800479 on Build GRCh38
rs955291995 Chr.12: 2800655-2800655 on Build GRCh38
rs750970764 Chr.12: 2800518-2800518 on Build GRCh38
rs372496355 Chr.12: 2800382-2800382 on Build GRCh38
rs750364400 Chr.12: 2800375-2800376 on Build GRCh38
rs751615524 Chr.12: 2800618-2800619 on Build GRCh38
rs367827487 Chr.12: 2800684-2800684 on Build GRCh38
rs941114838 Chr.12: 2800707-2800707 on Build GRCh38
rs1037871427 Chr.12: 2800642-2800642 on Build GRCh38
rs372262068 Chr.12: 2800623-2800623 on Build GRCh38
rs771776017 Chr.12: 2800478-2800478 on Build GRCh38
rs751745589 Chr.12: 2800547-2800547 on Build GRCh38
rs899312644 Chr.12: 2800393-2800393 on Build GRCh38
rs780835389 Chr.12: 2800434-2800434 on Build GRCh38
rs184543678 Chr.12: 2800544-2800544 on Build GRCh38
rs933691817 Chr.12: 2800572-2800572 on Build GRCh38
rs759178167 Chr.12: 2800477-2800477 on Build GRCh38
rs1037905007 Chr.12: 2800374-2800374 on Build GRCh38
rs754132895 Chr.12: 2800331-2800331 on Build GRCh38
rs368886036 Chr.12: 2800373-2800373 on Build GRCh38
rs1032845664 Chr.12: 2800553-2800553 on Build GRCh38
rs751800526 Chr.12: 2800433-2800433 on Build GRCh38
rs922512449 Chr.12: 2800629-2800629 on Build GRCh38
rs922325088 Chr.12: 2800326-2800326 on Build GRCh38
rs114029399 Chr.12: 2800342-2800342 on Build GRCh38
rs762248029 Chr.12: 2800485-2800485 on Build GRCh38
rs144630885 Chr.12: 2800468-2800468 on Build GRCh38
rs1014212097 Chr.12: 2800497-2800497 on Build GRCh38
rs765609799 Chr.12: 2800492-2800492 on Build GRCh38
rs982827966 Chr.12: 2800576-2800576 on Build GRCh38
rs757630947 Chr.12: 2800594-2800594 on Build GRCh38
rs150316795 Chr.12: 2800406-2800406 on Build GRCh38
rs908165102 Chr.12: 2800631-2800631 on Build GRCh38

View Full Product Details