Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2888357-2888858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHNO1
Entrez Gene ID
83695
Gene Name
RAD9-HUS1-RAD1 interacting nuclear orphan 1
Gene Aliases
C12orf32, HKMT1188, RHINO
UniGene
Hs.198853
Gene Chromosome Location
Chr.12: 2876258-2889525 on Build GRCh38
Transcripts
NM_001252499.2 NM_001252500.2 NM_001257097.1 NM_001257098.1 NR_027365.2 NR_046432.1 NR_046433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955016 c.695G>A p.S232N Chr.12: 2888437-2888437 on Build GRCh38
4848326 c.631G>C p.V211L Chr.12: 2888373-2888373 on Build GRCh38
3671049 c.646C>T p.R216* Chr.12: 2888388-2888388 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776598671 Chr.12: 2888476-2888476 on Build GRCh38
rs755472010 Chr.12: 2888728-2888728 on Build GRCh38
rs565175289 Chr.12: 2888729-2888729 on Build GRCh38
rs778820816 Chr.12: 2888423-2888423 on Build GRCh38
rs1051957503 Chr.12: 2888717-2888717 on Build GRCh38
rs757331711 Chr.12: 2888413-2888413 on Build GRCh38
rs933490746 Chr.12: 2888713-2888713 on Build GRCh38
rs756533975 Chr.12: 2888388-2888388 on Build GRCh38
rs1004870243 Chr.12: 2888436-2888436 on Build GRCh38
rs145725447 Chr.12: 2888737-2888737 on Build GRCh38
rs912446182 Chr.12: 2888690-2888690 on Build GRCh38
rs751267067 Chr.12: 2888495-2888495 on Build GRCh38
rs545276039 Chr.12: 2888721-2888721 on Build GRCh38
rs929797911 Chr.12: 2888653-2888653 on Build GRCh38
rs780428485 Chr.12: 2888466-2888466 on Build GRCh38
rs916250743 Chr.12: 2888523-2888523 on Build GRCh38
rs772536125 Chr.12: 2888488-2888488 on Build GRCh38
rs779936669 Chr.12: 2888391-2888391 on Build GRCh38
rs936883835 Chr.12: 2888685-2888685 on Build GRCh38
rs534876158 Chr.12: 2888465-2888465 on Build GRCh38
rs201886525 Chr.12: 2888608-2888608 on Build GRCh38
rs201547442 Chr.12: 2888385-2888385 on Build GRCh38
rs1034282155 Chr.12: 2888804-2888804 on Build GRCh38
rs761009314 Chr.12: 2888511-2888511 on Build GRCh38
rs992506261 Chr.12: 2888630-2888630 on Build GRCh38
rs1046917591 Chr.12: 2888663-2888663 on Build GRCh38
rs375116699 Chr.12: 2888711-2888711 on Build GRCh38
rs749811458 Chr.12: 2888397-2888397 on Build GRCh38
rs759947710 Chr.12: 2888814-2888814 on Build GRCh38
rs768003568 Chr.12: 2888469-2888469 on Build GRCh38
rs768621136 Chr.12: 2888475-2888475 on Build GRCh38
rs769790532 Chr.12: 2888486-2888486 on Build GRCh38
rs749256665 Chr.12: 2888426-2888426 on Build GRCh38
rs11062399 Chr.12: 2888773-2888773 on Build GRCh38
rs1013191564 Chr.12: 2888782-2888782 on Build GRCh38
rs899070919 Chr.12: 2888553-2888553 on Build GRCh38
rs751242303 Chr.12: 2888444-2888444 on Build GRCh38
rs1019976199 Chr.12: 2888671-2888671 on Build GRCh38
rs186352408 Chr.12: 2888570-2888570 on Build GRCh38
rs969140519 Chr.12: 2888587-2888587 on Build GRCh38
rs766680953 Chr.12: 2888631-2888631 on Build GRCh38
rs376902913 Chr.12: 2888490-2888490 on Build GRCh38
rs376987704 Chr.12: 2888392-2888392 on Build GRCh38
rs190965459 Chr.12: 2888613-2888613 on Build GRCh38
rs773928239 Chr.12: 2888614-2888614 on Build GRCh38
rs919712914 Chr.12: 2888652-2888652 on Build GRCh38
rs371145409 Chr.12: 2888708-2888708 on Build GRCh38
rs386759584 Chr.12: 2888773-2888774 on Build GRCh38
rs11062400 Chr.12: 2888774-2888774 on Build GRCh38
rs370452101 Chr.12: 2888533-2888533 on Build GRCh38
rs967124517 Chr.12: 2888677-2888677 on Build GRCh38
rs745984392 Chr.12: 2888434-2888434 on Build GRCh38
rs761613588 Chr.12: 2888478-2888478 on Build GRCh38
rs903306309 Chr.12: 2888731-2888731 on Build GRCh38
rs181653990 Chr.12: 2888524-2888524 on Build GRCh38
rs1001468904 Chr.12: 2888557-2888557 on Build GRCh38
rs978395473 Chr.12: 2888678-2888678 on Build GRCh38
rs925654958 Chr.12: 2888683-2888683 on Build GRCh38
rs148952518 Chr.12: 2888719-2888719 on Build GRCh38
rs895748439 Chr.12: 2888807-2888807 on Build GRCh38
rs1043516717 Chr.12: 2888730-2888730 on Build GRCh38

View Full Product Details