Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4367840-4368378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF23
Entrez Gene ID
8074
Gene Name
fibroblast growth factor 23
Gene Aliases
ADHR, FGFN, HPDR2, HYPF, PHPTC
UniGene
Hs.287370
Gene Chromosome Location
Chr.12: 4368227-4379728 on Build GRCh38
Transcripts
NM_020638.2

SNP Detail

SNP ID Chromosome Location
rs999625126 Chr.12: 4368124-4368124 on Build GRCh38
rs562751894 Chr.12: 4368131-4368131 on Build GRCh38
rs916638628 Chr.12: 4368009-4368009 on Build GRCh38
rs796624261 Chr.12: 4368188-4368188 on Build GRCh38
rs992308497 Chr.12: 4368001-4368001 on Build GRCh38
rs960845394 Chr.12: 4367971-4367971 on Build GRCh38
rs911032187 Chr.12: 4367963-4367963 on Build GRCh38
rs13312804 Chr.12: 4368089-4368089 on Build GRCh38
rs984893382 Chr.12: 4368217-4368217 on Build GRCh38
rs893043603 Chr.12: 4368007-4368007 on Build GRCh38
rs968666117 Chr.12: 4367904-4367904 on Build GRCh38
rs1043906493 Chr.12: 4368122-4368122 on Build GRCh38
rs760272001 Chr.12: 4368077-4368077 on Build GRCh38
rs889565055 Chr.12: 4367878-4367878 on Build GRCh38
rs973037164 Chr.12: 4368142-4368142 on Build GRCh38
rs577592559 Chr.12: 4367938-4367938 on Build GRCh38
rs137904258 Chr.12: 4368184-4368184 on Build GRCh38
rs972806956 Chr.12: 4367868-4367868 on Build GRCh38
rs1006538927 Chr.12: 4367885-4367885 on Build GRCh38
rs955041844 Chr.12: 4368201-4368201 on Build GRCh38
rs772588965 Chr.12: 4367947-4367947 on Build GRCh38
rs959498141 Chr.12: 4368101-4368101 on Build GRCh38
rs909220106 Chr.12: 4368049-4368049 on Build GRCh38
rs527720791 Chr.12: 4368297-4368297 on Build GRCh38
rs1017141958 Chr.12: 4368128-4368128 on Build GRCh38
rs1025701430 Chr.12: 4368173-4368174 on Build GRCh38
rs945927013 Chr.12: 4368339-4368339 on Build GRCh38
rs918485528 Chr.12: 4368345-4368345 on Build GRCh38
rs370918108 Chr.12: 4368185-4368188 on Build GRCh38
rs940728426 Chr.12: 4368055-4368055 on Build GRCh38
rs13312803 Chr.12: 4368123-4368123 on Build GRCh38
rs1050843267 Chr.12: 4367948-4367948 on Build GRCh38
rs780835137 Chr.12: 4368045-4368045 on Build GRCh38
rs779517976 Chr.12: 4368248-4368248 on Build GRCh38
rs560232851 Chr.12: 4368071-4368071 on Build GRCh38
rs889503988 Chr.12: 4368096-4368096 on Build GRCh38
rs572475257 Chr.12: 4368129-4368129 on Build GRCh38
rs547468001 Chr.12: 4368320-4368320 on Build GRCh38
rs145249503 Chr.12: 4368036-4368036 on Build GRCh38
rs933713599 Chr.12: 4367894-4367894 on Build GRCh38
rs1050935576 Chr.12: 4368092-4368092 on Build GRCh38
rs1021968433 Chr.12: 4367899-4367899 on Build GRCh38
rs1031701898 Chr.12: 4367969-4367969 on Build GRCh38
rs527640506 Chr.12: 4368188-4368189 on Build GRCh38
rs13312802 Chr.12: 4368203-4368203 on Build GRCh38
rs1042770792 Chr.12: 4368045-4368050 on Build GRCh38
rs541609260 Chr.12: 4368184-4368185 on Build GRCh38
rs942468250 Chr.12: 4368274-4368274 on Build GRCh38
rs773935723 Chr.12: 4368046-4368046 on Build GRCh38
rs918527833 Chr.12: 4368088-4368088 on Build GRCh38
rs969407606 Chr.12: 4368211-4368211 on Build GRCh38
rs558079364 Chr.12: 4368281-4368281 on Build GRCh38
rs771811777 Chr.12: 4367912-4367912 on Build GRCh38
rs1016797631 Chr.12: 4368228-4368228 on Build GRCh38
rs13312805 Chr.12: 4368042-4368042 on Build GRCh38
rs999778516 Chr.12: 4367958-4367958 on Build GRCh38
rs914441177 Chr.12: 4368338-4368338 on Build GRCh38
rs969459979 Chr.12: 4368035-4368035 on Build GRCh38
rs972497814 Chr.12: 4368311-4368311 on Build GRCh38
rs1006646633 Chr.12: 4368117-4368117 on Build GRCh38
rs942510769 Chr.12: 4367992-4367992 on Build GRCh38
rs1054337517 Chr.12: 4367998-4367998 on Build GRCh38

View Full Product Details