Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4487518-4488057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
UniGene
Hs.302977
Gene Chromosome Location
Chr.12: 4487730-4538508 on Build GRCh38
Transcripts
NM_001304811.1 NM_020374.3 XM_005253716.1

SNP Detail

SNP ID Chromosome Location
rs924646954 Chr.12: 4487787-4487787 on Build GRCh38
rs974423163 Chr.12: 4487828-4487828 on Build GRCh38
rs780261669 Chr.12: 4487664-4487664 on Build GRCh38
rs755971316 Chr.12: 4487899-4487899 on Build GRCh38
rs1008686624 Chr.12: 4487706-4487706 on Build GRCh38
rs71579224 Chr.12: 4487616-4487616 on Build GRCh38
rs796109282 Chr.12: 4487700-4487700 on Build GRCh38
rs71579222 Chr.12: 4487760-4487760 on Build GRCh38
rs189619256 Chr.12: 4487916-4487916 on Build GRCh38
rs138975476 Chr.12: 4487619-4487620 on Build GRCh38
rs540311809 Chr.12: 4487882-4487882 on Build GRCh38
rs962752572 Chr.12: 4487715-4487715 on Build GRCh38
rs934742052 Chr.12: 4487802-4487802 on Build GRCh38
rs891926099 Chr.12: 4487677-4487677 on Build GRCh38
rs975055128 Chr.12: 4487759-4487759 on Build GRCh38
rs913518879 Chr.12: 4488000-4488000 on Build GRCh38
rs369326900 Chr.12: 4487724-4487724 on Build GRCh38
rs915135933 Chr.12: 4487759-4487760 on Build GRCh38
rs948885533 Chr.12: 4487906-4487906 on Build GRCh38
rs71579223 Chr.12: 4487623-4487623 on Build GRCh38
rs138510649 Chr.12: 4487883-4487883 on Build GRCh38
rs749480129 Chr.12: 4487669-4487669 on Build GRCh38
rs778989666 Chr.12: 4487685-4487685 on Build GRCh38
rs780435026 Chr.12: 4487761-4487761 on Build GRCh38
rs139647281 Chr.12: 4488032-4488032 on Build GRCh38
rs995141600 Chr.12: 4487734-4487734 on Build GRCh38
rs575628496 Chr.12: 4487625-4487625 on Build GRCh38
rs868627056 Chr.12: 4487820-4487820 on Build GRCh38
rs1001522949 Chr.12: 4487686-4487686 on Build GRCh38
rs1026241018 Chr.12: 4487745-4487745 on Build GRCh38
rs773202732 Chr.12: 4487921-4487921 on Build GRCh38
rs559824807 Chr.12: 4487893-4487893 on Build GRCh38
rs1018786887 Chr.12: 4487738-4487738 on Build GRCh38
rs748278926 Chr.12: 4487695-4487695 on Build GRCh38
rs899374404 Chr.12: 4487988-4487988 on Build GRCh38
rs574044770 Chr.12: 4487605-4487605 on Build GRCh38
rs7134953 Chr.12: 4487678-4487678 on Build GRCh38
rs911650741 Chr.12: 4487581-4487581 on Build GRCh38
rs930898295 Chr.12: 4487993-4487993 on Build GRCh38
rs7793 Chr.12: 4487812-4487812 on Build GRCh38
rs564283277 Chr.12: 4487759-4487759 on Build GRCh38
rs147831244 Chr.12: 4487572-4487572 on Build GRCh38
rs964564015 Chr.12: 4487757-4487757 on Build GRCh38
rs868797155 Chr.12: 4487639-4487639 on Build GRCh38
rs181876363 Chr.12: 4487997-4487997 on Build GRCh38
rs1045786388 Chr.12: 4487679-4487679 on Build GRCh38
rs950547845 Chr.12: 4487543-4487543 on Build GRCh38

View Full Product Details