Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4489130-4489641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
UniGene
Hs.302977
Gene Chromosome Location
Chr.12: 4487730-4538508 on Build GRCh38
Transcripts
NM_001304811.1 NM_020374.3 XM_005253716.1

SNP Detail

SNP ID Chromosome Location
rs924982671 Chr.12: 4489571-4489571 on Build GRCh38
rs775338263 Chr.12: 4489383-4489383 on Build GRCh38
rs974635909 Chr.12: 4489220-4489220 on Build GRCh38
rs1045416205 Chr.12: 4489329-4489329 on Build GRCh38
rs748738860 Chr.12: 4489481-4489481 on Build GRCh38
rs191032379 Chr.12: 4489403-4489403 on Build GRCh38
rs1010035600 Chr.12: 4489408-4489408 on Build GRCh38
rs1013438660 Chr.12: 4489273-4489273 on Build GRCh38
rs538951788 Chr.12: 4489474-4489474 on Build GRCh38
rs186328265 Chr.12: 4489171-4489171 on Build GRCh38
rs1041442204 Chr.12: 4489480-4489480 on Build GRCh38
rs866531918 Chr.12: 4489301-4489301 on Build GRCh38
rs1043414840 Chr.12: 4489212-4489212 on Build GRCh38
rs987874285 Chr.12: 4489607-4489607 on Build GRCh38
rs74061917 Chr.12: 4489254-4489254 on Build GRCh38
rs11548151 Chr.12: 4489577-4489577 on Build GRCh38
rs185841049 Chr.12: 4489401-4489401 on Build GRCh38
rs907003233 Chr.12: 4489487-4489487 on Build GRCh38
rs565542837 Chr.12: 4489427-4489427 on Build GRCh38
rs115746793 Chr.12: 4489371-4489371 on Build GRCh38
rs983580545 Chr.12: 4489163-4489163 on Build GRCh38
rs924984927 Chr.12: 4489238-4489238 on Build GRCh38
rs538662422 Chr.12: 4489187-4489187 on Build GRCh38
rs922152704 Chr.12: 4489437-4489437 on Build GRCh38
rs1044791 Chr.12: 4489193-4489193 on Build GRCh38
rs976287893 Chr.12: 4489410-4489410 on Build GRCh38
rs577880857 Chr.12: 4489198-4489198 on Build GRCh38
rs796862282 Chr.12: 4489401-4489403 on Build GRCh38
rs982825198 Chr.12: 4489255-4489255 on Build GRCh38
rs976761734 Chr.12: 4489188-4489188 on Build GRCh38
rs953656209 Chr.12: 4489473-4489473 on Build GRCh38
rs114194187 Chr.12: 4489183-4489183 on Build GRCh38
rs908052692 Chr.12: 4489165-4489165 on Build GRCh38
rs183169384 Chr.12: 4489445-4489445 on Build GRCh38
rs576594045 Chr.12: 4489585-4489585 on Build GRCh38
rs912686215 Chr.12: 4489257-4489257 on Build GRCh38
rs979472870 Chr.12: 4489484-4489484 on Build GRCh38
rs750905456 Chr.12: 4489572-4489572 on Build GRCh38
rs1002935944 Chr.12: 4489532-4489532 on Build GRCh38
rs995750569 Chr.12: 4489609-4489609 on Build GRCh38
rs931887957 Chr.12: 4489391-4489391 on Build GRCh38
rs558323807 Chr.12: 4489379-4489379 on Build GRCh38
rs548139550 Chr.12: 4489488-4489488 on Build GRCh38
rs931610516 Chr.12: 4489202-4489202 on Build GRCh38
rs71579220 Chr.12: 4489243-4489243 on Build GRCh38
rs944016576 Chr.12: 4489283-4489283 on Build GRCh38
rs905123813 Chr.12: 4489366-4489366 on Build GRCh38
rs529786133 Chr.12: 4489399-4489399 on Build GRCh38
rs1015100430 Chr.12: 4489405-4489405 on Build GRCh38
rs959012146 Chr.12: 4489195-4489195 on Build GRCh38

View Full Product Details