Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6694395-6694887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIANP
Entrez Gene ID
196500
Gene Name
PILR alpha associated neural protein
Gene Aliases
C12orf53, LEDA1, PANP, leda-1
UniGene
Hs.44067
Gene Chromosome Location
Chr.12: 6689726-6700843 on Build GRCh38
Transcripts
NM_001244014.1 NM_001244015.1 NM_153685.3 XM_011520926.2

SNP Detail

SNP ID Chromosome Location
rs941892154 Chr.12: 6694715-6694715 on Build GRCh38
rs887447922 Chr.12: 6694570-6694570 on Build GRCh38
rs917646539 Chr.12: 6694527-6694527 on Build GRCh38
rs11064320 Chr.12: 6694730-6694730 on Build GRCh38
rs958231441 Chr.12: 6694813-6694813 on Build GRCh38
rs533191763 Chr.12: 6694810-6694810 on Build GRCh38
rs61742689 Chr.12: 6694631-6694631 on Build GRCh38
rs4764476 Chr.12: 6694515-6694515 on Build GRCh38
rs781010334 Chr.12: 6694602-6694602 on Build GRCh38
rs750023568 Chr.12: 6694450-6694450 on Build GRCh38
rs977484086 Chr.12: 6694748-6694748 on Build GRCh38
rs1006095599 Chr.12: 6694571-6694571 on Build GRCh38
rs1044707746 Chr.12: 6694552-6694552 on Build GRCh38
rs138966637 Chr.12: 6694691-6694691 on Build GRCh38
rs997221591 Chr.12: 6694539-6694539 on Build GRCh38
rs992931572 Chr.12: 6694780-6694780 on Build GRCh38
rs953467064 Chr.12: 6694481-6694481 on Build GRCh38
rs548396384 Chr.12: 6694583-6694585 on Build GRCh38
rs575210256 Chr.12: 6694449-6694449 on Build GRCh38
rs916092596 Chr.12: 6694781-6694781 on Build GRCh38
rs7953855 Chr.12: 6694690-6694690 on Build GRCh38
rs766070423 Chr.12: 6694490-6694490 on Build GRCh38
rs1036055321 Chr.12: 6694843-6694843 on Build GRCh38
rs953121090 Chr.12: 6694555-6694555 on Build GRCh38
rs550876860 Chr.12: 6694759-6694759 on Build GRCh38
rs970964496 Chr.12: 6694844-6694844 on Build GRCh38
rs925477617 Chr.12: 6694569-6694569 on Build GRCh38
rs556300137 Chr.12: 6694486-6694486 on Build GRCh38
rs375201088 Chr.12: 6694566-6694566 on Build GRCh38
rs373607165 Chr.12: 6694729-6694729 on Build GRCh38
rs546072891 Chr.12: 6694632-6694632 on Build GRCh38
rs770156798 Chr.12: 6694778-6694778 on Build GRCh38
rs1046407460 Chr.12: 6694614-6694614 on Build GRCh38
rs905307675 Chr.12: 6694443-6694443 on Build GRCh38
rs541298355 Chr.12: 6694840-6694840 on Build GRCh38
rs1036683062 Chr.12: 6694572-6694572 on Build GRCh38
rs947824686 Chr.12: 6694533-6694533 on Build GRCh38
rs182499849 Chr.12: 6694790-6694790 on Build GRCh38
rs914181159 Chr.12: 6694585-6694585 on Build GRCh38
rs960303771 Chr.12: 6694776-6694776 on Build GRCh38
rs988405777 Chr.12: 6694573-6694573 on Build GRCh38
rs1046463784 Chr.12: 6694438-6694438 on Build GRCh38
rs1030411324 Chr.12: 6694462-6694462 on Build GRCh38
rs1007325652 Chr.12: 6694787-6694787 on Build GRCh38
rs61745779 Chr.12: 6694461-6694461 on Build GRCh38
rs754821475 Chr.12: 6694595-6694595 on Build GRCh38
rs924928864 Chr.12: 6694520-6694520 on Build GRCh38
rs187530338 Chr.12: 6694827-6694827 on Build GRCh38
rs947772801 Chr.12: 6694789-6694789 on Build GRCh38
rs1021589903 Chr.12: 6694720-6694720 on Build GRCh38
rs751129271 Chr.12: 6694523-6694523 on Build GRCh38
rs564383396 Chr.12: 6694516-6694516 on Build GRCh38
rs1030190337 Chr.12: 6694621-6694621 on Build GRCh38
rs1057260878 Chr.12: 6694526-6694526 on Build GRCh38
rs938113996 Chr.12: 6694685-6694685 on Build GRCh38
rs530397361 Chr.12: 6694749-6694749 on Build GRCh38

View Full Product Details