Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6696364-6696862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIANP
Entrez Gene ID
196500
Gene Name
PILR alpha associated neural protein
Gene Aliases
C12orf53, LEDA1, PANP, leda-1
UniGene
Hs.44067
Gene Chromosome Location
Chr.12: 6689726-6700843 on Build GRCh38
Transcripts
NM_001244014.1 NM_001244015.1 NM_153685.3 XM_011520926.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358019 c.567C>G p.I189M Chr.12: 6696481-6696481 on Build GRCh38
5143441 c.536A>G p.Q179R Chr.12: 6696512-6696512 on Build GRCh38
3704374 c.580G>A p.A194T Chr.12: 6696468-6696468 on Build GRCh38
272408 c.532C>T p.P178S Chr.12: 6696516-6696516 on Build GRCh38
1363838 c.578T>C p.V193A Chr.12: 6696470-6696470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769647374 Chr.12: 6696493-6696493 on Build GRCh38
rs918600267 Chr.12: 6696786-6696786 on Build GRCh38
rs267603641 Chr.12: 6696524-6696524 on Build GRCh38
rs202197192 Chr.12: 6696560-6696560 on Build GRCh38
rs773799330 Chr.12: 6696645-6696645 on Build GRCh38
rs567609807 Chr.12: 6696798-6696798 on Build GRCh38
rs897382091 Chr.12: 6696409-6696409 on Build GRCh38
rs557813162 Chr.12: 6696794-6696794 on Build GRCh38
rs927383649 Chr.12: 6696755-6696755 on Build GRCh38
rs770756267 Chr.12: 6696532-6696532 on Build GRCh38
rs563314280 Chr.12: 6696535-6696535 on Build GRCh38
rs773992375 Chr.12: 6696533-6696533 on Build GRCh38
rs955926720 Chr.12: 6696671-6696671 on Build GRCh38
rs999843968 Chr.12: 6696639-6696639 on Build GRCh38
rs749057355 Chr.12: 6696506-6696506 on Build GRCh38
rs530959923 Chr.12: 6696682-6696682 on Build GRCh38
rs759170974 Chr.12: 6696537-6696537 on Build GRCh38
rs148472703 Chr.12: 6696780-6696780 on Build GRCh38
rs771554413 Chr.12: 6696549-6696549 on Build GRCh38
rs988693071 Chr.12: 6696687-6696687 on Build GRCh38
rs539796037 Chr.12: 6696609-6696609 on Build GRCh38
rs1022147694 Chr.12: 6696700-6696701 on Build GRCh38
rs893249065 Chr.12: 6696688-6696688 on Build GRCh38
rs762242748 Chr.12: 6696430-6696430 on Build GRCh38
rs930212379 Chr.12: 6696583-6696583 on Build GRCh38
rs535697991 Chr.12: 6696678-6696678 on Build GRCh38
rs774869296 Chr.12: 6696553-6696553 on Build GRCh38
rs767005445 Chr.12: 6696423-6696423 on Build GRCh38
rs144459517 Chr.12: 6696631-6696631 on Build GRCh38
rs781144093 Chr.12: 6696481-6696481 on Build GRCh38
rs370638218 Chr.12: 6696406-6696406 on Build GRCh38
rs1051057915 Chr.12: 6696598-6696598 on Build GRCh38
rs1051232179 Chr.12: 6696555-6696555 on Build GRCh38
rs990527843 Chr.12: 6696475-6696475 on Build GRCh38
rs766906348 Chr.12: 6696572-6696572 on Build GRCh38
rs12370203 Chr.12: 6696733-6696733 on Build GRCh38
rs752277941 Chr.12: 6696833-6696833 on Build GRCh38
rs891301095 Chr.12: 6696557-6696557 on Build GRCh38
rs971198765 Chr.12: 6696745-6696745 on Build GRCh38
rs927641209 Chr.12: 6696466-6696466 on Build GRCh38
rs750849345 Chr.12: 6696435-6696435 on Build GRCh38
rs116893637 Chr.12: 6696559-6696559 on Build GRCh38
rs966566260 Chr.12: 6696738-6696738 on Build GRCh38
rs1024217115 Chr.12: 6696743-6696743 on Build GRCh38
rs755071812 Chr.12: 6696477-6696477 on Build GRCh38
rs780391704 Chr.12: 6696471-6696471 on Build GRCh38
rs1051344393 Chr.12: 6696830-6696830 on Build GRCh38
rs891122341 Chr.12: 6696602-6696603 on Build GRCh38
rs188836221 Chr.12: 6696783-6696783 on Build GRCh38
rs1011126744 Chr.12: 6696689-6696689 on Build GRCh38
rs777492515 Chr.12: 6696496-6696496 on Build GRCh38
rs566813611 Chr.12: 6696644-6696644 on Build GRCh38
rs184456612 Chr.12: 6696793-6696793 on Build GRCh38
rs765633138 Chr.12: 6696431-6696431 on Build GRCh38
rs980373580 Chr.12: 6696750-6696750 on Build GRCh38
rs770299477 Chr.12: 6696571-6696571 on Build GRCh38
rs758645040 Chr.12: 6696454-6696454 on Build GRCh38
rs376198777 Chr.12: 6696478-6696478 on Build GRCh38
rs562628305 Chr.12: 6696425-6696425 on Build GRCh38
rs372943783 Chr.12: 6696472-6696472 on Build GRCh38
rs571926403 Chr.12: 6696695-6696695 on Build GRCh38
rs774043070 Chr.12: 6696397-6696397 on Build GRCh38
rs540218238 Chr.12: 6696820-6696820 on Build GRCh38
rs149239038 Chr.12: 6696483-6696483 on Build GRCh38
rs751007359 Chr.12: 6696570-6696570 on Build GRCh38
rs193224284 Chr.12: 6696784-6696784 on Build GRCh38

View Full Product Details