Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6814679-6815178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD4
Entrez Gene ID
920
Gene Name
CD4 molecule
Gene Aliases
CD4mut
UniGene
Hs.631659
Gene Chromosome Location
Chr.12: 6789472-6820810 on Build GRCh38
Transcripts
NM_000616.4 NM_001195014.2 NM_001195015.2 NM_001195016.2 NM_001195017.2 XM_017020228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3872317 c.522G>A p.L174L Chr.12: 6814907-6814907 on Build GRCh38
4804728 c.390C>T p.D130D Chr.12: 6814775-6814775 on Build GRCh38
6137845 c.511G>T p.V171L Chr.12: 6814896-6814896 on Build GRCh38
2099338 c.378T>C p.T126T Chr.12: 6814763-6814763 on Build GRCh38
3872318 c.573G>A p.K191K Chr.12: 6814958-6814958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782730611 Chr.12: 6814978-6814978 on Build GRCh38
rs782400591 Chr.12: 6814864-6814864 on Build GRCh38
rs781937995 Chr.12: 6814876-6814877 on Build GRCh38
rs757275036 Chr.12: 6815117-6815117 on Build GRCh38
rs782245179 Chr.12: 6814897-6814897 on Build GRCh38
rs200097903 Chr.12: 6815025-6815025 on Build GRCh38
rs782100522 Chr.12: 6814764-6814764 on Build GRCh38
rs782190107 Chr.12: 6814998-6814998 on Build GRCh38
rs201435903 Chr.12: 6814879-6814879 on Build GRCh38
rs782664798 Chr.12: 6814820-6814820 on Build GRCh38
rs782503327 Chr.12: 6814819-6814819 on Build GRCh38
rs199808793 Chr.12: 6815008-6815008 on Build GRCh38
rs782301334 Chr.12: 6814938-6814938 on Build GRCh38
rs782082944 Chr.12: 6815014-6815014 on Build GRCh38
rs1038620467 Chr.12: 6815007-6815007 on Build GRCh38
rs781853897 Chr.12: 6815112-6815112 on Build GRCh38
rs781786552 Chr.12: 6814803-6814803 on Build GRCh38
rs993168756 Chr.12: 6814816-6814816 on Build GRCh38
rs113196432 Chr.12: 6814721-6814721 on Build GRCh38
rs183019961 Chr.12: 6815013-6815013 on Build GRCh38
rs782353658 Chr.12: 6814947-6814947 on Build GRCh38
rs782344955 Chr.12: 6814838-6814838 on Build GRCh38
rs895780652 Chr.12: 6814714-6814714 on Build GRCh38
rs202028153 Chr.12: 6814824-6814824 on Build GRCh38
rs925753347 Chr.12: 6814935-6814935 on Build GRCh38
rs200102296 Chr.12: 6814713-6814713 on Build GRCh38
rs201315085 Chr.12: 6814941-6814941 on Build GRCh38
rs202208698 Chr.12: 6814791-6814791 on Build GRCh38
rs782241392 Chr.12: 6814826-6814826 on Build GRCh38
rs200162977 Chr.12: 6814896-6814896 on Build GRCh38
rs1017491203 Chr.12: 6814781-6814781 on Build GRCh38
rs781909754 Chr.12: 6814972-6814972 on Build GRCh38
rs782349263 Chr.12: 6814876-6814876 on Build GRCh38
rs781982696 Chr.12: 6814840-6814840 on Build GRCh38
rs782301761 Chr.12: 6815011-6815011 on Build GRCh38
rs28917504 Chr.12: 6814956-6814956 on Build GRCh38
rs200491526 Chr.12: 6814874-6814874 on Build GRCh38
rs201323005 Chr.12: 6814828-6814828 on Build GRCh38
rs201346195 Chr.12: 6814742-6814742 on Build GRCh38
rs781925104 Chr.12: 6814881-6814881 on Build GRCh38
rs782390734 Chr.12: 6814739-6814739 on Build GRCh38
rs377592890 Chr.12: 6814967-6814967 on Build GRCh38
rs782762639 Chr.12: 6815134-6815134 on Build GRCh38
rs994332105 Chr.12: 6815084-6815084 on Build GRCh38
rs782497073 Chr.12: 6814980-6814980 on Build GRCh38
rs140406047 Chr.12: 6814983-6814983 on Build GRCh38
rs782318207 Chr.12: 6815091-6815091 on Build GRCh38
rs200844936 Chr.12: 6814784-6814784 on Build GRCh38
rs200733746 Chr.12: 6814814-6814814 on Build GRCh38
rs782775415 Chr.12: 6814769-6814769 on Build GRCh38
rs782702450 Chr.12: 6814749-6814749 on Build GRCh38
rs267603648 Chr.12: 6814883-6814883 on Build GRCh38
rs143646863 Chr.12: 6814954-6814954 on Build GRCh38
rs201247136 Chr.12: 6815029-6815029 on Build GRCh38
rs143960807 Chr.12: 6814928-6814928 on Build GRCh38
rs191620221 Chr.12: 6814875-6814875 on Build GRCh38
rs781822450 Chr.12: 6814979-6814979 on Build GRCh38
rs78962090 Chr.12: 6814723-6814723 on Build GRCh38
rs10849524 Chr.12: 6815131-6815131 on Build GRCh38
rs11575099 Chr.12: 6814943-6814943 on Build GRCh38
rs149350418 Chr.12: 6814823-6814823 on Build GRCh38
rs782677887 Chr.12: 6814889-6814889 on Build GRCh38
rs782809525 Chr.12: 6815030-6815030 on Build GRCh38
rs546840687 Chr.12: 6815154-6815154 on Build GRCh38
rs782389403 Chr.12: 6814904-6814904 on Build GRCh38
rs782798801 Chr.12: 6814796-6814796 on Build GRCh38
rs781935563 Chr.12: 6814754-6814754 on Build GRCh38
rs782737779 Chr.12: 6815022-6815022 on Build GRCh38
rs201514796 Chr.12: 6815003-6815003 on Build GRCh38
rs782199835 Chr.12: 6814832-6814832 on Build GRCh38
rs202144555 Chr.12: 6814892-6814892 on Build GRCh38
rs199799041 Chr.12: 6814975-6814975 on Build GRCh38
rs201423104 Chr.12: 6814885-6814885 on Build GRCh38
rs906131850 Chr.12: 6814726-6814726 on Build GRCh38
rs782132916 Chr.12: 6814847-6814847 on Build GRCh38
rs782704721 Chr.12: 6815100-6815100 on Build GRCh38
rs782277816 Chr.12: 6814822-6814822 on Build GRCh38
rs199912144 Chr.12: 6814745-6814745 on Build GRCh38
rs782239864 Chr.12: 6814810-6814810 on Build GRCh38
rs781867321 Chr.12: 6814777-6814777 on Build GRCh38
rs782146438 Chr.12: 6815106-6815106 on Build GRCh38
rs28917505 Chr.12: 6814982-6814982 on Build GRCh38
rs143618857 Chr.12: 6814895-6814895 on Build GRCh38
rs782400130 Chr.12: 6815006-6815006 on Build GRCh38
rs200552269 Chr.12: 6815001-6815001 on Build GRCh38
rs200408853 Chr.12: 6815012-6815012 on Build GRCh38
rs782286025 Chr.12: 6814932-6814932 on Build GRCh38

View Full Product Details