Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6936197-6936719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATN1
Entrez Gene ID
1822
Gene Name
atrophin 1
Gene Aliases
B37, D12S755E, DRPLA, HRS, NOD
UniGene
Hs.143766
Gene Chromosome Location
Chr.12: 6924463-6942321 on Build GRCh38
Transcripts
NM_001007026.1 NM_001940.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695268 c.1054C>G p.L352V Chr.12: 6936321-6936321 on Build GRCh38
6606921 c.1269_1270insC p.K426fs*98 Chr.12: 6936536-6936537 on Build GRCh38
297161 c.1224delC p.H410fs*43 Chr.12: 6936491-6936491 on Build GRCh38
4664120 c.1450_1455delCAGCAA p.Q501_Q502delQQ Chr.12: 6936717-6936722 on Build GRCh38
7313462 c.987G>T p.L329L Chr.12: 6936254-6936254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782156467 Chr.12: 6936542-6936542 on Build GRCh38
rs782498743 Chr.12: 6936617-6936617 on Build GRCh38
rs781954263 Chr.12: 6936381-6936381 on Build GRCh38
rs782524107 Chr.12: 6936417-6936417 on Build GRCh38
rs200023232 Chr.12: 6936522-6936522 on Build GRCh38
rs144583520 Chr.12: 6936354-6936354 on Build GRCh38
rs781911792 Chr.12: 6936606-6936606 on Build GRCh38
rs782776362 Chr.12: 6936289-6936289 on Build GRCh38
rs782679725 Chr.12: 6936495-6936495 on Build GRCh38
rs782609349 Chr.12: 6936334-6936334 on Build GRCh38
rs782705125 Chr.12: 6936326-6936326 on Build GRCh38
rs782043111 Chr.12: 6936556-6936556 on Build GRCh38
rs782248103 Chr.12: 6936629-6936629 on Build GRCh38
rs782420180 Chr.12: 6936635-6936635 on Build GRCh38
rs782540175 Chr.12: 6936511-6936511 on Build GRCh38
rs782731596 Chr.12: 6936561-6936561 on Build GRCh38
rs782610032 Chr.12: 6936462-6936462 on Build GRCh38
rs782304278 Chr.12: 6936277-6936277 on Build GRCh38
rs370072335 Chr.12: 6936405-6936405 on Build GRCh38
rs143104309 Chr.12: 6936659-6936659 on Build GRCh38
rs781783154 Chr.12: 6936439-6936439 on Build GRCh38
rs782324322 Chr.12: 6936506-6936506 on Build GRCh38
rs782171710 Chr.12: 6936680-6936680 on Build GRCh38
rs376473719 Chr.12: 6936432-6936432 on Build GRCh38
rs368064415 Chr.12: 6936246-6936246 on Build GRCh38
rs782379477 Chr.12: 6936356-6936356 on Build GRCh38
rs782519252 Chr.12: 6936494-6936494 on Build GRCh38
rs782392359 Chr.12: 6936514-6936514 on Build GRCh38
rs782397024 Chr.12: 6936668-6936668 on Build GRCh38
rs373005029 Chr.12: 6936664-6936664 on Build GRCh38
rs781975264 Chr.12: 6936282-6936282 on Build GRCh38
rs1058045 Chr.12: 6936284-6936284 on Build GRCh38
rs782462592 Chr.12: 6936370-6936370 on Build GRCh38
rs369685207 Chr.12: 6936480-6936480 on Build GRCh38
rs781987472 Chr.12: 6936532-6936532 on Build GRCh38
rs782657125 Chr.12: 6936266-6936266 on Build GRCh38
rs782015159 Chr.12: 6936242-6936242 on Build GRCh38
rs782336388 Chr.12: 6936257-6936257 on Build GRCh38
rs138480639 Chr.12: 6936368-6936368 on Build GRCh38
rs377411487 Chr.12: 6936695-6936695 on Build GRCh38
rs782267331 Chr.12: 6936502-6936503 on Build GRCh38
rs782769630 Chr.12: 6936601-6936601 on Build GRCh38
rs369265872 Chr.12: 6936616-6936616 on Build GRCh38
rs782739392 Chr.12: 6936692-6936692 on Build GRCh38
rs1017325093 Chr.12: 6936636-6936636 on Build GRCh38
rs782242861 Chr.12: 6936267-6936267 on Build GRCh38
rs782699382 Chr.12: 6936379-6936379 on Build GRCh38
rs201404455 Chr.12: 6936355-6936355 on Build GRCh38
rs782479130 Chr.12: 6936442-6936442 on Build GRCh38
rs372352874 Chr.12: 6936358-6936358 on Build GRCh38
rs782523332 Chr.12: 6936537-6936537 on Build GRCh38
rs782647859 Chr.12: 6936622-6936622 on Build GRCh38
rs782132501 Chr.12: 6936385-6936385 on Build GRCh38
rs782802249 Chr.12: 6936681-6936681 on Build GRCh38
rs376389021 Chr.12: 6936578-6936578 on Build GRCh38
rs782012147 Chr.12: 6936268-6936268 on Build GRCh38
rs371878798 Chr.12: 6936403-6936403 on Build GRCh38
rs782018350 Chr.12: 6936375-6936375 on Build GRCh38
rs781805970 Chr.12: 6936677-6936677 on Build GRCh38
rs781923319 Chr.12: 6936510-6936510 on Build GRCh38
rs373311096 Chr.12: 6936553-6936553 on Build GRCh38
rs782070126 Chr.12: 6936378-6936378 on Build GRCh38
rs782062831 Chr.12: 6936272-6936272 on Build GRCh38
rs202113252 Chr.12: 6936324-6936324 on Build GRCh38
rs782052698 Chr.12: 6936690-6936690 on Build GRCh38
rs782152586 Chr.12: 6936371-6936371 on Build GRCh38
rs1032661597 Chr.12: 6936687-6936687 on Build GRCh38
rs782663869 Chr.12: 6936657-6936657 on Build GRCh38
rs374756105 Chr.12: 6936291-6936291 on Build GRCh38
rs142366172 Chr.12: 6936340-6936340 on Build GRCh38
rs782165640 Chr.12: 6936243-6936243 on Build GRCh38
rs781962975 Chr.12: 6936660-6936660 on Build GRCh38
rs782187750 Chr.12: 6936658-6936658 on Build GRCh38
rs941343780 Chr.12: 6936342-6936342 on Build GRCh38
rs782170559 Chr.12: 6936292-6936292 on Build GRCh38
rs782278123 Chr.12: 6936502-6936502 on Build GRCh38
rs782797360 Chr.12: 6936302-6936302 on Build GRCh38
rs782539379 Chr.12: 6936240-6936240 on Build GRCh38
rs137926388 Chr.12: 6936254-6936254 on Build GRCh38
rs781808775 Chr.12: 6936683-6936683 on Build GRCh38
rs781930389 Chr.12: 6936555-6936555 on Build GRCh38
rs782531474 Chr.12: 6936608-6936608 on Build GRCh38
rs782597674 Chr.12: 6936613-6936613 on Build GRCh38

View Full Product Details