Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6974460-6974956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NM_001320049.1 NM_006331.7 NR_135131.1

SNP Detail

SNP ID Chromosome Location
rs782486688 Chr.12: 6974577-6974577 on Build GRCh38
rs782241980 Chr.12: 6974504-6974504 on Build GRCh38
rs989289956 Chr.12: 6974843-6974843 on Build GRCh38
rs782304736 Chr.12: 6974524-6974524 on Build GRCh38
rs782498143 Chr.12: 6974770-6974770 on Build GRCh38
rs781966629 Chr.12: 6974597-6974597 on Build GRCh38
rs782661322 Chr.12: 6974703-6974703 on Build GRCh38
rs782438469 Chr.12: 6974579-6974579 on Build GRCh38
rs367740124 Chr.12: 6974653-6974653 on Build GRCh38
rs781924299 Chr.12: 6974740-6974740 on Build GRCh38
rs782563434 Chr.12: 6974586-6974586 on Build GRCh38
rs782226901 Chr.12: 6974688-6974688 on Build GRCh38
rs374746885 Chr.12: 6974512-6974512 on Build GRCh38
rs782423280 Chr.12: 6974728-6974728 on Build GRCh38
rs781786442 Chr.12: 6974668-6974668 on Build GRCh38
rs782536404 Chr.12: 6974536-6974536 on Build GRCh38
rs782494758 Chr.12: 6974700-6974700 on Build GRCh38
rs978599596 Chr.12: 6974760-6974760 on Build GRCh38
rs782169969 Chr.12: 6974634-6974634 on Build GRCh38
rs782241520 Chr.12: 6974615-6974616 on Build GRCh38
rs782608850 Chr.12: 6974785-6974785 on Build GRCh38
rs964594510 Chr.12: 6974733-6974733 on Build GRCh38
rs782644384 Chr.12: 6974578-6974578 on Build GRCh38
rs782600333 Chr.12: 6974503-6974503 on Build GRCh38
rs370505796 Chr.12: 6974533-6974533 on Build GRCh38
rs781936465 Chr.12: 6974525-6974525 on Build GRCh38
rs782112875 Chr.12: 6974602-6974602 on Build GRCh38
rs781831826 Chr.12: 6974574-6974574 on Build GRCh38
rs782538249 Chr.12: 6974654-6974654 on Build GRCh38
rs782456048 Chr.12: 6974564-6974564 on Build GRCh38
rs993652525 Chr.12: 6974613-6974613 on Build GRCh38
rs782018212 Chr.12: 6974621-6974621 on Build GRCh38
rs781860384 Chr.12: 6974545-6974545 on Build GRCh38
rs782018986 Chr.12: 6974558-6974558 on Build GRCh38
rs782453517 Chr.12: 6974678-6974678 on Build GRCh38
rs191745916 Chr.12: 6974915-6974915 on Build GRCh38
rs782808257 Chr.12: 6974667-6974667 on Build GRCh38
rs782648414 Chr.12: 6974603-6974604 on Build GRCh38
rs782169084 Chr.12: 6974562-6974562 on Build GRCh38
rs782273726 Chr.12: 6974722-6974722 on Build GRCh38
rs782190613 Chr.12: 6974589-6974589 on Build GRCh38
rs781939868 Chr.12: 6974645-6974645 on Build GRCh38
rs782394322 Chr.12: 6974650-6974650 on Build GRCh38
rs782267364 Chr.12: 6974868-6974868 on Build GRCh38
rs377600634 Chr.12: 6974534-6974534 on Build GRCh38
rs781910054 Chr.12: 6974563-6974563 on Build GRCh38
rs374737385 Chr.12: 6974635-6974635 on Build GRCh38
rs956187350 Chr.12: 6974841-6974841 on Build GRCh38
rs782580920 Chr.12: 6974565-6974565 on Build GRCh38
rs188194099 Chr.12: 6974857-6974857 on Build GRCh38
rs782233668 Chr.12: 6974864-6974864 on Build GRCh38
rs782218371 Chr.12: 6974686-6974686 on Build GRCh38
rs782418214 Chr.12: 6974585-6974585 on Build GRCh38
rs898919076 Chr.12: 6974587-6974587 on Build GRCh38
rs781943238 Chr.12: 6974738-6974738 on Build GRCh38
rs199566199 Chr.12: 6974618-6974618 on Build GRCh38
rs781812160 Chr.12: 6974646-6974646 on Build GRCh38
rs782353464 Chr.12: 6974618-6974619 on Build GRCh38

View Full Product Details