Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7133468-7133964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268904 c.236C>T p.A79V Chr.12: 7133621-7133621 on Build GRCh38
5528782 c.369C>T p.T123T Chr.12: 7133754-7133754 on Build GRCh38
6138124 c.308A>G p.K103R Chr.12: 7133693-7133693 on Build GRCh38
2101700 c.274C>T p.R92W Chr.12: 7133659-7133659 on Build GRCh38
5528784 c.339C>T p.D113D Chr.12: 7133724-7133724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778337212 Chr.12: 7133779-7133779 on Build GRCh38
rs1046447551 Chr.12: 7133869-7133869 on Build GRCh38
rs778011123 Chr.12: 7133590-7133590 on Build GRCh38
rs778924872 Chr.12: 7133683-7133683 on Build GRCh38
rs936133979 Chr.12: 7133831-7133831 on Build GRCh38
rs763703526 Chr.12: 7133540-7133540 on Build GRCh38
rs371850025 Chr.12: 7133805-7133805 on Build GRCh38
rs1055415943 Chr.12: 7133921-7133921 on Build GRCh38
rs375794985 Chr.12: 7133703-7133703 on Build GRCh38
rs749951518 Chr.12: 7133731-7133731 on Build GRCh38
rs192085039 Chr.12: 7133837-7133837 on Build GRCh38
rs1039859392 Chr.12: 7133527-7133527 on Build GRCh38
rs754859199 Chr.12: 7133629-7133629 on Build GRCh38
rs934250256 Chr.12: 7133509-7133509 on Build GRCh38
rs764913430 Chr.12: 7133730-7133730 on Build GRCh38
rs905223465 Chr.12: 7133870-7133870 on Build GRCh38
rs182971253 Chr.12: 7133847-7133847 on Build GRCh38
rs776397587 Chr.12: 7133712-7133712 on Build GRCh38
rs892461185 Chr.12: 7133564-7133564 on Build GRCh38
rs770193497 Chr.12: 7133577-7133577 on Build GRCh38
rs141936578 Chr.12: 7133640-7133640 on Build GRCh38
rs781710631 Chr.12: 7133655-7133655 on Build GRCh38
rs753394177 Chr.12: 7133542-7133542 on Build GRCh38
rs747418543 Chr.12: 7133557-7133557 on Build GRCh38
rs376456144 Chr.12: 7133873-7133873 on Build GRCh38
rs922245338 Chr.12: 7133790-7133790 on Build GRCh38
rs775187858 Chr.12: 7133610-7133610 on Build GRCh38
rs756135597 Chr.12: 7133640-7133641 on Build GRCh38
rs369094080 Chr.12: 7133742-7133742 on Build GRCh38
rs892715767 Chr.12: 7133848-7133848 on Build GRCh38
rs780569046 Chr.12: 7133551-7133551 on Build GRCh38
rs748515566 Chr.12: 7133568-7133568 on Build GRCh38
rs375695520 Chr.12: 7133619-7133619 on Build GRCh38
rs200533858 Chr.12: 7133682-7133682 on Build GRCh38
rs767944006 Chr.12: 7133739-7133739 on Build GRCh38
rs149627692 Chr.12: 7133731-7133732 on Build GRCh38
rs756182506 Chr.12: 7133639-7133639 on Build GRCh38
rs147540436 Chr.12: 7133550-7133550 on Build GRCh38
rs760494012 Chr.12: 7133710-7133710 on Build GRCh38
rs144627317 Chr.12: 7133740-7133740 on Build GRCh38
rs150647098 Chr.12: 7133671-7133671 on Build GRCh38
rs112073544 Chr.12: 7133515-7133515 on Build GRCh38
rs867144396 Chr.12: 7133889-7133889 on Build GRCh38
rs148147388 Chr.12: 7133627-7133627 on Build GRCh38
rs936867258 Chr.12: 7133845-7133845 on Build GRCh38
rs199554289 Chr.12: 7133561-7133561 on Build GRCh38
rs754248568 Chr.12: 7133787-7133787 on Build GRCh38
rs975154851 Chr.12: 7133841-7133841 on Build GRCh38
rs146392825 Chr.12: 7133623-7133623 on Build GRCh38
rs761520601 Chr.12: 7133724-7133724 on Build GRCh38
rs781478472 Chr.12: 7133563-7133563 on Build GRCh38
rs755786416 Chr.12: 7133530-7133530 on Build GRCh38
rs1025026175 Chr.12: 7133606-7133606 on Build GRCh38
rs368878804 Chr.12: 7133584-7133584 on Build GRCh38
rs898320756 Chr.12: 7133917-7133917 on Build GRCh38
rs763448776 Chr.12: 7133938-7133938 on Build GRCh38
rs767482993 Chr.12: 7133608-7133608 on Build GRCh38
rs367959543 Chr.12: 7133677-7133677 on Build GRCh38
rs911366732 Chr.12: 7133517-7133517 on Build GRCh38
rs762478857 Chr.12: 7133736-7133736 on Build GRCh38
rs755469584 Chr.12: 7133700-7133700 on Build GRCh38
rs974014487 Chr.12: 7133709-7133709 on Build GRCh38
rs757590929 Chr.12: 7133802-7133802 on Build GRCh38
rs986446485 Chr.12: 7133760-7133760 on Build GRCh38
rs772151021 Chr.12: 7133812-7133812 on Build GRCh38
rs748485897 Chr.12: 7133659-7133659 on Build GRCh38
rs201216710 Chr.12: 7133660-7133660 on Build GRCh38
rs764125918 Chr.12: 7133854-7133854 on Build GRCh38
rs139728882 Chr.12: 7133698-7133698 on Build GRCh38
rs912282901 Chr.12: 7133781-7133781 on Build GRCh38
rs367854375 Chr.12: 7133583-7133583 on Build GRCh38
rs1016800894 Chr.12: 7133898-7133898 on Build GRCh38
rs201731889 Chr.12: 7133591-7133591 on Build GRCh38
rs371847868 Chr.12: 7133844-7133844 on Build GRCh38
rs759447166 Chr.12: 7133595-7133595 on Build GRCh38
rs755479765 Chr.12: 7133642-7133642 on Build GRCh38
rs200598370 Chr.12: 7133522-7133522 on Build GRCh38
rs745956586 Chr.12: 7133806-7133806 on Build GRCh38

View Full Product Details