Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7135205-7135704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139941 c.451C>T p.R151C Chr.12: 7135394-7135394 on Build GRCh38
3465100 c.576C>T p.F192F Chr.12: 7135519-7135519 on Build GRCh38
943340 c.395_409del15 p.R134_V138delRVNDV Chr.12: 7135338-7135352 on Build GRCh38
4930000 c.442C>G p.R148G Chr.12: 7135385-7135385 on Build GRCh38
1513179 c.485G>A p.R162H Chr.12: 7135428-7135428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375784823 Chr.12: 7135282-7135282 on Build GRCh38
rs762377892 Chr.12: 7135302-7135302 on Build GRCh38
rs375653650 Chr.12: 7135454-7135454 on Build GRCh38
rs779033354 Chr.12: 7135327-7135327 on Build GRCh38
rs6488452 Chr.12: 7135290-7135290 on Build GRCh38
rs747634878 Chr.12: 7135568-7135568 on Build GRCh38
rs939004366 Chr.12: 7135245-7135245 on Build GRCh38
rs1011652728 Chr.12: 7135660-7135660 on Build GRCh38
rs149152741 Chr.12: 7135453-7135453 on Build GRCh38
rs750591063 Chr.12: 7135338-7135338 on Build GRCh38
rs79158916 Chr.12: 7135312-7135312 on Build GRCh38
rs761575660 Chr.12: 7135291-7135291 on Build GRCh38
rs368087485 Chr.12: 7135547-7135547 on Build GRCh38
rs977234727 Chr.12: 7135468-7135468 on Build GRCh38
rs777983572 Chr.12: 7135297-7135297 on Build GRCh38
rs751717161 Chr.12: 7135516-7135516 on Build GRCh38
rs147467911 Chr.12: 7135342-7135342 on Build GRCh38
rs902123235 Chr.12: 7135610-7135610 on Build GRCh38
rs754301712 Chr.12: 7135321-7135321 on Build GRCh38
rs745458695 Chr.12: 7135586-7135586 on Build GRCh38
rs756113801 Chr.12: 7135550-7135550 on Build GRCh38
rs749002728 Chr.12: 7135467-7135467 on Build GRCh38
rs772756740 Chr.12: 7135374-7135374 on Build GRCh38
rs777236671 Chr.12: 7135399-7135399 on Build GRCh38
rs143301050 Chr.12: 7135474-7135474 on Build GRCh38
rs748752481 Chr.12: 7135446-7135446 on Build GRCh38
rs977935449 Chr.12: 7135683-7135683 on Build GRCh38
rs370600730 Chr.12: 7135505-7135505 on Build GRCh38
rs771640655 Chr.12: 7135483-7135483 on Build GRCh38
rs751096948 Chr.12: 7135314-7135314 on Build GRCh38
rs751700027 Chr.12: 7135344-7135344 on Build GRCh38
rs750546549 Chr.12: 7135292-7135292 on Build GRCh38
rs140264751 Chr.12: 7135486-7135486 on Build GRCh38
rs375251480 Chr.12: 7135317-7135317 on Build GRCh38
rs757075797 Chr.12: 7135583-7135583 on Build GRCh38
rs770482600 Chr.12: 7135379-7135379 on Build GRCh38
rs187600268 Chr.12: 7135351-7135351 on Build GRCh38
rs778192570 Chr.12: 7135560-7135560 on Build GRCh38
rs376219915 Chr.12: 7135279-7135279 on Build GRCh38
rs770663115 Chr.12: 7135469-7135469 on Build GRCh38
rs749024109 Chr.12: 7135378-7135378 on Build GRCh38
rs778711365 Chr.12: 7135584-7135584 on Build GRCh38
rs948451473 Chr.12: 7135549-7135549 on Build GRCh38
rs138998893 Chr.12: 7135634-7135634 on Build GRCh38
rs368350140 Chr.12: 7135465-7135465 on Build GRCh38
rs374604825 Chr.12: 7135275-7135275 on Build GRCh38
rs772884379 Chr.12: 7135294-7135294 on Build GRCh38
rs762898070 Chr.12: 7135239-7135239 on Build GRCh38
rs185085991 Chr.12: 7135636-7135636 on Build GRCh38
rs764718194 Chr.12: 7135398-7135398 on Build GRCh38
rs769568594 Chr.12: 7135369-7135369 on Build GRCh38
rs2278822 Chr.12: 7135318-7135318 on Build GRCh38
rs1005015982 Chr.12: 7135599-7135599 on Build GRCh38
rs773182437 Chr.12: 7135240-7135240 on Build GRCh38
rs754688978 Chr.12: 7135543-7135543 on Build GRCh38
rs762440271 Chr.12: 7135491-7135491 on Build GRCh38
rs759093363 Chr.12: 7135385-7135385 on Build GRCh38
rs764679503 Chr.12: 7135405-7135405 on Build GRCh38
rs781061270 Chr.12: 7135352-7135352 on Build GRCh38
rs745401266 Chr.12: 7135476-7135476 on Build GRCh38
rs188363076 Chr.12: 7135652-7135652 on Build GRCh38
rs1040564675 Chr.12: 7135604-7135604 on Build GRCh38
rs899421429 Chr.12: 7135601-7135601 on Build GRCh38
rs753589029 Chr.12: 7135534-7135534 on Build GRCh38
rs145077643 Chr.12: 7135423-7135423 on Build GRCh38
rs147044460 Chr.12: 7135682-7135682 on Build GRCh38
rs149090453 Chr.12: 7135436-7135436 on Build GRCh38
rs763179688 Chr.12: 7135506-7135506 on Build GRCh38
rs757704345 Chr.12: 7135563-7135563 on Build GRCh38
rs754734394 Chr.12: 7135648-7135648 on Build GRCh38
rs779860561 Chr.12: 7135276-7135276 on Build GRCh38
rs770288033 Chr.12: 7135455-7135455 on Build GRCh38
rs372922249 Chr.12: 7135400-7135400 on Build GRCh38
rs758688276 Chr.12: 7135411-7135411 on Build GRCh38
rs915076959 Chr.12: 7135421-7135421 on Build GRCh38
rs774041468 Chr.12: 7135381-7135381 on Build GRCh38
rs765630565 Chr.12: 7135503-7135503 on Build GRCh38
rs749400529 Chr.12: 7135408-7135409 on Build GRCh38
rs371263668 Chr.12: 7135567-7135567 on Build GRCh38
rs761455824 Chr.12: 7135665-7135665 on Build GRCh38
rs755013198 Chr.12: 7135427-7135427 on Build GRCh38
rs1037918036 Chr.12: 7135556-7135556 on Build GRCh38
rs1020388347 Chr.12: 7135679-7135679 on Build GRCh38
rs117963905 Chr.12: 7135323-7135323 on Build GRCh38
rs570605012 Chr.12: 7135659-7135659 on Build GRCh38
rs142150232 Chr.12: 7135445-7135445 on Build GRCh38
rs200143604 Chr.12: 7135515-7135515 on Build GRCh38
rs34933247 Chr.12: 7135360-7135360 on Build GRCh38
rs370422712 Chr.12: 7135565-7135565 on Build GRCh38
rs766566628 Chr.12: 7135418-7135418 on Build GRCh38

View Full Product Details