Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7136738-7137236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3727198 c.1180G>A p.E394K Chr.12: 7137080-7137080 on Build GRCh38
297101 c.978C>T p.A326A Chr.12: 7136878-7136878 on Build GRCh38
404579 c.1034G>T p.W345L Chr.12: 7136934-7136934 on Build GRCh38
6296902 c.1022G>T p.S341I Chr.12: 7136922-7136922 on Build GRCh38
4633735 c.999G>A p.S333S Chr.12: 7136899-7136899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968836097 Chr.12: 7136803-7136803 on Build GRCh38
rs776274636 Chr.12: 7136987-7136987 on Build GRCh38
rs770510625 Chr.12: 7137135-7137135 on Build GRCh38
rs768207743 Chr.12: 7136778-7136778 on Build GRCh38
rs747870105 Chr.12: 7137124-7137124 on Build GRCh38
rs766543184 Chr.12: 7137083-7137083 on Build GRCh38
rs1031899524 Chr.12: 7136865-7136865 on Build GRCh38
rs781079377 Chr.12: 7137119-7137119 on Build GRCh38
rs777104227 Chr.12: 7137155-7137155 on Build GRCh38
rs182094956 Chr.12: 7137050-7137050 on Build GRCh38
rs768112843 Chr.12: 7136902-7136902 on Build GRCh38
rs776076434 Chr.12: 7137028-7137028 on Build GRCh38
rs957125101 Chr.12: 7136765-7136765 on Build GRCh38
rs956858627 Chr.12: 7137123-7137123 on Build GRCh38
rs1021772328 Chr.12: 7137200-7137200 on Build GRCh38
rs754037443 Chr.12: 7136923-7136923 on Build GRCh38
rs1040088215 Chr.12: 7136955-7136955 on Build GRCh38
rs754903769 Chr.12: 7137113-7137113 on Build GRCh38
rs2290236 Chr.12: 7137163-7137163 on Build GRCh38
rs779979604 Chr.12: 7137098-7137098 on Build GRCh38
rs375191507 Chr.12: 7136804-7136804 on Build GRCh38
rs768166019 Chr.12: 7137024-7137024 on Build GRCh38
rs745878740 Chr.12: 7136864-7136864 on Build GRCh38
rs746811144 Chr.12: 7136965-7136965 on Build GRCh38
rs776279570 Chr.12: 7136880-7136880 on Build GRCh38
rs1007231622 Chr.12: 7137186-7137186 on Build GRCh38
rs778088083 Chr.12: 7136844-7136844 on Build GRCh38
rs775306150 Chr.12: 7136868-7136868 on Build GRCh38
rs763705492 Chr.12: 7136802-7136802 on Build GRCh38
rs769680335 Chr.12: 7136898-7136898 on Build GRCh38
rs927171605 Chr.12: 7136854-7136854 on Build GRCh38
rs761345333 Chr.12: 7136819-7136819 on Build GRCh38
rs761139498 Chr.12: 7137049-7137049 on Build GRCh38
rs867263612 Chr.12: 7137022-7137022 on Build GRCh38
rs753130631 Chr.12: 7136903-7136903 on Build GRCh38
rs752044597 Chr.12: 7136826-7136826 on Build GRCh38
rs747899801 Chr.12: 7136988-7136988 on Build GRCh38
rs757410687 Chr.12: 7136926-7136926 on Build GRCh38
rs368454189 Chr.12: 7137160-7137160 on Build GRCh38
rs749363456 Chr.12: 7136845-7136845 on Build GRCh38
rs909988156 Chr.12: 7136985-7136985 on Build GRCh38
rs374178133 Chr.12: 7136785-7136785 on Build GRCh38
rs779918532 Chr.12: 7136960-7136960 on Build GRCh38
rs148348543 Chr.12: 7137089-7137089 on Build GRCh38
rs367923285 Chr.12: 7136825-7136825 on Build GRCh38
rs758379738 Chr.12: 7136952-7136952 on Build GRCh38
rs2290235 Chr.12: 7136964-7136964 on Build GRCh38
rs778732773 Chr.12: 7136858-7136858 on Build GRCh38
rs371509493 Chr.12: 7137045-7137045 on Build GRCh38
rs753077527 Chr.12: 7136835-7136835 on Build GRCh38
rs747006823 Chr.12: 7137105-7137105 on Build GRCh38
rs764858222 Chr.12: 7136893-7136893 on Build GRCh38
rs370885083 Chr.12: 7136899-7136899 on Build GRCh38
rs149615701 Chr.12: 7136837-7136837 on Build GRCh38
rs745906828 Chr.12: 7136942-7136942 on Build GRCh38
rs768382086 Chr.12: 7136979-7136979 on Build GRCh38
rs144317360 Chr.12: 7136990-7136990 on Build GRCh38
rs779119496 Chr.12: 7136927-7136927 on Build GRCh38
rs761333916 Chr.12: 7137141-7137141 on Build GRCh38
rs200951848 Chr.12: 7137061-7137061 on Build GRCh38
rs768320674 Chr.12: 7136874-7136874 on Build GRCh38
rs772825293 Chr.12: 7136994-7136994 on Build GRCh38
rs761243186 Chr.12: 7136890-7136890 on Build GRCh38
rs777008868 Chr.12: 7136871-7136871 on Build GRCh38
rs763717646 Chr.12: 7137088-7137088 on Build GRCh38
rs764419705 Chr.12: 7136920-7136920 on Build GRCh38
rs756503676 Chr.12: 7136913-7136913 on Build GRCh38
rs773330447 Chr.12: 7136941-7136941 on Build GRCh38
rs770980080 Chr.12: 7136857-7136857 on Build GRCh38
rs773988935 Chr.12: 7137139-7137139 on Build GRCh38
rs775254382 Chr.12: 7136780-7136780 on Build GRCh38
rs765356936 Chr.12: 7137073-7137073 on Build GRCh38
rs755424732 Chr.12: 7136833-7136833 on Build GRCh38
rs993119763 Chr.12: 7136894-7136894 on Build GRCh38
rs769015353 Chr.12: 7137152-7137152 on Build GRCh38
rs781731281 Chr.12: 7136834-7136834 on Build GRCh38
rs376026535 Chr.12: 7137129-7137129 on Build GRCh38
rs201397184 Chr.12: 7137162-7137162 on Build GRCh38
rs762499119 Chr.12: 7136998-7136998 on Build GRCh38
rs367782237 Chr.12: 7136968-7136968 on Build GRCh38
rs772695527 Chr.12: 7137132-7137132 on Build GRCh38
rs901602538 Chr.12: 7136969-7136969 on Build GRCh38
rs771994677 Chr.12: 7136867-7136867 on Build GRCh38

View Full Product Details