Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7137876-7138388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747234 c.1258G>A p.A420T Chr.12: 7138002-7138002 on Build GRCh38
4845057 c.1303T>C p.F435L Chr.12: 7138047-7138047 on Build GRCh38
4044737 c.1281T>C p.L427L Chr.12: 7138025-7138025 on Build GRCh38
549819 c.1292G>A p.R431H Chr.12: 7138036-7138036 on Build GRCh38
1299860 c.1223C>G p.S408C Chr.12: 7137967-7137967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758576395 Chr.12: 7138331-7138331 on Build GRCh38
rs749011347 Chr.12: 7137967-7137967 on Build GRCh38
rs1032125660 Chr.12: 7138273-7138274 on Build GRCh38
rs568295762 Chr.12: 7138250-7138250 on Build GRCh38
rs371860737 Chr.12: 7138030-7138030 on Build GRCh38
rs772566349 Chr.12: 7138344-7138345 on Build GRCh38
rs978602933 Chr.12: 7138351-7138351 on Build GRCh38
rs375311499 Chr.12: 7137943-7137943 on Build GRCh38
rs777180875 Chr.12: 7138046-7138046 on Build GRCh38
rs376591370 Chr.12: 7138035-7138035 on Build GRCh38
rs901128027 Chr.12: 7138172-7138172 on Build GRCh38
rs771171096 Chr.12: 7138111-7138111 on Build GRCh38
rs754988397 Chr.12: 7137947-7137947 on Build GRCh38
rs770113027 Chr.12: 7137992-7137992 on Build GRCh38
rs1018784390 Chr.12: 7138321-7138321 on Build GRCh38
rs142696668 Chr.12: 7138096-7138096 on Build GRCh38
rs780745938 Chr.12: 7138310-7138310 on Build GRCh38
rs535754781 Chr.12: 7138352-7138352 on Build GRCh38
rs748669780 Chr.12: 7138097-7138097 on Build GRCh38
rs774043979 Chr.12: 7137984-7137984 on Build GRCh38
rs752563612 Chr.12: 7137957-7137957 on Build GRCh38
rs376514416 Chr.12: 7138099-7138099 on Build GRCh38
rs778609307 Chr.12: 7137976-7137976 on Build GRCh38
rs769370309 Chr.12: 7138343-7138344 on Build GRCh38
rs941533229 Chr.12: 7138273-7138273 on Build GRCh38
rs988460879 Chr.12: 7138231-7138231 on Build GRCh38
rs780251590 Chr.12: 7138312-7138312 on Build GRCh38
rs781625555 Chr.12: 7138018-7138018 on Build GRCh38
rs781344032 Chr.12: 7138227-7138227 on Build GRCh38
rs778662140 Chr.12: 7138052-7138052 on Build GRCh38
rs1044572388 Chr.12: 7138251-7138251 on Build GRCh38
rs149602492 Chr.12: 7138342-7138343 on Build GRCh38
rs763200937 Chr.12: 7138014-7138014 on Build GRCh38
rs938135795 Chr.12: 7138143-7138143 on Build GRCh38
rs916922740 Chr.12: 7138237-7138237 on Build GRCh38
rs754806687 Chr.12: 7138274-7138274 on Build GRCh38
rs139911367 Chr.12: 7138021-7138021 on Build GRCh38
rs757781176 Chr.12: 7138091-7138091 on Build GRCh38
rs372177991 Chr.12: 7137942-7137942 on Build GRCh38
rs773561186 Chr.12: 7138008-7138008 on Build GRCh38
rs147478372 Chr.12: 7137989-7137989 on Build GRCh38
rs756068728 Chr.12: 7138036-7138036 on Build GRCh38
rs776964256 Chr.12: 7137987-7137987 on Build GRCh38
rs764954014 Chr.12: 7138280-7138280 on Build GRCh38
rs985974755 Chr.12: 7138123-7138123 on Build GRCh38
rs781112610 Chr.12: 7138162-7138162 on Build GRCh38
rs749601988 Chr.12: 7138110-7138110 on Build GRCh38
rs143159646 Chr.12: 7137977-7137977 on Build GRCh38
rs953316415 Chr.12: 7138221-7138221 on Build GRCh38
rs994407621 Chr.12: 7138297-7138297 on Build GRCh38
rs368707350 Chr.12: 7137959-7137959 on Build GRCh38
rs750673387 Chr.12: 7138350-7138350 on Build GRCh38
rs1015996984 Chr.12: 7137900-7137900 on Build GRCh38
rs988293828 Chr.12: 7138276-7138277 on Build GRCh38
rs751658003 Chr.12: 7138019-7138019 on Build GRCh38
rs766986566 Chr.12: 7138027-7138027 on Build GRCh38
rs752305732 Chr.12: 7137990-7137990 on Build GRCh38
rs753656821 Chr.12: 7138041-7138041 on Build GRCh38
rs1006969873 Chr.12: 7138320-7138320 on Build GRCh38
rs1037156226 Chr.12: 7138168-7138168 on Build GRCh38
rs1029648494 Chr.12: 7138298-7138298 on Build GRCh38
rs888646474 Chr.12: 7138160-7138160 on Build GRCh38
rs934284383 Chr.12: 7138204-7138204 on Build GRCh38
rs746525430 Chr.12: 7138163-7138163 on Build GRCh38
rs777585973 Chr.12: 7137960-7137960 on Build GRCh38
rs905996748 Chr.12: 7138267-7138267 on Build GRCh38
rs949827530 Chr.12: 7138248-7138248 on Build GRCh38
rs780939894 Chr.12: 7137948-7137948 on Build GRCh38
rs926874851 Chr.12: 7138130-7138130 on Build GRCh38
rs750584681 Chr.12: 7137917-7137917 on Build GRCh38
rs762510627 Chr.12: 7137945-7137945 on Build GRCh38
rs754472628 Chr.12: 7138293-7138293 on Build GRCh38
rs777174345 Chr.12: 7137915-7137915 on Build GRCh38
rs745417202 Chr.12: 7138089-7138089 on Build GRCh38
rs779418176 Chr.12: 7138129-7138129 on Build GRCh38
rs964552030 Chr.12: 7138346-7138346 on Build GRCh38
rs964938857 Chr.12: 7138144-7138144 on Build GRCh38
rs867604375 Chr.12: 7138095-7138095 on Build GRCh38
rs926300200 Chr.12: 7138335-7138335 on Build GRCh38
rs369285236 Chr.12: 7138037-7138037 on Build GRCh38
rs182097706 Chr.12: 7138174-7138174 on Build GRCh38
rs773614200 Chr.12: 7138101-7138101 on Build GRCh38
rs763148039 Chr.12: 7137924-7137924 on Build GRCh38

View Full Product Details