Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7373234-7373742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163L1
Entrez Gene ID
283316
Gene Name
CD163 molecule like 1
Gene Aliases
CD163B, M160, SCARI2
UniGene
Hs.631727
Gene Chromosome Location
Chr.12: 7320510-7444185 on Build GRCh38
Transcripts
NM_001297650.1 NM_174941.5 XM_011520616.1 XM_011520617.2 XM_011520618.2 XM_011520619.1 XM_011520620.1 XM_011520621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6073607 c.3469G>C p.E1157Q Chr.12: 7373581-7373581 on Build GRCh38
3987204 c.3645G>A p.T1215T Chr.12: 7373405-7373405 on Build GRCh38
4544352 c.3552G>A p.L1184L Chr.12: 7373498-7373498 on Build GRCh38
1705027 c.3569G>A p.G1190E Chr.12: 7373481-7373481 on Build GRCh38
1364225 c.3579C>T p.S1193S Chr.12: 7373471-7373471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750251476 Chr.12: 7373658-7373658 on Build GRCh38
rs762793760 Chr.12: 7373666-7373666 on Build GRCh38
rs371023443 Chr.12: 7373349-7373349 on Build GRCh38
rs762781774 Chr.12: 7373470-7373470 on Build GRCh38
rs941058764 Chr.12: 7373284-7373284 on Build GRCh38
rs368371081 Chr.12: 7373273-7373273 on Build GRCh38
rs745466357 Chr.12: 7373478-7373478 on Build GRCh38
rs752511808 Chr.12: 7373300-7373300 on Build GRCh38
rs778666311 Chr.12: 7373471-7373471 on Build GRCh38
rs759665574 Chr.12: 7373425-7373425 on Build GRCh38
rs1038428895 Chr.12: 7373297-7373297 on Build GRCh38
rs368648151 Chr.12: 7373468-7373468 on Build GRCh38
rs746578351 Chr.12: 7373567-7373567 on Build GRCh38
rs771585037 Chr.12: 7373520-7373520 on Build GRCh38
rs199578853 Chr.12: 7373510-7373510 on Build GRCh38
rs749669056 Chr.12: 7373511-7373511 on Build GRCh38
rs761887748 Chr.12: 7373642-7373642 on Build GRCh38
rs372850983 Chr.12: 7373617-7373617 on Build GRCh38
rs779199170 Chr.12: 7373593-7373593 on Build GRCh38
rs770716204 Chr.12: 7373504-7373504 on Build GRCh38
rs751670195 Chr.12: 7373645-7373645 on Build GRCh38
rs760838710 Chr.12: 7373619-7373619 on Build GRCh38
rs1000907748 Chr.12: 7373482-7373482 on Build GRCh38
rs763298117 Chr.12: 7373400-7373400 on Build GRCh38
rs749248792 Chr.12: 7373551-7373551 on Build GRCh38
rs371280203 Chr.12: 7373449-7373449 on Build GRCh38
rs764137580 Chr.12: 7373533-7373533 on Build GRCh38
rs771308162 Chr.12: 7373583-7373583 on Build GRCh38
rs995260440 Chr.12: 7373316-7373316 on Build GRCh38
rs149044372 Chr.12: 7373488-7373488 on Build GRCh38
rs139734826 Chr.12: 7373536-7373536 on Build GRCh38
rs748553409 Chr.12: 7373387-7373387 on Build GRCh38
rs767456913 Chr.12: 7373438-7373438 on Build GRCh38
rs775920670 Chr.12: 7373616-7373616 on Build GRCh38
rs144886397 Chr.12: 7373467-7373467 on Build GRCh38
rs751148361 Chr.12: 7373685-7373685 on Build GRCh38
rs765962896 Chr.12: 7373679-7373679 on Build GRCh38
rs980322713 Chr.12: 7373549-7373549 on Build GRCh38
rs769336948 Chr.12: 7373258-7373258 on Build GRCh38
rs986491959 Chr.12: 7373711-7373711 on Build GRCh38
rs749072705 Chr.12: 7373337-7373337 on Build GRCh38
rs766524619 Chr.12: 7373487-7373488 on Build GRCh38
rs11053518 Chr.12: 7373269-7373269 on Build GRCh38
rs142968338 Chr.12: 7373553-7373553 on Build GRCh38
rs898340806 Chr.12: 7373289-7373289 on Build GRCh38
rs886884623 Chr.12: 7373591-7373591 on Build GRCh38
rs918375123 Chr.12: 7373341-7373341 on Build GRCh38
rs951195637 Chr.12: 7373374-7373374 on Build GRCh38
rs117904047 Chr.12: 7373530-7373530 on Build GRCh38
rs770351398 Chr.12: 7373501-7373501 on Build GRCh38
rs746496144 Chr.12: 7373500-7373500 on Build GRCh38
rs781765194 Chr.12: 7373364-7373364 on Build GRCh38
rs199702335 Chr.12: 7373627-7373627 on Build GRCh38
rs777666138 Chr.12: 7373313-7373313 on Build GRCh38
rs141611631 Chr.12: 7373525-7373525 on Build GRCh38
rs746217746 Chr.12: 7373597-7373597 on Build GRCh38
rs781202762 Chr.12: 7373283-7373283 on Build GRCh38
rs973803162 Chr.12: 7373629-7373629 on Build GRCh38
rs34390248 Chr.12: 7373570-7373570 on Build GRCh38
rs779749979 Chr.12: 7373566-7373566 on Build GRCh38
rs963721200 Chr.12: 7373410-7373410 on Build GRCh38
rs995333837 Chr.12: 7373564-7373564 on Build GRCh38
rs777222857 Chr.12: 7373375-7373375 on Build GRCh38
rs11368626 Chr.12: 7373711-7373712 on Build GRCh38
rs755335200 Chr.12: 7373365-7373365 on Build GRCh38
rs918220956 Chr.12: 7373661-7373661 on Build GRCh38
rs971196157 Chr.12: 7373541-7373541 on Build GRCh38
rs1025430923 Chr.12: 7373373-7373373 on Build GRCh38
rs948687888 Chr.12: 7373537-7373537 on Build GRCh38
rs375149226 Chr.12: 7373393-7373393 on Build GRCh38
rs772252828 Chr.12: 7373598-7373598 on Build GRCh38
rs760836443 Chr.12: 7373532-7373532 on Build GRCh38
rs375934529 Chr.12: 7373569-7373569 on Build GRCh38
rs768813525 Chr.12: 7373632-7373632 on Build GRCh38
rs371725390 Chr.12: 7373544-7373544 on Build GRCh38
rs929743341 Chr.12: 7373677-7373677 on Build GRCh38
rs778259990 Chr.12: 7373571-7373571 on Build GRCh38
rs774379913 Chr.12: 7373406-7373406 on Build GRCh38
rs765189788 Chr.12: 7373650-7373650 on Build GRCh38
rs34600496 Chr.12: 7373551-7373552 on Build GRCh38
rs374489185 Chr.12: 7373457-7373457 on Build GRCh38
rs749755256 Chr.12: 7373581-7373581 on Build GRCh38
rs368002565 Chr.12: 7373405-7373405 on Build GRCh38
rs773604158 Chr.12: 7373395-7373395 on Build GRCh38
rs368402880 Chr.12: 7373636-7373636 on Build GRCh38
rs764888319 Chr.12: 7373548-7373548 on Build GRCh38
rs778692830 Chr.12: 7373350-7373350 on Build GRCh38
rs779478020 Chr.12: 7373496-7373496 on Build GRCh38
rs745439396 Chr.12: 7373524-7373524 on Build GRCh38

View Full Product Details