Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8039740-8040254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
UniGene
Hs.120844
Gene Chromosome Location
Chr.12: 8032680-8055522 on Build GRCh38
Transcripts
NM_018416.2 XM_011520760.2 XM_011520761.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841183 c.273G>T p.E91D Chr.12: 8040105-8040105 on Build GRCh38
6895308 c.94C>T p.Q32* Chr.12: 8039926-8039926 on Build GRCh38
5909233 c.61C>T p.R21* Chr.12: 8039893-8039893 on Build GRCh38
3465982 c.242C>T p.S81F Chr.12: 8040074-8040074 on Build GRCh38
7307206 c.1A>C p.M1L Chr.12: 8039833-8039833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202211207 Chr.12: 8039836-8039836 on Build GRCh38
rs781311522 Chr.12: 8039924-8039924 on Build GRCh38
rs757505817 Chr.12: 8039781-8039781 on Build GRCh38
rs374823580 Chr.12: 8040191-8040191 on Build GRCh38
rs377340764 Chr.12: 8040087-8040087 on Build GRCh38
rs12580604 Chr.12: 8040201-8040201 on Build GRCh38
rs372183229 Chr.12: 8039933-8039933 on Build GRCh38
rs747981281 Chr.12: 8039844-8039844 on Build GRCh38
rs752615890 Chr.12: 8039927-8039927 on Build GRCh38
rs757005116 Chr.12: 8040150-8040150 on Build GRCh38
rs767648355 Chr.12: 8039967-8039967 on Build GRCh38
rs148966560 Chr.12: 8040024-8040024 on Build GRCh38
rs201469015 Chr.12: 8039950-8039950 on Build GRCh38
rs755989740 Chr.12: 8039929-8039929 on Build GRCh38
rs759695512 Chr.12: 8039961-8039961 on Build GRCh38
rs750670202 Chr.12: 8039894-8039894 on Build GRCh38
rs1019306444 Chr.12: 8040086-8040086 on Build GRCh38
rs751570983 Chr.12: 8039903-8039903 on Build GRCh38
rs147737639 Chr.12: 8039951-8039951 on Build GRCh38
rs781695258 Chr.12: 8040049-8040049 on Build GRCh38
rs952617945 Chr.12: 8040193-8040193 on Build GRCh38
rs749748296 Chr.12: 8040211-8040211 on Build GRCh38
rs965791384 Chr.12: 8039838-8039838 on Build GRCh38
rs767775080 Chr.12: 8040111-8040111 on Build GRCh38
rs765518292 Chr.12: 8039892-8039892 on Build GRCh38
rs369499195 Chr.12: 8040017-8040017 on Build GRCh38
rs932023917 Chr.12: 8040014-8040014 on Build GRCh38
rs777420472 Chr.12: 8039931-8039931 on Build GRCh38
rs376535944 Chr.12: 8040003-8040003 on Build GRCh38
rs538093373 Chr.12: 8040175-8040175 on Build GRCh38
rs762193187 Chr.12: 8039863-8039863 on Build GRCh38
rs754472539 Chr.12: 8040195-8040195 on Build GRCh38
rs747023744 Chr.12: 8039795-8039795 on Build GRCh38
rs59574600 Chr.12: 8039818-8039818 on Build GRCh38
rs368273372 Chr.12: 8039782-8039782 on Build GRCh38
rs745455298 Chr.12: 8040013-8040013 on Build GRCh38
rs552576369 Chr.12: 8040188-8040190 on Build GRCh38
rs771259531 Chr.12: 8040213-8040213 on Build GRCh38
rs756022459 Chr.12: 8039943-8039943 on Build GRCh38
rs139454481 Chr.12: 8039854-8039854 on Build GRCh38
rs745388112 Chr.12: 8039941-8039941 on Build GRCh38
rs143696902 Chr.12: 8040135-8040135 on Build GRCh38
rs377097169 Chr.12: 8039784-8039784 on Build GRCh38
rs748720437 Chr.12: 8040026-8040026 on Build GRCh38
rs201952649 Chr.12: 8040112-8040112 on Build GRCh38
rs751765056 Chr.12: 8039956-8039956 on Build GRCh38
rs201738561 Chr.12: 8039975-8039975 on Build GRCh38
rs764560182 Chr.12: 8039856-8039856 on Build GRCh38
rs769672438 Chr.12: 8039847-8039847 on Build GRCh38
rs965387632 Chr.12: 8040098-8040098 on Build GRCh38
rs759823116 Chr.12: 8040100-8040100 on Build GRCh38
rs945571125 Chr.12: 8040227-8040227 on Build GRCh38
rs921371432 Chr.12: 8039904-8039904 on Build GRCh38
rs149967567 Chr.12: 8039952-8039952 on Build GRCh38
rs766547030 Chr.12: 8039898-8039898 on Build GRCh38
rs765462945 Chr.12: 8039783-8039783 on Build GRCh38
rs12580580 Chr.12: 8039994-8039994 on Build GRCh38
rs770880535 Chr.12: 8039946-8039946 on Build GRCh38
rs193084823 Chr.12: 8040054-8040054 on Build GRCh38
rs748837550 Chr.12: 8039849-8039849 on Build GRCh38
rs753556858 Chr.12: 8039996-8039996 on Build GRCh38
rs763073110 Chr.12: 8039896-8039896 on Build GRCh38
rs780430492 Chr.12: 8039809-8039809 on Build GRCh38
rs763956521 Chr.12: 8040122-8040122 on Build GRCh38
rs763258765 Chr.12: 8040060-8040060 on Build GRCh38
rs144780651 Chr.12: 8039971-8039971 on Build GRCh38
rs765291949 Chr.12: 8039780-8039780 on Build GRCh38
rs756932068 Chr.12: 8040214-8040215 on Build GRCh38
rs925597042 Chr.12: 8040035-8040035 on Build GRCh38
rs369271501 Chr.12: 8039955-8039955 on Build GRCh38
rs746500310 Chr.12: 8040192-8040192 on Build GRCh38
rs773731922 Chr.12: 8039852-8039852 on Build GRCh38
rs751635111 Chr.12: 8039859-8039859 on Build GRCh38
rs770536517 Chr.12: 8039850-8039850 on Build GRCh38
rs369154050 Chr.12: 8039821-8039821 on Build GRCh38
rs771262165 Chr.12: 8040084-8040084 on Build GRCh38
rs200461151 Chr.12: 8039794-8039794 on Build GRCh38
rs1013267549 Chr.12: 8040170-8040170 on Build GRCh38
rs774855324 Chr.12: 8040214-8040214 on Build GRCh38
rs773004970 Chr.12: 8039848-8039848 on Build GRCh38
rs207472641 Chr.12: 8039824-8039824 on Build GRCh38
rs760748528 Chr.12: 8040113-8040113 on Build GRCh38
rs758509800 Chr.12: 8039789-8039789 on Build GRCh38
rs200593250 Chr.12: 8040172-8040172 on Build GRCh38
rs777680047 Chr.12: 8039912-8039912 on Build GRCh38
rs770462460 Chr.12: 8039939-8039939 on Build GRCh38
rs145677680 Chr.12: 8039940-8039940 on Build GRCh38
rs758039940 Chr.12: 8040180-8040180 on Build GRCh38

View Full Product Details