Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9692926-9693471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2D
Entrez Gene ID
29121
Gene Name
C-type lectin domain family 2 member D
Gene Aliases
CLAX, LLT1, OCIL
UniGene
Hs.268326
Gene Chromosome Location
Chr.12: 9669708-9699555 on Build GRCh38
Transcripts
NM_001004419.4 NM_001197317.2 NM_001197318.2 NM_001197319.2 NM_013269.5 NR_036693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2046963 c.465A>T p.L155F Chr.12: 9693037-9693037 on Build GRCh38
239340 c.520C>T p.R174* Chr.12: 9693092-9693092 on Build GRCh38
7361497 c.479A>G p.D160G Chr.12: 9693051-9693051 on Build GRCh38
5423020 c.482G>T p.G161V Chr.12: 9693054-9693054 on Build GRCh38
4499183 c.536C>T p.P179L Chr.12: 9693108-9693108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772296283 Chr.12: 9693416-9693416 on Build GRCh38
rs758600880 Chr.12: 9692962-9692962 on Build GRCh38
rs1049470158 Chr.12: 9693096-9693096 on Build GRCh38
rs758481352 Chr.12: 9693406-9693406 on Build GRCh38
rs1001747765 Chr.12: 9693436-9693436 on Build GRCh38
rs943696298 Chr.12: 9693431-9693431 on Build GRCh38
rs944852963 Chr.12: 9693282-9693282 on Build GRCh38
rs377194446 Chr.12: 9693092-9693092 on Build GRCh38
rs769620489 Chr.12: 9693160-9693160 on Build GRCh38
rs200535095 Chr.12: 9693025-9693025 on Build GRCh38
rs775069923 Chr.12: 9693114-9693114 on Build GRCh38
rs763706366 Chr.12: 9693208-9693208 on Build GRCh38
rs771780259 Chr.12: 9693022-9693022 on Build GRCh38
rs751974042 Chr.12: 9692970-9692970 on Build GRCh38
rs1032991060 Chr.12: 9693011-9693011 on Build GRCh38
rs745808977 Chr.12: 9693144-9693144 on Build GRCh38
rs985175406 Chr.12: 9693407-9693410 on Build GRCh38
rs374099267 Chr.12: 9693125-9693125 on Build GRCh38
rs912187365 Chr.12: 9693212-9693212 on Build GRCh38
rs202185044 Chr.12: 9693093-9693093 on Build GRCh38
rs770807283 Chr.12: 9693040-9693040 on Build GRCh38
rs375780896 Chr.12: 9692993-9692993 on Build GRCh38
rs148171592 Chr.12: 9693323-9693323 on Build GRCh38
rs777426491 Chr.12: 9693123-9693123 on Build GRCh38
rs756637586 Chr.12: 9693111-9693111 on Build GRCh38
rs778334177 Chr.12: 9693007-9693007 on Build GRCh38
rs1053170922 Chr.12: 9693445-9693445 on Build GRCh38
rs143831127 Chr.12: 9693246-9693248 on Build GRCh38
rs971369440 Chr.12: 9693381-9693381 on Build GRCh38
rs762205003 Chr.12: 9693085-9693085 on Build GRCh38
rs142016017 Chr.12: 9693113-9693113 on Build GRCh38
rs770684592 Chr.12: 9693137-9693137 on Build GRCh38
rs960025575 Chr.12: 9693239-9693239 on Build GRCh38
rs772561684 Chr.12: 9693014-9693015 on Build GRCh38
rs1039214777 Chr.12: 9693295-9693295 on Build GRCh38
rs757037875 Chr.12: 9693242-9693242 on Build GRCh38
rs768032874 Chr.12: 9693095-9693095 on Build GRCh38
rs547230059 Chr.12: 9693350-9693350 on Build GRCh38
rs757776802 Chr.12: 9692980-9692980 on Build GRCh38
rs765341201 Chr.12: 9693059-9693059 on Build GRCh38
rs759434767 Chr.12: 9693054-9693054 on Build GRCh38
rs1001586318 Chr.12: 9693226-9693226 on Build GRCh38
rs1011506743 Chr.12: 9693175-9693175 on Build GRCh38
rs746821195 Chr.12: 9693035-9693035 on Build GRCh38
rs764419979 Chr.12: 9693089-9693089 on Build GRCh38
rs759992812 Chr.12: 9692952-9692953 on Build GRCh38
rs764546380 Chr.12: 9692969-9692969 on Build GRCh38
rs1024156491 Chr.12: 9693363-9693363 on Build GRCh38
rs1035809468 Chr.12: 9693238-9693238 on Build GRCh38
rs918261134 Chr.12: 9693150-9693150 on Build GRCh38
rs753391841 Chr.12: 9693194-9693194 on Build GRCh38
rs35664274 Chr.12: 9693431-9693432 on Build GRCh38
rs1005315554 Chr.12: 9693272-9693272 on Build GRCh38
rs548202184 Chr.12: 9693407-9693407 on Build GRCh38
rs768798597 Chr.12: 9693439-9693439 on Build GRCh38
rs778598723 Chr.12: 9693241-9693241 on Build GRCh38
rs1000575628 Chr.12: 9692995-9692995 on Build GRCh38
rs369608070 Chr.12: 9693138-9693138 on Build GRCh38
rs929510713 Chr.12: 9693164-9693164 on Build GRCh38
rs370902376 Chr.12: 9692992-9692992 on Build GRCh38
rs770852339 Chr.12: 9693347-9693347 on Build GRCh38
rs377414804 Chr.12: 9693127-9693127 on Build GRCh38
rs775493389 Chr.12: 9693073-9693073 on Build GRCh38
rs922133663 Chr.12: 9693032-9693032 on Build GRCh38
rs763146114 Chr.12: 9693166-9693166 on Build GRCh38
rs779191942 Chr.12: 9693141-9693141 on Build GRCh38
rs377519152 Chr.12: 9692954-9692954 on Build GRCh38
rs116932414 Chr.12: 9693107-9693107 on Build GRCh38
rs775578373 Chr.12: 9693162-9693162 on Build GRCh38
rs747821975 Chr.12: 9693009-9693009 on Build GRCh38
rs900560591 Chr.12: 9693309-9693309 on Build GRCh38
rs1015161684 Chr.12: 9693290-9693290 on Build GRCh38
rs905699258 Chr.12: 9693214-9693214 on Build GRCh38
rs185264640 Chr.12: 9693112-9693112 on Build GRCh38
rs777701407 Chr.12: 9693320-9693320 on Build GRCh38
rs776665801 Chr.12: 9693041-9693041 on Build GRCh38
rs754479014 Chr.12: 9692994-9692994 on Build GRCh38

View Full Product Details