Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9980500-9981000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12A
Entrez Gene ID
160364
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
CD371, CLL-1, CLL1, DCAL-2, MICL
UniGene
Hs.190519
Gene Chromosome Location
Chr.12: 9951316-9995209 on Build GRCh38
Transcripts
NM_001207010.1 NM_001300730.1 NM_138337.5 NM_201623.3 XM_005253324.2 XM_006719033.3 XM_006719035.3 XM_006719036.3 XM_011520570.1 XM_011520571.2 XM_011520573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2062924 c.527C>A p.A176D Chr.12: 9980699-9980699 on Build GRCh38
3416369 c.501C>T p.S167S Chr.12: 9980703-9980703 on Build GRCh38
6224292 c.490T>C p.W164R Chr.12: 9980662-9980662 on Build GRCh38
5732302 c.413T>A p.I138N Chr.12: 9980615-9980615 on Build GRCh38
5732301 c.443T>A p.I148N Chr.12: 9980615-9980615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556889668 Chr.12: 9980650-9980650 on Build GRCh38
rs745496829 Chr.12: 9980672-9980672 on Build GRCh38
rs879929758 Chr.12: 9980872-9980872 on Build GRCh38
rs201771552 Chr.12: 9980636-9980636 on Build GRCh38
rs762649944 Chr.12: 9980933-9980933 on Build GRCh38
rs990684666 Chr.12: 9980760-9980760 on Build GRCh38
rs367580037 Chr.12: 9980609-9980609 on Build GRCh38
rs916358588 Chr.12: 9980811-9980811 on Build GRCh38
rs978028772 Chr.12: 9980685-9980685 on Build GRCh38
rs894330300 Chr.12: 9980895-9980895 on Build GRCh38
rs766697200 Chr.12: 9980622-9980622 on Build GRCh38
rs771814161 Chr.12: 9980570-9980572 on Build GRCh38
rs748174264 Chr.12: 9980719-9980719 on Build GRCh38
rs527698507 Chr.12: 9980909-9980909 on Build GRCh38
rs774089563 Chr.12: 9980712-9980712 on Build GRCh38
rs889945612 Chr.12: 9980943-9980943 on Build GRCh38
rs774368524 Chr.12: 9980586-9980586 on Build GRCh38
rs765703572 Chr.12: 9980784-9980784 on Build GRCh38
rs779567838 Chr.12: 9980652-9980653 on Build GRCh38
rs750629465 Chr.12: 9980603-9980603 on Build GRCh38
rs1024234189 Chr.12: 9980945-9980945 on Build GRCh38
rs371939657 Chr.12: 9980756-9980756 on Build GRCh38
rs768511603 Chr.12: 9980697-9980697 on Build GRCh38
rs1037273318 Chr.12: 9980897-9980897 on Build GRCh38
rs761741957 Chr.12: 9980595-9980595 on Build GRCh38
rs986279097 Chr.12: 9980591-9980591 on Build GRCh38
rs779886163 Chr.12: 9980687-9980687 on Build GRCh38
rs151295066 Chr.12: 9980680-9980680 on Build GRCh38
rs903931635 Chr.12: 9980862-9980862 on Build GRCh38
rs576696196 Chr.12: 9980745-9980745 on Build GRCh38
rs576435099 Chr.12: 9980723-9980723 on Build GRCh38
rs565104657 Chr.12: 9980903-9980903 on Build GRCh38
rs945402697 Chr.12: 9980804-9980804 on Build GRCh38
rs780910855 Chr.12: 9980670-9980670 on Build GRCh38
rs1028887241 Chr.12: 9980621-9980621 on Build GRCh38
rs764763986 Chr.12: 9980813-9980813 on Build GRCh38
rs907584731 Chr.12: 9980855-9980855 on Build GRCh38
rs749044290 Chr.12: 9980689-9980689 on Build GRCh38
rs755873492 Chr.12: 9980684-9980684 on Build GRCh38
rs772022047 Chr.12: 9980731-9980731 on Build GRCh38
rs762954723 Chr.12: 9980549-9980549 on Build GRCh38
rs759747432 Chr.12: 9980773-9980773 on Build GRCh38
rs868614121 Chr.12: 9980681-9980681 on Build GRCh38
rs768438975 Chr.12: 9980584-9980584 on Build GRCh38
rs118068222 Chr.12: 9980955-9980955 on Build GRCh38
rs35776889 Chr.12: 9980599-9980599 on Build GRCh38
rs756721238 Chr.12: 9980661-9980661 on Build GRCh38
rs1049934159 Chr.12: 9980937-9980937 on Build GRCh38
rs368908261 Chr.12: 9980743-9980743 on Build GRCh38
rs993161367 Chr.12: 9980960-9980960 on Build GRCh38
rs753248286 Chr.12: 9980660-9980660 on Build GRCh38
rs773319750 Chr.12: 9980741-9980741 on Build GRCh38
rs1037243490 Chr.12: 9980596-9980596 on Build GRCh38
rs1002274315 Chr.12: 9980865-9980865 on Build GRCh38
rs1049277437 Chr.12: 9980830-9980830 on Build GRCh38
rs371694731 Chr.12: 9980645-9980645 on Build GRCh38
rs375771982 Chr.12: 9980658-9980658 on Build GRCh38
rs117182256 Chr.12: 9980879-9980879 on Build GRCh38

View Full Product Details