Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9984405-9984904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12A
Entrez Gene ID
160364
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
CD371, CLL-1, CLL1, DCAL-2, MICL
UniGene
Hs.190519
Gene Chromosome Location
Chr.12: 9951316-9995209 on Build GRCh38
Transcripts
NM_001207010.1 NM_138337.5 NM_201623.3 XM_005253324.2 XM_006719033.3 XM_006719035.3 XM_011520570.1 XM_011520571.2 XM_011520573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608404 c.691G>T p.D231Y Chr.12: 9984889-9984889 on Build GRCh38
4608405 c.661G>T p.D221Y Chr.12: 9984889-9984889 on Build GRCh38
231273 c.648A>G p.I216M Chr.12: 9984876-9984876 on Build GRCh38
2062956 c.655G>A p.A219T Chr.12: 9984883-9984883 on Build GRCh38
1476029 c.654C>T p.N218N Chr.12: 9984882-9984882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149078588 Chr.12: 9984437-9984437 on Build GRCh38
rs372644156 Chr.12: 9984511-9984511 on Build GRCh38
rs773604916 Chr.12: 9984873-9984873 on Build GRCh38
rs898229049 Chr.12: 9984707-9984710 on Build GRCh38
rs1044877123 Chr.12: 9984551-9984551 on Build GRCh38
rs907118406 Chr.12: 9984685-9984685 on Build GRCh38
rs990150184 Chr.12: 9984504-9984504 on Build GRCh38
rs1054189334 Chr.12: 9984538-9984538 on Build GRCh38
rs557114129 Chr.12: 9984443-9984443 on Build GRCh38
rs573477257 Chr.12: 9984776-9984776 on Build GRCh38
rs891833549 Chr.12: 9984605-9984605 on Build GRCh38
rs914570412 Chr.12: 9984794-9984794 on Build GRCh38
rs684134 Chr.12: 9984660-9984660 on Build GRCh38
rs476844 Chr.12: 9984697-9984697 on Build GRCh38
rs111971159 Chr.12: 9984546-9984546 on Build GRCh38
rs202219981 Chr.12: 9984844-9984844 on Build GRCh38
rs980599226 Chr.12: 9984835-9984835 on Build GRCh38
rs1028467728 Chr.12: 9984730-9984730 on Build GRCh38
rs997010816 Chr.12: 9984715-9984715 on Build GRCh38
rs912147743 Chr.12: 9984494-9984494 on Build GRCh38
rs914641536 Chr.12: 9984509-9984509 on Build GRCh38
rs1045710921 Chr.12: 9984665-9984665 on Build GRCh38
rs905046566 Chr.12: 9984580-9984580 on Build GRCh38
rs143298062 Chr.12: 9984447-9984447 on Build GRCh38
rs148321036 Chr.12: 9984815-9984815 on Build GRCh38
rs900662832 Chr.12: 9984539-9984539 on Build GRCh38
rs1039616358 Chr.12: 9984523-9984523 on Build GRCh38
rs769295853 Chr.12: 9984762-9984763 on Build GRCh38
rs990097693 Chr.12: 9984784-9984784 on Build GRCh38
rs190193785 Chr.12: 9984530-9984530 on Build GRCh38
rs924574835 Chr.12: 9984459-9984459 on Build GRCh38
rs986874062 Chr.12: 9984857-9984857 on Build GRCh38
rs962515810 Chr.12: 9984716-9984716 on Build GRCh38
rs1001425954 Chr.12: 9984695-9984695 on Build GRCh38
rs936108071 Chr.12: 9984543-9984543 on Build GRCh38
rs553342552 Chr.12: 9984702-9984702 on Build GRCh38
rs986453307 Chr.12: 9984455-9984455 on Build GRCh38
rs955730557 Chr.12: 9984752-9984752 on Build GRCh38
rs1056404891 Chr.12: 9984645-9984645 on Build GRCh38
rs557770544 Chr.12: 9984566-9984566 on Build GRCh38
rs1025360340 Chr.12: 9984777-9984777 on Build GRCh38
rs995240615 Chr.12: 9984761-9984761 on Build GRCh38

View Full Product Details