Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9997076-9997584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC1B
Entrez Gene ID
51266
Gene Name
C-type lectin domain family 1 member B
Gene Aliases
1810061I13Rik, CLEC2, CLEC2B, PRO1384, QDED721
UniGene
Hs.409794
Gene Chromosome Location
Chr.12: 9986119-10001894 on Build GRCh38
Transcripts
NM_001099431.1 NM_016509.3 XM_005253382.4 XM_011520685.2 XM_011520686.2 XM_017019395.1 XM_017019396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
360770 c.186A>G p.L62L Chr.12: 9997257-9997257 on Build GRCh38
160034 c.242G>C p.C81S Chr.12: 9997201-9997201 on Build GRCh38
6070668 c.228A>T p.L76F Chr.12: 9997215-9997215 on Build GRCh38
3667071 c.212G>A p.G71E Chr.12: 9997231-9997231 on Build GRCh38
377849 c.208A>G p.T70A Chr.12: 9997235-9997235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199500766 Chr.12: 9997325-9997325 on Build GRCh38
rs940180435 Chr.12: 9997505-9997505 on Build GRCh38
rs771971567 Chr.12: 9997166-9997166 on Build GRCh38
rs770915286 Chr.12: 9997314-9997314 on Build GRCh38
rs76073479 Chr.12: 9997503-9997503 on Build GRCh38
rs761327813 Chr.12: 9997269-9997269 on Build GRCh38
rs1054205468 Chr.12: 9997531-9997531 on Build GRCh38
rs781150132 Chr.12: 9997265-9997267 on Build GRCh38
rs753895219 Chr.12: 9997319-9997319 on Build GRCh38
rs760732267 Chr.12: 9997188-9997188 on Build GRCh38
rs778135932 Chr.12: 9997122-9997122 on Build GRCh38
rs908707499 Chr.12: 9997192-9997192 on Build GRCh38
rs200797571 Chr.12: 9997207-9997207 on Build GRCh38
rs192256546 Chr.12: 9997190-9997190 on Build GRCh38
rs1033897489 Chr.12: 9997379-9997379 on Build GRCh38
rs968384974 Chr.12: 9997402-9997402 on Build GRCh38
rs920689553 Chr.12: 9997478-9997478 on Build GRCh38
rs142926357 Chr.12: 9997347-9997347 on Build GRCh38
rs776714738 Chr.12: 9997249-9997249 on Build GRCh38
rs1004195867 Chr.12: 9997376-9997376 on Build GRCh38
rs765414250 Chr.12: 9997253-9997253 on Build GRCh38
rs748828577 Chr.12: 9997154-9997154 on Build GRCh38
rs200687802 Chr.12: 9997215-9997215 on Build GRCh38
rs199498047 Chr.12: 9997193-9997193 on Build GRCh38
rs770929569 Chr.12: 9997247-9997247 on Build GRCh38
rs769273710 Chr.12: 9997149-9997149 on Build GRCh38
rs583903 Chr.12: 9997252-9997252 on Build GRCh38
rs1022182077 Chr.12: 9997393-9997393 on Build GRCh38
rs763278564 Chr.12: 9997259-9997259 on Build GRCh38
rs776694737 Chr.12: 9997321-9997321 on Build GRCh38
rs765309726 Chr.12: 9997202-9997202 on Build GRCh38
rs950763516 Chr.12: 9997493-9997493 on Build GRCh38
rs572790397 Chr.12: 9997448-9997448 on Build GRCh38
rs944692568 Chr.12: 9997513-9997513 on Build GRCh38
rs532487075 Chr.12: 9997528-9997528 on Build GRCh38
rs777577912 Chr.12: 9997307-9997307 on Build GRCh38
rs754744736 Chr.12: 9997455-9997455 on Build GRCh38
rs980423901 Chr.12: 9997435-9997435 on Build GRCh38
rs201255744 Chr.12: 9997241-9997241 on Build GRCh38
rs776122981 Chr.12: 9997256-9997256 on Build GRCh38
rs548647712 Chr.12: 9997238-9997238 on Build GRCh38
rs921830315 Chr.12: 9997529-9997529 on Build GRCh38
rs188723247 Chr.12: 9997208-9997208 on Build GRCh38
rs1039368282 Chr.12: 9997526-9997526 on Build GRCh38
rs553040178 Chr.12: 9997357-9997357 on Build GRCh38
rs768144561 Chr.12: 9997155-9997155 on Build GRCh38
rs373866361 Chr.12: 9997224-9997224 on Build GRCh38
rs369761749 Chr.12: 9997111-9997111 on Build GRCh38
rs537562078 Chr.12: 9997275-9997275 on Build GRCh38
rs773259821 Chr.12: 9997187-9997187 on Build GRCh38
rs764333177 Chr.12: 9997265-9997265 on Build GRCh38
rs534748167 Chr.12: 9997217-9997217 on Build GRCh38
rs768480984 Chr.12: 9997305-9997305 on Build GRCh38
rs775550521 Chr.12: 9997258-9997258 on Build GRCh38
rs745864498 Chr.12: 9997323-9997323 on Build GRCh38
rs754612828 Chr.12: 9997142-9997143 on Build GRCh38
rs199596128 Chr.12: 9997100-9997100 on Build GRCh38
rs771729842 Chr.12: 9997237-9997237 on Build GRCh38
rs753029379 Chr.12: 9997205-9997205 on Build GRCh38
rs17734363 Chr.12: 9997539-9997539 on Build GRCh38
rs959529644 Chr.12: 9997400-9997400 on Build GRCh38
rs1045434726 Chr.12: 9997538-9997538 on Build GRCh38
rs199593756 Chr.12: 9997136-9997136 on Build GRCh38
rs747895438 Chr.12: 9997231-9997231 on Build GRCh38
rs584856 Chr.12: 9997470-9997470 on Build GRCh38
rs761451862 Chr.12: 9997162-9997162 on Build GRCh38
rs752032462 Chr.12: 9997209-9997209 on Build GRCh38
rs772185159 Chr.12: 9997496-9997496 on Build GRCh38
rs73050636 Chr.12: 9997348-9997348 on Build GRCh38
rs779580668 Chr.12: 9997150-9997150 on Build GRCh38
rs767948976 Chr.12: 9997273-9997273 on Build GRCh38
rs1009430839 Chr.12: 9997351-9997351 on Build GRCh38
rs764508871 Chr.12: 9997279-9997279 on Build GRCh38
rs911875494 Chr.12: 9997512-9997512 on Build GRCh38
rs748742795 Chr.12: 9997218-9997218 on Build GRCh38
rs200577312 Chr.12: 9997242-9997242 on Build GRCh38
rs935850934 Chr.12: 9997530-9997530 on Build GRCh38
rs754436234 Chr.12: 9997276-9997276 on Build GRCh38
rs184365704 Chr.12: 9997191-9997191 on Build GRCh38
rs181441498 Chr.12: 9997537-9997537 on Build GRCh38
rs774070473 Chr.12: 9997156-9997156 on Build GRCh38
rs746734460 Chr.12: 9997308-9997308 on Build GRCh38
rs778428448 Chr.12: 9997229-9997229 on Build GRCh38
rs574966728 Chr.12: 9997532-9997532 on Build GRCh38
rs750682107 Chr.12: 9997453-9997453 on Build GRCh38
rs373257247 Chr.12: 9997133-9997133 on Build GRCh38
rs751872895 Chr.12: 9997267-9997267 on Build GRCh38
rs960394255 Chr.12: 9997458-9997458 on Build GRCh38
rs956577112 Chr.12: 9997117-9997117 on Build GRCh38
rs999948129 Chr.12: 9997264-9997264 on Build GRCh38

View Full Product Details