Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10051900-10052400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC9A
Entrez Gene ID
283420
Gene Name
C-type lectin domain family 9 member A
Gene Aliases
CD370, DNGR-1, DNGR1, UNQ9341
UniGene
Hs.531189
Gene Chromosome Location
Chr.12: 10030677-10066030 on Build GRCh38
Transcripts
NM_207345.3

SNP Detail

SNP ID Chromosome Location
rs558496589 Chr.12: 10052123-10052123 on Build GRCh38
rs1009508670 Chr.12: 10052090-10052090 on Build GRCh38
rs777944635 Chr.12: 10052333-10052333 on Build GRCh38
rs756553847 Chr.12: 10052275-10052275 on Build GRCh38
rs1035366005 Chr.12: 10052329-10052329 on Build GRCh38
rs565949156 Chr.12: 10052137-10052137 on Build GRCh38
rs570090764 Chr.12: 10052052-10052052 on Build GRCh38
rs748744454 Chr.12: 10052263-10052263 on Build GRCh38
rs546543871 Chr.12: 10051976-10051980 on Build GRCh38
rs1011911573 Chr.12: 10051967-10051967 on Build GRCh38
rs890511818 Chr.12: 10052126-10052126 on Build GRCh38
rs967643656 Chr.12: 10052144-10052144 on Build GRCh38
rs908048029 Chr.12: 10052044-10052044 on Build GRCh38
rs899170451 Chr.12: 10052340-10052340 on Build GRCh38
rs147563432 Chr.12: 10052281-10052281 on Build GRCh38
rs940624329 Chr.12: 10052317-10052317 on Build GRCh38
rs111938378 Chr.12: 10052286-10052286 on Build GRCh38
rs545562856 Chr.12: 10052345-10052345 on Build GRCh38
rs1031478923 Chr.12: 10051936-10051936 on Build GRCh38
rs942218025 Chr.12: 10052063-10052066 on Build GRCh38
rs973664414 Chr.12: 10052355-10052355 on Build GRCh38
rs970160844 Chr.12: 10051992-10051992 on Build GRCh38
rs908036317 Chr.12: 10052346-10052346 on Build GRCh38
rs185748056 Chr.12: 10052343-10052343 on Build GRCh38
rs953565904 Chr.12: 10052088-10052088 on Build GRCh38
rs538820888 Chr.12: 10052121-10052121 on Build GRCh38
rs773538913 Chr.12: 10052311-10052311 on Build GRCh38
rs755325237 Chr.12: 10052039-10052039 on Build GRCh38
rs766079716 Chr.12: 10051946-10051946 on Build GRCh38
rs567631272 Chr.12: 10052007-10052007 on Build GRCh38
rs1029531356 Chr.12: 10052057-10052057 on Build GRCh38
rs921944437 Chr.12: 10052093-10052093 on Build GRCh38
rs534571120 Chr.12: 10052226-10052226 on Build GRCh38
rs1019176362 Chr.12: 10052111-10052111 on Build GRCh38
rs899226173 Chr.12: 10052027-10052027 on Build GRCh38
rs372658817 Chr.12: 10052065-10052065 on Build GRCh38
rs928396293 Chr.12: 10052036-10052036 on Build GRCh38
rs927861808 Chr.12: 10052316-10052316 on Build GRCh38
rs547800097 Chr.12: 10051947-10051947 on Build GRCh38
rs963912228 Chr.12: 10052354-10052354 on Build GRCh38
rs377607088 Chr.12: 10051952-10051952 on Build GRCh38
rs906468993 Chr.12: 10051971-10051971 on Build GRCh38
rs1038395737 Chr.12: 10052063-10052063 on Build GRCh38
rs999573826 Chr.12: 10052203-10052203 on Build GRCh38
rs1051792959 Chr.12: 10052117-10052117 on Build GRCh38
rs530141005 Chr.12: 10052014-10052014 on Build GRCh38
rs1004821358 Chr.12: 10052021-10052021 on Build GRCh38
rs192255561 Chr.12: 10052253-10052253 on Build GRCh38
rs1021287435 Chr.12: 10051975-10051975 on Build GRCh38
rs959742919 Chr.12: 10052330-10052330 on Build GRCh38
rs557010546 Chr.12: 10052310-10052310 on Build GRCh38
rs1011483938 Chr.12: 10052274-10052274 on Build GRCh38
rs983587515 Chr.12: 10052041-10052041 on Build GRCh38
rs923610584 Chr.12: 10052238-10052238 on Build GRCh38
rs186748023 Chr.12: 10052026-10052026 on Build GRCh38
rs1001582438 Chr.12: 10052299-10052299 on Build GRCh38

View Full Product Details