Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10071194-10071705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC1A
Entrez Gene ID
51267
Gene Name
C-type lectin domain family 1 member A
Gene Aliases
CLEC-1, CLEC1
UniGene
Hs.29549
Gene Chromosome Location
Chr.12: 10069504-10099065 on Build GRCh38
Transcripts
NM_001297748.1 NM_001297749.1 NM_001297750.1 NM_001297751.1 NM_016511.3 XM_011520687.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365011 c.667C>A p.H223N Chr.12: 10071509-10071509 on Build GRCh38
3782694 c.806T>C p.L269P Chr.12: 10071370-10071370 on Build GRCh38
1358208 c.761G>A p.R254H Chr.12: 10071415-10071415 on Build GRCh38
5432947 c.697A>G p.S233G Chr.12: 10071479-10071479 on Build GRCh38
1582119 c.685A>T p.T229S Chr.12: 10071491-10071491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77910243 Chr.12: 10071474-10071474 on Build GRCh38
rs765751236 Chr.12: 10071320-10071320 on Build GRCh38
rs915830593 Chr.12: 10071601-10071601 on Build GRCh38
rs766684811 Chr.12: 10071304-10071304 on Build GRCh38
rs1008472910 Chr.12: 10071678-10071678 on Build GRCh38
rs535626443 Chr.12: 10071629-10071629 on Build GRCh38
rs376070327 Chr.12: 10071330-10071330 on Build GRCh38
rs758395531 Chr.12: 10071411-10071411 on Build GRCh38
rs373331560 Chr.12: 10071496-10071496 on Build GRCh38
rs74882389 Chr.12: 10071675-10071675 on Build GRCh38
rs755758524 Chr.12: 10071382-10071382 on Build GRCh38
rs560020652 Chr.12: 10071275-10071275 on Build GRCh38
rs368197625 Chr.12: 10071244-10071244 on Build GRCh38
rs767163642 Chr.12: 10071469-10071470 on Build GRCh38
rs759380673 Chr.12: 10071490-10071490 on Build GRCh38
rs760254855 Chr.12: 10071515-10071515 on Build GRCh38
rs147882348 Chr.12: 10071415-10071415 on Build GRCh38
rs566897074 Chr.12: 10071562-10071562 on Build GRCh38
rs202247186 Chr.12: 10071521-10071521 on Build GRCh38
rs776906519 Chr.12: 10071416-10071416 on Build GRCh38
rs771148686 Chr.12: 10071361-10071361 on Build GRCh38
rs770610492 Chr.12: 10071560-10071560 on Build GRCh38
rs758214516 Chr.12: 10071497-10071497 on Build GRCh38
rs757368915 Chr.12: 10071518-10071518 on Build GRCh38
rs780216083 Chr.12: 10071540-10071540 on Build GRCh38
rs138110293 Chr.12: 10071422-10071422 on Build GRCh38
rs756784472 Chr.12: 10071328-10071328 on Build GRCh38
rs753580135 Chr.12: 10071489-10071489 on Build GRCh38
rs1031354911 Chr.12: 10071249-10071249 on Build GRCh38
rs770467406 Chr.12: 10071298-10071298 on Build GRCh38
rs910131540 Chr.12: 10071645-10071645 on Build GRCh38
rs962988265 Chr.12: 10071652-10071652 on Build GRCh38
rs763104181 Chr.12: 10071483-10071483 on Build GRCh38
rs772538258 Chr.12: 10071469-10071469 on Build GRCh38
rs370181561 Chr.12: 10071441-10071441 on Build GRCh38
rs997480502 Chr.12: 10071405-10071405 on Build GRCh38
rs751056732 Chr.12: 10071323-10071323 on Build GRCh38
rs1055695204 Chr.12: 10071574-10071574 on Build GRCh38
rs770036112 Chr.12: 10071528-10071528 on Build GRCh38
rs777828931 Chr.12: 10071508-10071508 on Build GRCh38
rs532974560 Chr.12: 10071235-10071235 on Build GRCh38
rs144671005 Chr.12: 10071402-10071402 on Build GRCh38
rs186622934 Chr.12: 10071546-10071546 on Build GRCh38
rs749978919 Chr.12: 10071378-10071378 on Build GRCh38
rs777014722 Chr.12: 10071311-10071311 on Build GRCh38
rs777650508 Chr.12: 10071412-10071412 on Build GRCh38
rs781369888 Chr.12: 10071520-10071520 on Build GRCh38
rs765283534 Chr.12: 10071284-10071284 on Build GRCh38
rs748082893 Chr.12: 10071408-10071408 on Build GRCh38
rs893719347 Chr.12: 10071498-10071498 on Build GRCh38
rs749100279 Chr.12: 10071333-10071333 on Build GRCh38
rs753671101 Chr.12: 10071400-10071400 on Build GRCh38
rs150379337 Chr.12: 10071362-10071362 on Build GRCh38
rs778092254 Chr.12: 10071227-10071227 on Build GRCh38
rs192695290 Chr.12: 10071630-10071630 on Build GRCh38
rs548539727 Chr.12: 10071345-10071345 on Build GRCh38
rs746200242 Chr.12: 10071419-10071419 on Build GRCh38
rs140826615 Chr.12: 10071346-10071346 on Build GRCh38
rs754904871 Chr.12: 10071339-10071339 on Build GRCh38
rs1028498417 Chr.12: 10071667-10071667 on Build GRCh38
rs769208893 Chr.12: 10071468-10071468 on Build GRCh38
rs890271740 Chr.12: 10071444-10071444 on Build GRCh38
rs575534504 Chr.12: 10071657-10071657 on Build GRCh38
rs1017503934 Chr.12: 10071371-10071371 on Build GRCh38
rs769029658 Chr.12: 10071545-10071545 on Build GRCh38
rs267603270 Chr.12: 10071458-10071458 on Build GRCh38
rs559643340 Chr.12: 10071680-10071680 on Build GRCh38
rs775901940 Chr.12: 10071368-10071368 on Build GRCh38
rs763510048 Chr.12: 10071357-10071357 on Build GRCh38
rs773837440 Chr.12: 10071301-10071314 on Build GRCh38
rs1038468946 Chr.12: 10071638-10071638 on Build GRCh38
rs372371337 Chr.12: 10071547-10071547 on Build GRCh38
rs759954835 Chr.12: 10071363-10071363 on Build GRCh38
rs763327379 Chr.12: 10071445-10071445 on Build GRCh38
rs969091100 Chr.12: 10071595-10071595 on Build GRCh38
rs767168660 Chr.12: 10071329-10071329 on Build GRCh38
rs779048525 Chr.12: 10071344-10071344 on Build GRCh38
rs778958795 Chr.12: 10071406-10071406 on Build GRCh38
rs371555733 Chr.12: 10071351-10071351 on Build GRCh38
rs1004693190 Chr.12: 10071632-10071632 on Build GRCh38
rs910133768 Chr.12: 10071386-10071386 on Build GRCh38
rs772081392 Chr.12: 10071349-10071349 on Build GRCh38
rs764888596 Chr.12: 10071492-10071492 on Build GRCh38
rs369664338 Chr.12: 10071294-10071294 on Build GRCh38
rs996974257 Chr.12: 10071644-10071644 on Build GRCh38
rs751589821 Chr.12: 10071513-10071513 on Build GRCh38
rs773458345 Chr.12: 10071359-10071359 on Build GRCh38
rs752181413 Chr.12: 10071477-10071478 on Build GRCh38
rs762235923 Chr.12: 10071293-10071294 on Build GRCh38
rs568326585 Chr.12: 10071375-10071375 on Build GRCh38
rs1010528420 Chr.12: 10071516-10071516 on Build GRCh38
rs749429527 Chr.12: 10071394-10071394 on Build GRCh38
rs373203262 Chr.12: 10071316-10071316 on Build GRCh38
rs898637218 Chr.12: 10071641-10071641 on Build GRCh38
rs987105621 Chr.12: 10071430-10071430 on Build GRCh38
rs949995266 Chr.12: 10071625-10071625 on Build GRCh38
rs766142843 Chr.12: 10071396-10071396 on Build GRCh38
rs999780931 Chr.12: 10071453-10071453 on Build GRCh38
rs749674889 Chr.12: 10071541-10071541 on Build GRCh38
rs747130047 Chr.12: 10071413-10071413 on Build GRCh38
rs766890740 Chr.12: 10071376-10071376 on Build GRCh38

View Full Product Details