Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10218061-10218608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABARAPL1
Entrez Gene ID
23710
Gene Name
GABA type A receptor associated protein like 1
Gene Aliases
APG8-LIKE, APG8L, ATG8, ATG8B, ATG8L, GEC1
UniGene
Hs.524250
Gene Chromosome Location
Chr.12: 10212841-10223128 on Build GRCh38
Transcripts
NM_031412.2 XM_005253344.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2064044 c.133G>A p.D45N Chr.12: 10218105-10218105 on Build GRCh38
361663 c.100G>A p.E34K Chr.12: 10218072-10218072 on Build GRCh38
4686062 c.146A>G p.Y49C Chr.12: 10218118-10218118 on Build GRCh38
4038115 c.119G>T p.R40M Chr.12: 10218091-10218091 on Build GRCh38
4038116 c.124C>T p.P42S Chr.12: 10218096-10218096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893311128 Chr.12: 10218398-10218398 on Build GRCh38
rs749513430 Chr.12: 10218096-10218096 on Build GRCh38
rs916196610 Chr.12: 10218206-10218206 on Build GRCh38
rs750357116 Chr.12: 10218189-10218189 on Build GRCh38
rs1033137790 Chr.12: 10218345-10218345 on Build GRCh38
rs902374173 Chr.12: 10218567-10218567 on Build GRCh38
rs112240800 Chr.12: 10218142-10218142 on Build GRCh38
rs763787334 Chr.12: 10218174-10218174 on Build GRCh38
rs773703548 Chr.12: 10218160-10218160 on Build GRCh38
rs1050378427 Chr.12: 10218300-10218300 on Build GRCh38
rs930523938 Chr.12: 10218265-10218265 on Build GRCh38
rs767228007 Chr.12: 10218187-10218187 on Build GRCh38
rs549900109 Chr.12: 10218488-10218488 on Build GRCh38
rs576180251 Chr.12: 10218201-10218201 on Build GRCh38
rs927749030 Chr.12: 10218234-10218234 on Build GRCh38
rs961732819 Chr.12: 10218559-10218559 on Build GRCh38
rs750838031 Chr.12: 10218165-10218165 on Build GRCh38
rs1023232636 Chr.12: 10218496-10218496 on Build GRCh38
rs1036194546 Chr.12: 10218249-10218249 on Build GRCh38
rs772352999 Chr.12: 10218156-10218156 on Build GRCh38
rs199873121 Chr.12: 10218175-10218175 on Build GRCh38
rs191909922 Chr.12: 10218299-10218299 on Build GRCh38
rs5003527 Chr.12: 10218311-10218311 on Build GRCh38
rs73058099 Chr.12: 10218164-10218164 on Build GRCh38
rs903301257 Chr.12: 10218332-10218332 on Build GRCh38
rs931036027 Chr.12: 10218348-10218348 on Build GRCh38
rs137990668 Chr.12: 10218316-10218316 on Build GRCh38
rs981778495 Chr.12: 10218534-10218534 on Build GRCh38
rs981799838 Chr.12: 10218222-10218222 on Build GRCh38
rs542420336 Chr.12: 10218166-10218166 on Build GRCh38
rs769039978 Chr.12: 10218100-10218100 on Build GRCh38
rs901257543 Chr.12: 10218107-10218107 on Build GRCh38
rs1010297490 Chr.12: 10218550-10218550 on Build GRCh38
rs143491194 Chr.12: 10218344-10218344 on Build GRCh38
rs545001290 Chr.12: 10218214-10218214 on Build GRCh38
rs920443702 Chr.12: 10218254-10218254 on Build GRCh38
rs377539572 Chr.12: 10218155-10218155 on Build GRCh38
rs552254392 Chr.12: 10218396-10218396 on Build GRCh38
rs867337221 Chr.12: 10218572-10218572 on Build GRCh38
rs374900507 Chr.12: 10218167-10218167 on Build GRCh38
rs940522880 Chr.12: 10218236-10218236 on Build GRCh38
rs959051822 Chr.12: 10218213-10218213 on Build GRCh38
rs774834736 Chr.12: 10218111-10218111 on Build GRCh38
rs969449776 Chr.12: 10218521-10218521 on Build GRCh38
rs889092690 Chr.12: 10218302-10218302 on Build GRCh38
rs780050909 Chr.12: 10218092-10218092 on Build GRCh38
rs991756443 Chr.12: 10218196-10218196 on Build GRCh38
rs116151512 Chr.12: 10218566-10218566 on Build GRCh38
rs1040988231 Chr.12: 10218537-10218538 on Build GRCh38
rs969091752 Chr.12: 10218217-10218217 on Build GRCh38
rs750150173 Chr.12: 10218354-10218354 on Build GRCh38
rs762565419 Chr.12: 10218173-10218173 on Build GRCh38
rs183987495 Chr.12: 10218514-10218514 on Build GRCh38
rs376106898 Chr.12: 10218204-10218204 on Build GRCh38
rs565154354 Chr.12: 10218232-10218232 on Build GRCh38
rs942736629 Chr.12: 10218406-10218407 on Build GRCh38
rs1034731211 Chr.12: 10218544-10218544 on Build GRCh38
rs765371487 Chr.12: 10218148-10218149 on Build GRCh38

View Full Product Details