Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10698949-10699473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YBX3
Entrez Gene ID
8531
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA, CSDA1, DBPA, ZONAB
UniGene
Hs.221889
Gene Chromosome Location
Chr.12: 10699077-10723448 on Build GRCh38
Transcripts
NM_001145426.1 NM_003651.4 XM_017020122.1 XM_017020123.1

SNP Detail

SNP ID Chromosome Location
rs182161188 Chr.12: 10699187-10699187 on Build GRCh38
rs574422614 Chr.12: 10699215-10699216 on Build GRCh38
rs931998455 Chr.12: 10699130-10699130 on Build GRCh38
rs980381871 Chr.12: 10699000-10699000 on Build GRCh38
rs144995295 Chr.12: 10699202-10699202 on Build GRCh38
rs908917735 Chr.12: 10699421-10699421 on Build GRCh38
rs933142673 Chr.12: 10699153-10699153 on Build GRCh38
rs943261547 Chr.12: 10699132-10699132 on Build GRCh38
rs760748488 Chr.12: 10698986-10698986 on Build GRCh38
rs566815448 Chr.12: 10699374-10699374 on Build GRCh38
rs565028231 Chr.12: 10699212-10699212 on Build GRCh38
rs1039361753 Chr.12: 10699133-10699133 on Build GRCh38
rs758570114 Chr.12: 10699073-10699073 on Build GRCh38
rs1022315185 Chr.12: 10698991-10698991 on Build GRCh38
rs960222780 Chr.12: 10699333-10699333 on Build GRCh38
rs1015530167 Chr.12: 10699332-10699332 on Build GRCh38
rs565350302 Chr.12: 10699170-10699170 on Build GRCh38
rs942148401 Chr.12: 10699058-10699058 on Build GRCh38
rs569374300 Chr.12: 10699422-10699422 on Build GRCh38
rs143459919 Chr.12: 10699430-10699430 on Build GRCh38
rs928978586 Chr.12: 10699075-10699075 on Build GRCh38
rs960601037 Chr.12: 10699308-10699308 on Build GRCh38
rs2166066 Chr.12: 10699059-10699059 on Build GRCh38
rs1036344170 Chr.12: 10699221-10699221 on Build GRCh38
rs1053024620 Chr.12: 10699432-10699432 on Build GRCh38
rs974715398 Chr.12: 10699423-10699423 on Build GRCh38
rs756880530 Chr.12: 10699134-10699134 on Build GRCh38
rs984866666 Chr.12: 10699408-10699408 on Build GRCh38
rs7270 Chr.12: 10699173-10699173 on Build GRCh38
rs1035319702 Chr.12: 10699278-10699279 on Build GRCh38
rs909134849 Chr.12: 10699131-10699131 on Build GRCh38
rs558104675 Chr.12: 10699014-10699014 on Build GRCh38
rs1011547603 Chr.12: 10699183-10699183 on Build GRCh38
rs199851560 Chr.12: 10699213-10699213 on Build GRCh38
rs538466176 Chr.12: 10699425-10699425 on Build GRCh38
rs984432013 Chr.12: 10699121-10699121 on Build GRCh38
rs554103407 Chr.12: 10699026-10699026 on Build GRCh38
rs15224 Chr.12: 10699397-10699397 on Build GRCh38
rs577942054 Chr.12: 10699020-10699020 on Build GRCh38
rs943936067 Chr.12: 10699156-10699156 on Build GRCh38
rs960191566 Chr.12: 10699050-10699050 on Build GRCh38
rs549119868 Chr.12: 10699410-10699410 on Build GRCh38
rs1043145 Chr.12: 10699158-10699158 on Build GRCh38
rs963565430 Chr.12: 10699364-10699364 on Build GRCh38
rs15223 Chr.12: 10699234-10699234 on Build GRCh38
rs980682636 Chr.12: 10699023-10699023 on Build GRCh38
rs918995904 Chr.12: 10699069-10699069 on Build GRCh38
rs775069536 Chr.12: 10699356-10699357 on Build GRCh38
rs921963927 Chr.12: 10699125-10699125 on Build GRCh38
rs11788 Chr.12: 10699281-10699281 on Build GRCh38
rs186322454 Chr.12: 10699445-10699445 on Build GRCh38
rs150632392 Chr.12: 10699025-10699025 on Build GRCh38
rs1052853508 Chr.12: 10699446-10699446 on Build GRCh38
rs1005131443 Chr.12: 10699317-10699317 on Build GRCh38
rs527272689 Chr.12: 10699298-10699298 on Build GRCh38
rs1001202021 Chr.12: 10699226-10699226 on Build GRCh38
rs1043053 Chr.12: 10699306-10699306 on Build GRCh38
rs752110036 Chr.12: 10699441-10699441 on Build GRCh38
rs879125735 Chr.12: 10699395-10699395 on Build GRCh38
rs547038399 Chr.12: 10699345-10699345 on Build GRCh38
rs139786389 Chr.12: 10699123-10699123 on Build GRCh38
rs972877242 Chr.12: 10699343-10699343 on Build GRCh38

View Full Product Details