Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10709771-10710293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YBX3
Entrez Gene ID
8531
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA, CSDA1, DBPA, ZONAB
UniGene
Hs.221889
Gene Chromosome Location
Chr.12: 10699077-10723448 on Build GRCh38
Transcripts
NM_001145426.1 NM_003651.4 XM_017020122.1 XM_017020123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561864 c.732G>A p.Q244Q Chr.12: 10709956-10709956 on Build GRCh38
6048437 c.679delC p.Q227fs*41 Chr.12: 10710009-10710009 on Build GRCh38
94103 c.655G>C p.A219P Chr.12: 10710033-10710033 on Build GRCh38
4038359 c.603C>T p.S201S Chr.12: 10710085-10710085 on Build GRCh38
4038352 c.669G>A p.L223L Chr.12: 10710019-10710019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747231634 Chr.12: 10709977-10709977 on Build GRCh38
rs763659772 Chr.12: 10709870-10709870 on Build GRCh38
rs778673433 Chr.12: 10709964-10709964 on Build GRCh38
rs748124096 Chr.12: 10709918-10709918 on Build GRCh38
rs755480728 Chr.12: 10710237-10710237 on Build GRCh38
rs779527951 Chr.12: 10710112-10710112 on Build GRCh38
rs905234548 Chr.12: 10709832-10709832 on Build GRCh38
rs769433743 Chr.12: 10710146-10710146 on Build GRCh38
rs535031876 Chr.12: 10709897-10709897 on Build GRCh38
rs75023229 Chr.12: 10709982-10709982 on Build GRCh38
rs752037830 Chr.12: 10710233-10710233 on Build GRCh38
rs768078848 Chr.12: 10709975-10709975 on Build GRCh38
rs765951122 Chr.12: 10710035-10710035 on Build GRCh38
rs1051287909 Chr.12: 10710058-10710059 on Build GRCh38
rs775300769 Chr.12: 10709937-10709937 on Build GRCh38
rs201917017 Chr.12: 10710065-10710065 on Build GRCh38
rs960888346 Chr.12: 10710239-10710239 on Build GRCh38
rs764939583 Chr.12: 10710049-10710049 on Build GRCh38
rs754964598 Chr.12: 10709999-10710010 on Build GRCh38
rs532948106 Chr.12: 10709811-10709811 on Build GRCh38
rs779768785 Chr.12: 10710092-10710094 on Build GRCh38
rs552270508 Chr.12: 10710197-10710197 on Build GRCh38
rs766718869 Chr.12: 10710208-10710208 on Build GRCh38
rs750967897 Chr.12: 10709871-10709871 on Build GRCh38
rs748121463 Chr.12: 10709971-10709971 on Build GRCh38
rs766772943 Chr.12: 10709859-10709859 on Build GRCh38
rs771079117 Chr.12: 10709979-10709979 on Build GRCh38
rs976522004 Chr.12: 10709945-10709945 on Build GRCh38
rs746144133 Chr.12: 10710024-10710024 on Build GRCh38
rs373462346 Chr.12: 10709920-10709920 on Build GRCh38
rs780051688 Chr.12: 10709896-10709896 on Build GRCh38
rs374011603 Chr.12: 10709974-10709974 on Build GRCh38
rs750604266 Chr.12: 10710085-10710085 on Build GRCh38
rs772781595 Chr.12: 10709989-10709989 on Build GRCh38
rs202001385 Chr.12: 10709983-10709983 on Build GRCh38
rs546593313 Chr.12: 10710138-10710138 on Build GRCh38
rs776273673 Chr.12: 10710120-10710120 on Build GRCh38
rs765605343 Chr.12: 10709938-10709938 on Build GRCh38
rs144215756 Chr.12: 10709996-10709996 on Build GRCh38
rs1021622718 Chr.12: 10710184-10710184 on Build GRCh38
rs929280330 Chr.12: 10710002-10710002 on Build GRCh38
rs1003626107 Chr.12: 10709836-10709836 on Build GRCh38
rs201350858 Chr.12: 10709980-10709980 on Build GRCh38
rs145107056 Chr.12: 10709990-10709990 on Build GRCh38
rs770331515 Chr.12: 10710119-10710119 on Build GRCh38
rs749620675 Chr.12: 10710104-10710104 on Build GRCh38
rs766730575 Chr.12: 10709829-10709831 on Build GRCh38
rs143435496 Chr.12: 10710029-10710029 on Build GRCh38
rs879241862 Chr.12: 10710133-10710134 on Build GRCh38
rs996491320 Chr.12: 10710091-10710092 on Build GRCh38
rs1045160439 Chr.12: 10709828-10709828 on Build GRCh38
rs990546340 Chr.12: 10709802-10709802 on Build GRCh38
rs200303641 Chr.12: 10709984-10709984 on Build GRCh38
rs760023584 Chr.12: 10709863-10709863 on Build GRCh38
rs775987486 Chr.12: 10710030-10710030 on Build GRCh38
rs752619424 Chr.12: 10710005-10710005 on Build GRCh38
rs112672703 Chr.12: 10710115-10710115 on Build GRCh38
rs897816131 Chr.12: 10709852-10709852 on Build GRCh38
rs145577602 Chr.12: 10710032-10710032 on Build GRCh38
rs763611652 Chr.12: 10709985-10709985 on Build GRCh38
rs773389370 Chr.12: 10709921-10709921 on Build GRCh38
rs964175437 Chr.12: 10709936-10709936 on Build GRCh38
rs771275269 Chr.12: 10709924-10709924 on Build GRCh38
rs761368552 Chr.12: 10709939-10709939 on Build GRCh38
rs775150221 Chr.12: 10710147-10710147 on Build GRCh38
rs1008107036 Chr.12: 10709922-10709922 on Build GRCh38
rs142609417 Chr.12: 10710063-10710063 on Build GRCh38
rs754821476 Chr.12: 10709959-10709959 on Build GRCh38
rs112334247 Chr.12: 10710117-10710117 on Build GRCh38
rs148867879 Chr.12: 10710033-10710033 on Build GRCh38
rs780436425 Chr.12: 10710101-10710101 on Build GRCh38
rs760609365 Chr.12: 10709946-10709946 on Build GRCh38
rs752426919 Chr.12: 10710058-10710058 on Build GRCh38
rs1056423388 Chr.12: 10709845-10709845 on Build GRCh38
rs946632013 Chr.12: 10709825-10709825 on Build GRCh38
rs1044909151 Chr.12: 10709827-10709827 on Build GRCh38
rs756841153 Chr.12: 10709874-10709874 on Build GRCh38
rs750133676 Chr.12: 10709876-10709876 on Build GRCh38
rs146058214 Chr.12: 10710014-10710014 on Build GRCh38
rs755949009 Chr.12: 10709892-10709892 on Build GRCh38
rs182510736 Chr.12: 10710015-10710015 on Build GRCh38
rs779100941 Chr.12: 10709831-10709832 on Build GRCh38
rs754350067 Chr.12: 10709860-10709860 on Build GRCh38
rs139977998 Chr.12: 10710018-10710018 on Build GRCh38
rs150841551 Chr.12: 10709940-10709940 on Build GRCh38
rs753555662 Chr.12: 10709991-10709991 on Build GRCh38
rs750014171 Chr.12: 10710059-10710059 on Build GRCh38
rs754694114 Chr.12: 10709995-10709995 on Build GRCh38
rs546627986 Chr.12: 10709830-10709830 on Build GRCh38
rs891373010 Chr.12: 10710093-10710093 on Build GRCh38
rs753515446 Chr.12: 10709953-10709953 on Build GRCh38
rs533063116 Chr.12: 10710111-10710111 on Build GRCh38
rs756291706 Chr.12: 10710095-10710095 on Build GRCh38
rs760939638 Chr.12: 10709923-10709923 on Build GRCh38
rs968402717 Chr.12: 10710166-10710166 on Build GRCh38
rs751363775 Chr.12: 10710129-10710130 on Build GRCh38
rs1021956992 Chr.12: 10710158-10710158 on Build GRCh38
rs987764502 Chr.12: 10710152-10710152 on Build GRCh38
rs139489938 Chr.12: 10709942-10709942 on Build GRCh38
rs908380470 Chr.12: 10710266-10710266 on Build GRCh38
rs761915154 Chr.12: 10710141-10710143 on Build GRCh38
rs757254428 Chr.12: 10710084-10710084 on Build GRCh38
rs759293477 Chr.12: 10710046-10710046 on Build GRCh38
rs776931399 Chr.12: 10709925-10709925 on Build GRCh38
rs992043552 Chr.12: 10710243-10710243 on Build GRCh38
rs375833820 Chr.12: 10710040-10710040 on Build GRCh38
rs764489597 Chr.12: 10710017-10710017 on Build GRCh38
rs758400154 Chr.12: 10710009-10710009 on Build GRCh38
rs377551329 Chr.12: 10709979-10709980 on Build GRCh38
rs779760100 Chr.12: 10709950-10709950 on Build GRCh38
rs767129316 Chr.12: 10709987-10709987 on Build GRCh38
rs776082214 Chr.12: 10709802-10709802 on Build GRCh38
rs566332275 Chr.12: 10709862-10709862 on Build GRCh38
rs778919347 Chr.12: 10709915-10709915 on Build GRCh38
rs750133551 Chr.12: 10709863-10709863 on Build GRCh38
rs552970132 Chr.12: 10710010-10710010 on Build GRCh38
rs369687324 Chr.12: 10709869-10709869 on Build GRCh38
rs1029364661 Chr.12: 10709947-10709947 on Build GRCh38

View Full Product Details