Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12479966-12480464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP16
Entrez Gene ID
80824
Gene Name
dual specificity phosphatase 16
Gene Aliases
MKP-7, MKP7
UniGene
Hs.536535
Gene Chromosome Location
Chr.12: 12473282-12562839 on Build GRCh38
Transcripts
NM_030640.2 XM_005253488.2 XM_006719155.2 XM_011520856.1 XM_011520857.1 XM_017019988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5840506 c.740G>A p.G247E Chr.12: 12480298-12480298 on Build GRCh38
936847 c.721C>A p.L241I Chr.12: 12480317-12480317 on Build GRCh38
6888836 c.705C>T p.A235A Chr.12: 12480333-12480333 on Build GRCh38
6650333 c.754G>A p.A252T Chr.12: 12480284-12480284 on Build GRCh38
5721904 c.712G>A p.G238R Chr.12: 12480326-12480326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775301924 Chr.12: 12480343-12480343 on Build GRCh38
rs781428998 Chr.12: 12480275-12480275 on Build GRCh38
rs755061848 Chr.12: 12480011-12480011 on Build GRCh38
rs779271793 Chr.12: 12480389-12480389 on Build GRCh38
rs1023389785 Chr.12: 12480148-12480148 on Build GRCh38
rs762630809 Chr.12: 12480345-12480345 on Build GRCh38
rs765033344 Chr.12: 12480247-12480247 on Build GRCh38
rs760082960 Chr.12: 12480220-12480220 on Build GRCh38
rs759246968 Chr.12: 12480327-12480327 on Build GRCh38
rs894013528 Chr.12: 12480404-12480404 on Build GRCh38
rs927896341 Chr.12: 12480006-12480006 on Build GRCh38
rs748648386 Chr.12: 12480304-12480304 on Build GRCh38
rs781245093 Chr.12: 12480172-12480172 on Build GRCh38
rs999808215 Chr.12: 12480103-12480103 on Build GRCh38
rs756346269 Chr.12: 12480284-12480284 on Build GRCh38
rs371480325 Chr.12: 12480325-12480325 on Build GRCh38
rs374500188 Chr.12: 12480273-12480273 on Build GRCh38
rs969513121 Chr.12: 12480154-12480154 on Build GRCh38
rs746426793 Chr.12: 12480424-12480424 on Build GRCh38
rs111441407 Chr.12: 12480065-12480065 on Build GRCh38
rs769027707 Chr.12: 12480210-12480210 on Build GRCh38
rs559342984 Chr.12: 12480130-12480130 on Build GRCh38
rs1036227119 Chr.12: 12480043-12480043 on Build GRCh38
rs369332270 Chr.12: 12480233-12480233 on Build GRCh38
rs892513755 Chr.12: 12480049-12480049 on Build GRCh38
rs901155192 Chr.12: 12480101-12480101 on Build GRCh38
rs191380076 Chr.12: 12480160-12480160 on Build GRCh38
rs543576643 Chr.12: 12480102-12480102 on Build GRCh38
rs372134887 Chr.12: 12480191-12480191 on Build GRCh38
rs192154248 Chr.12: 12480436-12480436 on Build GRCh38
rs138696462 Chr.12: 12480056-12480056 on Build GRCh38
rs186923562 Chr.12: 12480369-12480369 on Build GRCh38
rs377306900 Chr.12: 12480395-12480395 on Build GRCh38
rs986035848 Chr.12: 12480243-12480243 on Build GRCh38
rs746091774 Chr.12: 12480276-12480276 on Build GRCh38
rs779311638 Chr.12: 12480296-12480296 on Build GRCh38
rs769435110 Chr.12: 12480329-12480329 on Build GRCh38
rs769913393 Chr.12: 12480057-12480057 on Build GRCh38
rs769384819 Chr.12: 12480374-12480376 on Build GRCh38
rs1006906006 Chr.12: 12480069-12480069 on Build GRCh38
rs752478246 Chr.12: 12480252-12480252 on Build GRCh38
rs770356139 Chr.12: 12480323-12480323 on Build GRCh38
rs116234926 Chr.12: 12480416-12480416 on Build GRCh38
rs763903335 Chr.12: 12480263-12480263 on Build GRCh38
rs370998173 Chr.12: 12480405-12480405 on Build GRCh38
rs756469271 Chr.12: 12480367-12480367 on Build GRCh38
rs112572663 Chr.12: 12480351-12480351 on Build GRCh38
rs959723495 Chr.12: 12480085-12480086 on Build GRCh38
rs10845554 Chr.12: 12480016-12480016 on Build GRCh38
rs769256023 Chr.12: 12480291-12480291 on Build GRCh38
rs754066742 Chr.12: 12480382-12480382 on Build GRCh38
rs376440488 Chr.12: 12480208-12480208 on Build GRCh38
rs528429295 Chr.12: 12480364-12480364 on Build GRCh38
rs183586245 Chr.12: 12480189-12480189 on Build GRCh38
rs749641178 Chr.12: 12480289-12480289 on Build GRCh38
rs763897841 Chr.12: 12480349-12480349 on Build GRCh38
rs376852745 Chr.12: 12480285-12480285 on Build GRCh38
rs78272409 Chr.12: 12480426-12480426 on Build GRCh38
rs549253287 Chr.12: 12480272-12480272 on Build GRCh38
rs746034512 Chr.12: 12480176-12480176 on Build GRCh38
rs61758451 Chr.12: 12480270-12480270 on Build GRCh38
rs920464377 Chr.12: 12480053-12480053 on Build GRCh38
rs765864926 Chr.12: 12480222-12480222 on Build GRCh38
rs772820997 Chr.12: 12480218-12480218 on Build GRCh38
rs571936505 Chr.12: 12480387-12480387 on Build GRCh38
rs10845555 Chr.12: 12480353-12480353 on Build GRCh38
rs767506454 Chr.12: 12480359-12480359 on Build GRCh38
rs1041421488 Chr.12: 12480085-12480085 on Build GRCh38
rs147500389 Chr.12: 12480109-12480109 on Build GRCh38
rs759125027 Chr.12: 12480246-12480246 on Build GRCh38
rs770314001 Chr.12: 12480178-12480178 on Build GRCh38
rs781154712 Chr.12: 12480012-12480012 on Build GRCh38
rs185655921 Chr.12: 12480123-12480123 on Build GRCh38
rs541023115 Chr.12: 12480127-12480127 on Build GRCh38

View Full Product Details