Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12643396-12643898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBL2
Entrez Gene ID
1389
Gene Name
cAMP responsive element binding protein like 2
Gene Aliases
-
UniGene
Hs.591156
Gene Chromosome Location
Chr.12: 12611833-12645108 on Build GRCh38
Transcripts
NM_001310.3

SNP Detail

SNP ID Chromosome Location
rs182096274 Chr.12: 12643577-12643577 on Build GRCh38
rs1041196696 Chr.12: 12643855-12643855 on Build GRCh38
rs16908285 Chr.12: 12643504-12643504 on Build GRCh38
rs577675349 Chr.12: 12643496-12643496 on Build GRCh38
rs945127761 Chr.12: 12643603-12643603 on Build GRCh38
rs150823914 Chr.12: 12643608-12643608 on Build GRCh38
rs921167758 Chr.12: 12643753-12643753 on Build GRCh38
rs955331214 Chr.12: 12643790-12643790 on Build GRCh38
rs551017702 Chr.12: 12643682-12643682 on Build GRCh38
rs904683449 Chr.12: 12643625-12643625 on Build GRCh38
rs569234794 Chr.12: 12643722-12643722 on Build GRCh38
rs898460501 Chr.12: 12643466-12643466 on Build GRCh38
rs190627899 Chr.12: 12643425-12643425 on Build GRCh38
rs536272838 Chr.12: 12643759-12643759 on Build GRCh38
rs967658849 Chr.12: 12643661-12643662 on Build GRCh38
rs1056431752 Chr.12: 12643663-12643663 on Build GRCh38
rs1037032150 Chr.12: 12643442-12643442 on Build GRCh38
rs1010402725 Chr.12: 12643637-12643637 on Build GRCh38
rs559100445 Chr.12: 12643430-12643430 on Build GRCh38
rs745764038 Chr.12: 12643706-12643706 on Build GRCh38
rs778581591 Chr.12: 12643699-12643699 on Build GRCh38
rs959497312 Chr.12: 12643685-12643685 on Build GRCh38
rs139822805 Chr.12: 12643762-12643762 on Build GRCh38
rs889868262 Chr.12: 12643787-12643787 on Build GRCh38
rs530185673 Chr.12: 12643771-12643771 on Build GRCh38
rs561195552 Chr.12: 12643568-12643568 on Build GRCh38
rs986849579 Chr.12: 12643837-12643837 on Build GRCh38
rs1024325926 Chr.12: 12643697-12643697 on Build GRCh38
rs933635367 Chr.12: 12643570-12643570 on Build GRCh38
rs71755460 Chr.12: 12643688-12643692 on Build GRCh38
rs187139091 Chr.12: 12643585-12643585 on Build GRCh38
rs919940425 Chr.12: 12643741-12643742 on Build GRCh38
rs1014639180 Chr.12: 12643691-12643691 on Build GRCh38
rs779729821 Chr.12: 12643604-12643605 on Build GRCh38
rs1017009403 Chr.12: 12643720-12643720 on Build GRCh38
rs542629034 Chr.12: 12643530-12643530 on Build GRCh38
rs758692917 Chr.12: 12643665-12643666 on Build GRCh38
rs921233549 Chr.12: 12643521-12643521 on Build GRCh38
rs530485804 Chr.12: 12643516-12643516 on Build GRCh38
rs192367415 Chr.12: 12643640-12643640 on Build GRCh38
rs972614188 Chr.12: 12643714-12643714 on Build GRCh38
rs944241296 Chr.12: 12643797-12643797 on Build GRCh38
rs758294210 Chr.12: 12643848-12643848 on Build GRCh38
rs561137067 Chr.12: 12643602-12643602 on Build GRCh38
rs979608903 Chr.12: 12643674-12643674 on Build GRCh38
rs991999202 Chr.12: 12643686-12643686 on Build GRCh38
rs1050811601 Chr.12: 12643580-12643580 on Build GRCh38
rs962517522 Chr.12: 12643742-12643742 on Build GRCh38
rs975298479 Chr.12: 12643748-12643748 on Build GRCh38
rs545044150 Chr.12: 12643503-12643503 on Build GRCh38
rs1050296723 Chr.12: 12643783-12643783 on Build GRCh38
rs148879504 Chr.12: 12643584-12643584 on Build GRCh38
rs139266534 Chr.12: 12643661-12643665 on Build GRCh38

View Full Product Details