Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12821564-12822067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX47
Entrez Gene ID
51202
Gene Name
DEAD-box helicase 47
Gene Aliases
E4-DBP, HQ0256, MSTP162, RRP3
UniGene
Hs.719938
Gene Chromosome Location
Chr.12: 12813346-12829981 on Build GRCh38
Transcripts
NM_016355.3 NM_201224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4040085 c.405A>G p.Q135Q Chr.12: 12821689-12821689 on Build GRCh38
5980257 c.479C>T p.T160M Chr.12: 12822001-12822001 on Build GRCh38
5423475 c.479C>G p.T160R Chr.12: 12822001-12822001 on Build GRCh38
6641869 c.419_420insA p.P143fs*13 Chr.12: 12821703-12821704 on Build GRCh38
1360046 c.418G>T p.A140S Chr.12: 12821702-12821702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763236842 Chr.12: 12821711-12821711 on Build GRCh38
rs778762116 Chr.12: 12821629-12821629 on Build GRCh38
rs375208102 Chr.12: 12822005-12822005 on Build GRCh38
rs756363526 Chr.12: 12821947-12821947 on Build GRCh38
rs528402003 Chr.12: 12821918-12821918 on Build GRCh38
rs370792014 Chr.12: 12821761-12821761 on Build GRCh38
rs924343922 Chr.12: 12821899-12821899 on Build GRCh38
rs759209795 Chr.12: 12821768-12821768 on Build GRCh38
rs748648681 Chr.12: 12821976-12821976 on Build GRCh38
rs544765535 Chr.12: 12821838-12821838 on Build GRCh38
rs756460669 Chr.12: 12821827-12821827 on Build GRCh38
rs200563142 Chr.12: 12821908-12821909 on Build GRCh38
rs1056521586 Chr.12: 12821740-12821740 on Build GRCh38
rs1004052232 Chr.12: 12821738-12821738 on Build GRCh38
rs988302655 Chr.12: 12821909-12821909 on Build GRCh38
rs563244474 Chr.12: 12821873-12821873 on Build GRCh38
rs939334420 Chr.12: 12821966-12821966 on Build GRCh38
rs766042025 Chr.12: 12821625-12821625 on Build GRCh38
rs374718117 Chr.12: 12821944-12821944 on Build GRCh38
rs759480182 Chr.12: 12822015-12822015 on Build GRCh38
rs375771363 Chr.12: 12821678-12821678 on Build GRCh38
rs769181817 Chr.12: 12821714-12821714 on Build GRCh38
rs757275803 Chr.12: 12822042-12822042 on Build GRCh38
rs767293465 Chr.12: 12822043-12822043 on Build GRCh38
rs1045498655 Chr.12: 12821715-12821715 on Build GRCh38
rs745880212 Chr.12: 12821946-12821946 on Build GRCh38
rs761602436 Chr.12: 12821908-12821908 on Build GRCh38
rs764180410 Chr.12: 12821921-12821921 on Build GRCh38
rs199796225 Chr.12: 12821932-12821932 on Build GRCh38
rs368226365 Chr.12: 12821754-12821754 on Build GRCh38
rs931327805 Chr.12: 12821650-12821650 on Build GRCh38
rs774876536 Chr.12: 12821968-12821968 on Build GRCh38
rs78868516 Chr.12: 12821605-12821605 on Build GRCh38
rs911227584 Chr.12: 12821940-12821940 on Build GRCh38
rs770402879 Chr.12: 12821977-12821977 on Build GRCh38
rs1052639194 Chr.12: 12821639-12821639 on Build GRCh38
rs780647542 Chr.12: 12821654-12821655 on Build GRCh38
rs565158085 Chr.12: 12821986-12821986 on Build GRCh38
rs758420900 Chr.12: 12821635-12821635 on Build GRCh38
rs116740548 Chr.12: 12821656-12821656 on Build GRCh38
rs751608372 Chr.12: 12822033-12822033 on Build GRCh38
rs1017712906 Chr.12: 12821886-12821886 on Build GRCh38
rs769535432 Chr.12: 12821703-12821704 on Build GRCh38
rs780100620 Chr.12: 12821951-12821951 on Build GRCh38
rs1020714851 Chr.12: 12821785-12821785 on Build GRCh38
rs891929729 Chr.12: 12821970-12821970 on Build GRCh38
rs371547419 Chr.12: 12821941-12821941 on Build GRCh38
rs777782474 Chr.12: 12821640-12821640 on Build GRCh38
rs200512479 Chr.12: 12822002-12822002 on Build GRCh38
rs561946857 Chr.12: 12822001-12822001 on Build GRCh38
rs771117201 Chr.12: 12821660-12821660 on Build GRCh38
rs542258891 Chr.12: 12821879-12821879 on Build GRCh38
rs959310809 Chr.12: 12821871-12821871 on Build GRCh38
rs760226325 Chr.12: 12821741-12821741 on Build GRCh38
rs111772662 Chr.12: 12821874-12821874 on Build GRCh38
rs1049749034 Chr.12: 12821924-12821924 on Build GRCh38
rs915533003 Chr.12: 12821670-12821670 on Build GRCh38
rs770985836 Chr.12: 12821751-12821751 on Build GRCh38
rs946990993 Chr.12: 12821913-12821913 on Build GRCh38
rs560523469 Chr.12: 12821908-12821908 on Build GRCh38
rs774790402 Chr.12: 12821735-12821735 on Build GRCh38
rs1056956157 Chr.12: 12821969-12821969 on Build GRCh38
rs897661961 Chr.12: 12821751-12821751 on Build GRCh38
rs201410550 Chr.12: 12821628-12821628 on Build GRCh38
rs973201600 Chr.12: 12821896-12821899 on Build GRCh38
rs1009707032 Chr.12: 12821776-12821776 on Build GRCh38
rs369606390 Chr.12: 12821997-12821997 on Build GRCh38
rs781312919 Chr.12: 12821673-12821673 on Build GRCh38
rs144619888 Chr.12: 12821987-12821987 on Build GRCh38
rs200830048 Chr.12: 12821919-12821919 on Build GRCh38
rs768341957 Chr.12: 12821640-12821640 on Build GRCh38
rs1000556607 Chr.12: 12821812-12821812 on Build GRCh38
rs76984957 Chr.12: 12821609-12821609 on Build GRCh38
rs747112674 Chr.12: 12821646-12821646 on Build GRCh38
rs747937625 Chr.12: 12821657-12821659 on Build GRCh38
rs770116819 Chr.12: 12821689-12821689 on Build GRCh38
rs774480731 Chr.12: 12821760-12821761 on Build GRCh38
rs967692689 Chr.12: 12821792-12821792 on Build GRCh38
rs752632291 Chr.12: 12821634-12821634 on Build GRCh38
rs373439760 Chr.12: 12821591-12821591 on Build GRCh38
rs768985701 Chr.12: 12821963-12821963 on Build GRCh38
rs762848552 Chr.12: 12821745-12821749 on Build GRCh38
rs773153676 Chr.12: 12821739-12821757 on Build GRCh38
rs993576284 Chr.12: 12821993-12821993 on Build GRCh38
rs753505766 Chr.12: 12821627-12821627 on Build GRCh38
rs763978447 Chr.12: 12822013-12822013 on Build GRCh38
rs746101128 Chr.12: 12821687-12821687 on Build GRCh38
rs752917505 Chr.12: 12821783-12821783 on Build GRCh38
rs915397238 Chr.12: 12821910-12821910 on Build GRCh38
rs760138465 Chr.12: 12821610-12821610 on Build GRCh38
rs140772750 Chr.12: 12821702-12821702 on Build GRCh38
rs781320169 Chr.12: 12821945-12821945 on Build GRCh38
rs965718836 Chr.12: 12821891-12821891 on Build GRCh38
rs976250793 Chr.12: 12821895-12821895 on Build GRCh38

View Full Product Details