Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12824402-12824911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX47
Entrez Gene ID
51202
Gene Name
DEAD-box helicase 47
Gene Aliases
E4-DBP, HQ0256, MSTP162, RRP3
UniGene
Hs.719938
Gene Chromosome Location
Chr.12: 12813346-12829981 on Build GRCh38
Transcripts
NM_016355.3 NM_201224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468004 c.937G>T p.A313S Chr.12: 12824579-12824579 on Build GRCh38
1360049 c.905G>A p.R302H Chr.12: 12824547-12824547 on Build GRCh38
227325 c.1021C>T p.P341S Chr.12: 12824663-12824663 on Build GRCh38
7400739 c.1009A>G p.N337D Chr.12: 12824651-12824651 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537851583 Chr.12: 12824794-12824794 on Build GRCh38
rs373915147 Chr.12: 12824687-12824687 on Build GRCh38
rs374732094 Chr.12: 12824793-12824793 on Build GRCh38
rs373242027 Chr.12: 12824606-12824606 on Build GRCh38
rs1004113194 Chr.12: 12824817-12824817 on Build GRCh38
rs201910057 Chr.12: 12824605-12824605 on Build GRCh38
rs750046911 Chr.12: 12824565-12824565 on Build GRCh38
rs777068535 Chr.12: 12824493-12824493 on Build GRCh38
rs556188211 Chr.12: 12824802-12824802 on Build GRCh38
rs1006339928 Chr.12: 12824584-12824584 on Build GRCh38
rs962800922 Chr.12: 12824458-12824458 on Build GRCh38
rs778178055 Chr.12: 12824612-12824612 on Build GRCh38
rs754375134 Chr.12: 12824567-12824567 on Build GRCh38
rs200857940 Chr.12: 12824670-12824670 on Build GRCh38
rs370055269 Chr.12: 12824596-12824596 on Build GRCh38
rs761267089 Chr.12: 12824547-12824547 on Build GRCh38
rs779886782 Chr.12: 12824591-12824591 on Build GRCh38
rs770114327 Chr.12: 12824744-12824744 on Build GRCh38
rs377300940 Chr.12: 12824630-12824630 on Build GRCh38
rs754563602 Chr.12: 12824882-12824882 on Build GRCh38
rs371959457 Chr.12: 12824692-12824692 on Build GRCh38
rs569098116 Chr.12: 12824686-12824686 on Build GRCh38
rs777061310 Chr.12: 12824621-12824621 on Build GRCh38
rs781226586 Chr.12: 12824728-12824728 on Build GRCh38
rs146075815 Chr.12: 12824832-12824832 on Build GRCh38
rs975167936 Chr.12: 12824465-12824465 on Build GRCh38
rs957240389 Chr.12: 12824712-12824712 on Build GRCh38
rs377764365 Chr.12: 12824594-12824594 on Build GRCh38
rs929306510 Chr.12: 12824861-12824861 on Build GRCh38
rs760106173 Chr.12: 12824520-12824520 on Build GRCh38
rs983452391 Chr.12: 12824864-12824864 on Build GRCh38
rs765599134 Chr.12: 12824523-12824523 on Build GRCh38
rs142329297 Chr.12: 12824652-12824652 on Build GRCh38
rs187363680 Chr.12: 12824446-12824446 on Build GRCh38
rs975577051 Chr.12: 12824868-12824868 on Build GRCh38
rs1055846882 Chr.12: 12824666-12824666 on Build GRCh38
rs760386675 Chr.12: 12824668-12824668 on Build GRCh38
rs780850943 Chr.12: 12824697-12824698 on Build GRCh38
rs777984556 Chr.12: 12824705-12824705 on Build GRCh38
rs982478387 Chr.12: 12824440-12824440 on Build GRCh38
rs951304269 Chr.12: 12824759-12824759 on Build GRCh38
rs190667675 Chr.12: 12824549-12824549 on Build GRCh38
rs757532709 Chr.12: 12824723-12824723 on Build GRCh38
rs751105557 Chr.12: 12824546-12824546 on Build GRCh38
rs79126384 Chr.12: 12824885-12824885 on Build GRCh38
rs990016463 Chr.12: 12824461-12824461 on Build GRCh38
rs771003969 Chr.12: 12824616-12824616 on Build GRCh38
rs775954244 Chr.12: 12824635-12824635 on Build GRCh38
rs755931822 Chr.12: 12824582-12824582 on Build GRCh38
rs1010878 Chr.12: 12824466-12824466 on Build GRCh38
rs200518902 Chr.12: 12824691-12824691 on Build GRCh38
rs566247945 Chr.12: 12824615-12824615 on Build GRCh38
rs760842816 Chr.12: 12824430-12824431 on Build GRCh38
rs772908860 Chr.12: 12824662-12824662 on Build GRCh38
rs778954535 Chr.12: 12824603-12824603 on Build GRCh38
rs1045832799 Chr.12: 12824476-12824476 on Build GRCh38
rs754679587 Chr.12: 12824671-12824671 on Build GRCh38
rs758616133 Chr.12: 12824693-12824693 on Build GRCh38
rs1000700016 Chr.12: 12824659-12824659 on Build GRCh38
rs969930936 Chr.12: 12824782-12824782 on Build GRCh38
rs562084101 Chr.12: 12824485-12824485 on Build GRCh38
rs769095775 Chr.12: 12824757-12824757 on Build GRCh38
rs746297459 Chr.12: 12824731-12824731 on Build GRCh38
rs749783872 Chr.12: 12824751-12824751 on Build GRCh38
rs137865216 Chr.12: 12824638-12824638 on Build GRCh38
rs776044052 Chr.12: 12824747-12824747 on Build GRCh38
rs375365365 Chr.12: 12824464-12824464 on Build GRCh38
rs765879487 Chr.12: 12824669-12824669 on Build GRCh38
rs145266858 Chr.12: 12824583-12824583 on Build GRCh38
rs746997685 Chr.12: 12824715-12824715 on Build GRCh38
rs535729055 Chr.12: 12824862-12824862 on Build GRCh38
rs746182295 Chr.12: 12824627-12824627 on Build GRCh38
rs767704629 Chr.12: 12824696-12824696 on Build GRCh38

View Full Product Details