Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14434239-14434761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP
Entrez Gene ID
55729
Gene Name
activating transcription factor 7 interacting protein
Gene Aliases
AM, ATF-IP, MCAF, MCAF1, p621
UniGene
Hs.504856
Gene Chromosome Location
Chr.12: 14365632-14502935 on Build GRCh38
Transcripts
NM_001286514.1 NM_001286515.1 NM_018179.4 NM_181352.1 XM_005253424.3 XM_006719108.3 XM_006719109.3 XM_011520754.2 XM_011520755.2 XM_011520756.2 XM_017019638.1 XM_017019639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777968 c.1586A>G p.K529R Chr.12: 14434367-14434367 on Build GRCh38
6553055 c.1631C>T p.P544L Chr.12: 14434412-14434412 on Build GRCh38
6462437 c.1640A>G p.E547G Chr.12: 14434418-14434418 on Build GRCh38
937499 c.1593T>C p.P531P Chr.12: 14434371-14434371 on Build GRCh38
1128636 c.1589A>G p.K530R Chr.12: 14434367-14434367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008200515 Chr.12: 14434566-14434566 on Build GRCh38
rs752237315 Chr.12: 14434467-14434467 on Build GRCh38
rs556322838 Chr.12: 14434351-14434351 on Build GRCh38
rs151223624 Chr.12: 14434419-14434419 on Build GRCh38
rs1019715642 Chr.12: 14434568-14434568 on Build GRCh38
rs762280482 Chr.12: 14434333-14434333 on Build GRCh38
rs766105973 Chr.12: 14434386-14434386 on Build GRCh38
rs769937687 Chr.12: 14434290-14434290 on Build GRCh38
rs764643907 Chr.12: 14434323-14434323 on Build GRCh38
rs763718127 Chr.12: 14434700-14434700 on Build GRCh38
rs558163255 Chr.12: 14434491-14434491 on Build GRCh38
rs752325672 Chr.12: 14434382-14434382 on Build GRCh38
rs201782951 Chr.12: 14434429-14434429 on Build GRCh38
rs145246872 Chr.12: 14434364-14434364 on Build GRCh38
rs3213764 Chr.12: 14434367-14434367 on Build GRCh38
rs765045149 Chr.12: 14434461-14434461 on Build GRCh38
rs769122316 Chr.12: 14434431-14434431 on Build GRCh38
rs559708687 Chr.12: 14434628-14434628 on Build GRCh38
rs142252576 Chr.12: 14434383-14434383 on Build GRCh38
rs999644201 Chr.12: 14434267-14434267 on Build GRCh38
rs12815453 Chr.12: 14434513-14434513 on Build GRCh38
rs200546272 Chr.12: 14434502-14434503 on Build GRCh38
rs769696154 Chr.12: 14434404-14434404 on Build GRCh38
rs774676266 Chr.12: 14434329-14434329 on Build GRCh38
rs898645264 Chr.12: 14434515-14434515 on Build GRCh38
rs1025595611 Chr.12: 14434719-14434719 on Build GRCh38
rs576603428 Chr.12: 14434492-14434492 on Build GRCh38
rs368031485 Chr.12: 14434474-14434474 on Build GRCh38
rs184213558 Chr.12: 14434537-14434537 on Build GRCh38
rs752401259 Chr.12: 14434545-14434545 on Build GRCh38
rs767337598 Chr.12: 14434526-14434526 on Build GRCh38
rs924498957 Chr.12: 14434710-14434710 on Build GRCh38
rs761387457 Chr.12: 14434460-14434462 on Build GRCh38
rs1053865020 Chr.12: 14434564-14434564 on Build GRCh38
rs751275460 Chr.12: 14434456-14434461 on Build GRCh38
rs951258392 Chr.12: 14434729-14434735 on Build GRCh38
rs571905678 Chr.12: 14434399-14434399 on Build GRCh38
rs372036691 Chr.12: 14434369-14434369 on Build GRCh38
rs757104709 Chr.12: 14434394-14434394 on Build GRCh38
rs3213765 Chr.12: 14434678-14434678 on Build GRCh38
rs370379417 Chr.12: 14434355-14434355 on Build GRCh38
rs763454677 Chr.12: 14434316-14434316 on Build GRCh38
rs759217971 Chr.12: 14434460-14434460 on Build GRCh38
rs948388659 Chr.12: 14434490-14434490 on Build GRCh38
rs375070737 Chr.12: 14434395-14434395 on Build GRCh38
rs113216097 Chr.12: 14434636-14434636 on Build GRCh38
rs762036579 Chr.12: 14434439-14434439 on Build GRCh38
rs1052004152 Chr.12: 14434522-14434522 on Build GRCh38
rs775917225 Chr.12: 14434310-14434310 on Build GRCh38
rs746767096 Chr.12: 14434387-14434387 on Build GRCh38
rs1004354888 Chr.12: 14434494-14434494 on Build GRCh38
rs894166797 Chr.12: 14434480-14434480 on Build GRCh38
rs1012381485 Chr.12: 14434574-14434574 on Build GRCh38
rs746113846 Chr.12: 14434396-14434396 on Build GRCh38
rs776083974 Chr.12: 14434456-14434456 on Build GRCh38
rs767941425 Chr.12: 14434452-14434452 on Build GRCh38
rs1044745156 Chr.12: 14434617-14434617 on Build GRCh38
rs1028351607 Chr.12: 14434278-14434278 on Build GRCh38
rs541083326 Chr.12: 14434606-14434606 on Build GRCh38
rs562297113 Chr.12: 14434632-14434632 on Build GRCh38
rs552493657 Chr.12: 14434738-14434738 on Build GRCh38
rs189018238 Chr.12: 14434538-14434538 on Build GRCh38
rs1005406279 Chr.12: 14434730-14434730 on Build GRCh38
rs780765215 Chr.12: 14434473-14434474 on Build GRCh38
rs1025485985 Chr.12: 14434432-14434432 on Build GRCh38

View Full Product Details