Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14446718-14447215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP
Entrez Gene ID
55729
Gene Name
activating transcription factor 7 interacting protein
Gene Aliases
AM, ATF-IP, MCAF, MCAF1, p621
UniGene
Hs.504856
Gene Chromosome Location
Chr.12: 14365632-14502935 on Build GRCh38
Transcripts
NM_001286514.1 NM_001286515.1 NM_018179.4 NM_181352.1 XM_005253424.3 XM_006719108.3 XM_006719109.3 XM_011520754.2 XM_011520755.2 XM_011520756.2 XM_017019638.1 XM_017019639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515467 c.1985G>T p.R662I Chr.12: 14447046-14447046 on Build GRCh38
6014857 c.1930G>T p.A644S Chr.12: 14446988-14446988 on Build GRCh38
6284790 c.1968C>A p.A656A Chr.12: 14447026-14447026 on Build GRCh38
6370817 c.1945A>G p.T649A Chr.12: 14447006-14447006 on Build GRCh38
2153895 c.1975G>T p.D659Y Chr.12: 14447033-14447033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200558203 Chr.12: 14447091-14447091 on Build GRCh38
rs764087216 Chr.12: 14446944-14446944 on Build GRCh38
rs796095066 Chr.12: 14446971-14446972 on Build GRCh38
rs377536644 Chr.12: 14447075-14447075 on Build GRCh38
rs764436591 Chr.12: 14447073-14447073 on Build GRCh38
rs754256192 Chr.12: 14447150-14447150 on Build GRCh38
rs768878081 Chr.12: 14446980-14446981 on Build GRCh38
rs778569644 Chr.12: 14446993-14446993 on Build GRCh38
rs994657868 Chr.12: 14447188-14447188 on Build GRCh38
rs539053096 Chr.12: 14446971-14446971 on Build GRCh38
rs1039194192 Chr.12: 14447171-14447171 on Build GRCh38
rs150124510 Chr.12: 14447014-14447014 on Build GRCh38
rs1045789553 Chr.12: 14447119-14447119 on Build GRCh38
rs764957949 Chr.12: 14447058-14447058 on Build GRCh38
rs761145954 Chr.12: 14446973-14446974 on Build GRCh38
rs775816811 Chr.12: 14446971-14446973 on Build GRCh38
rs902378328 Chr.12: 14446905-14446905 on Build GRCh38
rs879045182 Chr.12: 14446831-14446831 on Build GRCh38
rs936149447 Chr.12: 14446922-14446922 on Build GRCh38
rs576897520 Chr.12: 14447167-14447167 on Build GRCh38
rs397735482 Chr.12: 14446984-14446985 on Build GRCh38
rs756456015 Chr.12: 14446963-14446963 on Build GRCh38
rs1055129060 Chr.12: 14446927-14446927 on Build GRCh38
rs181206739 Chr.12: 14446937-14446937 on Build GRCh38
rs766583343 Chr.12: 14447070-14447070 on Build GRCh38
rs771635172 Chr.12: 14446757-14446757 on Build GRCh38
rs777816775 Chr.12: 14446969-14446970 on Build GRCh38
rs754284471 Chr.12: 14447104-14447104 on Build GRCh38
rs373695319 Chr.12: 14446970-14446970 on Build GRCh38
rs200891089 Chr.12: 14446985-14446985 on Build GRCh38
rs183653258 Chr.12: 14446845-14446845 on Build GRCh38
rs928846700 Chr.12: 14446799-14446799 on Build GRCh38
rs747757826 Chr.12: 14446997-14446997 on Build GRCh38
rs544088884 Chr.12: 14446922-14446922 on Build GRCh38
rs770819685 Chr.12: 14447042-14447042 on Build GRCh38
rs757826003 Chr.12: 14446947-14446947 on Build GRCh38
rs1027545824 Chr.12: 14446866-14446866 on Build GRCh38
rs761255181 Chr.12: 14446849-14446853 on Build GRCh38
rs887498551 Chr.12: 14446842-14446842 on Build GRCh38
rs566466245 Chr.12: 14447067-14447067 on Build GRCh38
rs982093555 Chr.12: 14446781-14446781 on Build GRCh38
rs536040153 Chr.12: 14447005-14447005 on Build GRCh38
rs948930281 Chr.12: 14447092-14447092 on Build GRCh38
rs1037087383 Chr.12: 14447173-14447173 on Build GRCh38
rs369795486 Chr.12: 14446906-14446906 on Build GRCh38
rs1038656558 Chr.12: 14446869-14446869 on Build GRCh38
rs911545868 Chr.12: 14446896-14446896 on Build GRCh38
rs961868098 Chr.12: 14446842-14446842 on Build GRCh38
rs746194923 Chr.12: 14446950-14446950 on Build GRCh38
rs897668459 Chr.12: 14447176-14447176 on Build GRCh38
rs112974161 Chr.12: 14446932-14446932 on Build GRCh38
rs957404302 Chr.12: 14446917-14446917 on Build GRCh38
rs985784242 Chr.12: 14446874-14446874 on Build GRCh38
rs908967223 Chr.12: 14447153-14447154 on Build GRCh38
rs538573327 Chr.12: 14446971-14446972 on Build GRCh38
rs780275692 Chr.12: 14447099-14447099 on Build GRCh38
rs5796595 Chr.12: 14446970-14446971 on Build GRCh38
rs189250102 Chr.12: 14446912-14446912 on Build GRCh38
rs1004337278 Chr.12: 14447169-14447169 on Build GRCh38
rs750757439 Chr.12: 14447132-14447133 on Build GRCh38
rs751757585 Chr.12: 14446946-14446946 on Build GRCh38
rs192539043 Chr.12: 14446835-14446835 on Build GRCh38
rs948964720 Chr.12: 14446771-14446771 on Build GRCh38
rs762024704 Chr.12: 14447085-14447085 on Build GRCh38
rs557269876 Chr.12: 14447012-14447012 on Build GRCh38
rs137919163 Chr.12: 14446949-14446949 on Build GRCh38
rs953069456 Chr.12: 14446873-14446873 on Build GRCh38
rs950897147 Chr.12: 14447126-14447126 on Build GRCh38
rs759330230 Chr.12: 14447049-14447049 on Build GRCh38
rs977138387 Chr.12: 14446910-14446910 on Build GRCh38
rs768088286 Chr.12: 14447090-14447090 on Build GRCh38
rs776585263 Chr.12: 14447044-14447044 on Build GRCh38
rs983558798 Chr.12: 14447153-14447153 on Build GRCh38
rs755825016 Chr.12: 14446968-14446969 on Build GRCh38
rs929457106 Chr.12: 14446813-14446813 on Build GRCh38
rs756432909 Chr.12: 14447098-14447098 on Build GRCh38
rs771821995 Chr.12: 14447000-14447000 on Build GRCh38

View Full Product Details