Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14672731-14673242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_004963.3 XM_011520631.2

Gene Symbol
GUCY2C
Entrez Gene ID
2984
Gene Name
guanylate cyclase 2C
Gene Aliases
DIAR6, GUC2C, MECIL, MUCIL, STAR
UniGene
Hs.524278
Gene Chromosome Location
Chr.12: 14612632-14696625 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_004963.3 XM_011520631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223524 c.1085G>A p.G362E Chr.12: 14672958-14672958 on Build GRCh38
5986511 c.1085-7_1085-6insGGACA p.? Chr.12: 14672964-14672965 on Build GRCh38
6857326 c.1148A>G p.Y383C Chr.12: 14672895-14672895 on Build GRCh38
5840646 c.1094G>A p.G365D Chr.12: 14672949-14672949 on Build GRCh38
937535 c.1170+1G>T p.? Chr.12: 14672872-14672872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751020136 Chr.12: 14672768-14672768 on Build GRCh38
rs113458749 Chr.12: 14673053-14673053 on Build GRCh38
rs376604442 Chr.12: 14672910-14672910 on Build GRCh38
rs977011577 Chr.12: 14672778-14672778 on Build GRCh38
rs372300028 Chr.12: 14672868-14672868 on Build GRCh38
rs1027216857 Chr.12: 14672959-14672959 on Build GRCh38
rs781247649 Chr.12: 14672940-14672940 on Build GRCh38
rs762254572 Chr.12: 14672953-14672953 on Build GRCh38
rs560213516 Chr.12: 14672757-14672757 on Build GRCh38
rs764796248 Chr.12: 14672887-14672887 on Build GRCh38
rs2287170 Chr.12: 14673037-14673037 on Build GRCh38
rs754955870 Chr.12: 14672871-14672871 on Build GRCh38
rs769668898 Chr.12: 14672946-14672946 on Build GRCh38
rs775846475 Chr.12: 14672866-14672866 on Build GRCh38
rs531167526 Chr.12: 14672806-14672806 on Build GRCh38
rs925889993 Chr.12: 14673045-14673045 on Build GRCh38
rs543817365 Chr.12: 14673026-14673026 on Build GRCh38
rs757273773 Chr.12: 14672939-14672939 on Build GRCh38
rs762336239 Chr.12: 14672875-14672875 on Build GRCh38
rs752242360 Chr.12: 14672977-14672977 on Build GRCh38
rs375279933 Chr.12: 14672882-14672882 on Build GRCh38
rs770802438 Chr.12: 14672830-14672830 on Build GRCh38
rs781123886 Chr.12: 14673188-14673188 on Build GRCh38
rs937028293 Chr.12: 14672917-14672917 on Build GRCh38
rs751808848 Chr.12: 14673050-14673050 on Build GRCh38
rs141094437 Chr.12: 14672858-14672858 on Build GRCh38
rs749532194 Chr.12: 14672950-14672950 on Build GRCh38
rs973179353 Chr.12: 14672804-14672804 on Build GRCh38
rs953155551 Chr.12: 14673189-14673189 on Build GRCh38
rs368359900 Chr.12: 14672863-14672863 on Build GRCh38
rs897309666 Chr.12: 14673195-14673195 on Build GRCh38
rs566095994 Chr.12: 14673218-14673218 on Build GRCh38
rs375970677 Chr.12: 14672979-14672979 on Build GRCh38
rs2287171 Chr.12: 14673022-14673022 on Build GRCh38
rs550888916 Chr.12: 14673079-14673079 on Build GRCh38
rs1003497218 Chr.12: 14673064-14673064 on Build GRCh38
rs766818659 Chr.12: 14672976-14672976 on Build GRCh38
rs746904140 Chr.12: 14672928-14672928 on Build GRCh38
rs779816104 Chr.12: 14672904-14672905 on Build GRCh38
rs769892940 Chr.12: 14672864-14672864 on Build GRCh38
rs544933759 Chr.12: 14672951-14672951 on Build GRCh38
rs895568785 Chr.12: 14672922-14672922 on Build GRCh38
rs147742759 Chr.12: 14672919-14672919 on Build GRCh38
rs1036109163 Chr.12: 14673148-14673149 on Build GRCh38
rs745482074 Chr.12: 14672860-14672860 on Build GRCh38
rs767807024 Chr.12: 14672877-14672877 on Build GRCh38
rs985744836 Chr.12: 14673210-14673210 on Build GRCh38
rs587776905 Chr.12: 14672883-14672883 on Build GRCh38
rs961910918 Chr.12: 14673148-14673148 on Build GRCh38
rs752435021 Chr.12: 14672890-14672890 on Build GRCh38
rs371683641 Chr.12: 14672975-14672975 on Build GRCh38
rs757960893 Chr.12: 14672891-14672891 on Build GRCh38
rs758284597 Chr.12: 14672876-14672876 on Build GRCh38
rs767899334 Chr.12: 14672958-14672958 on Build GRCh38
rs776294687 Chr.12: 14672857-14672857 on Build GRCh38
rs151144116 Chr.12: 14672760-14672760 on Build GRCh38
rs369342368 Chr.12: 14672968-14672968 on Build GRCh38
rs936994435 Chr.12: 14673054-14673054 on Build GRCh38
rs991565273 Chr.12: 14673069-14673069 on Build GRCh38
rs973532018 Chr.12: 14673150-14673150 on Build GRCh38
rs949830598 Chr.12: 14673119-14673119 on Build GRCh38
rs552241443 Chr.12: 14672942-14672942 on Build GRCh38
rs573384164 Chr.12: 14672775-14672775 on Build GRCh38
rs552864042 Chr.12: 14672905-14672905 on Build GRCh38
rs757607029 Chr.12: 14673051-14673051 on Build GRCh38
rs920374966 Chr.12: 14673178-14673178 on Build GRCh38
rs746745578 Chr.12: 14672922-14672923 on Build GRCh38
rs965851060 Chr.12: 14673015-14673015 on Build GRCh38
rs750770871 Chr.12: 14672880-14672880 on Build GRCh38
rs752009818 Chr.12: 14672869-14672869 on Build GRCh38
rs913116348 Chr.12: 14672784-14672785 on Build GRCh38
rs11833202 Chr.12: 14673031-14673031 on Build GRCh38
rs201135486 Chr.12: 14672949-14672949 on Build GRCh38
rs763638677 Chr.12: 14672985-14672985 on Build GRCh38

View Full Product Details