Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14805760-14806271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
NM_175874.3 XM_005253322.4 XM_017018871.1 XM_017018872.1 XM_017018873.1 XM_017018874.1 NM_001013698.2 XM_017019312.1

Gene Symbol
SMCO3
Entrez Gene ID
440087
Gene Name
single-pass membrane protein with coiled-coil domains 3
Gene Aliases
C12orf69
UniGene
Hs.220931
Gene Chromosome Location
Chr.12: 14804650-14816761 on Build GRCh38
Transcripts
NM_175874.3 XM_005253322.4 XM_017018871.1 XM_017018872.1 XM_017018873.1 XM_017018874.1 NM_001013698.2 XM_017019312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4040581 c.624A>C p.K208N Chr.12: 14806057-14806057 on Build GRCh38
6136250 c.470C>A p.A157D Chr.12: 14806211-14806211 on Build GRCh38
4454196 c.432A>G p.I144M Chr.12: 14806249-14806249 on Build GRCh38
6318691 c.425C>A p.T142N Chr.12: 14806256-14806256 on Build GRCh38
1360423 c.530G>A p.R177H Chr.12: 14806151-14806151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75463413 Chr.12: 14806140-14806140 on Build GRCh38
rs369424819 Chr.12: 14805996-14805996 on Build GRCh38
rs375827971 Chr.12: 14806004-14806004 on Build GRCh38
rs768362318 Chr.12: 14806165-14806166 on Build GRCh38
rs568291123 Chr.12: 14805895-14805895 on Build GRCh38
rs765037681 Chr.12: 14805982-14805982 on Build GRCh38
rs1011365294 Chr.12: 14805973-14805973 on Build GRCh38
rs777248662 Chr.12: 14806151-14806151 on Build GRCh38
rs745444977 Chr.12: 14806160-14806160 on Build GRCh38
rs539521431 Chr.12: 14806202-14806202 on Build GRCh38
rs762068540 Chr.12: 14806217-14806217 on Build GRCh38
rs770717856 Chr.12: 14805956-14805956 on Build GRCh38
rs376267349 Chr.12: 14805990-14805990 on Build GRCh38
rs780721771 Chr.12: 14806157-14806157 on Build GRCh38
rs77248584 Chr.12: 14806002-14806002 on Build GRCh38
rs530744026 Chr.12: 14805997-14805997 on Build GRCh38
rs576597535 Chr.12: 14805875-14805875 on Build GRCh38
rs372371636 Chr.12: 14805824-14805824 on Build GRCh38
rs775312369 Chr.12: 14805989-14805989 on Build GRCh38
rs372866941 Chr.12: 14806001-14806001 on Build GRCh38
rs892448704 Chr.12: 14805969-14805969 on Build GRCh38
rs982683899 Chr.12: 14805786-14805786 on Build GRCh38
rs771578940 Chr.12: 14806047-14806047 on Build GRCh38
rs1053816710 Chr.12: 14805791-14805791 on Build GRCh38
rs761752523 Chr.12: 14806097-14806097 on Build GRCh38
rs750575247 Chr.12: 14806137-14806137 on Build GRCh38
rs374450426 Chr.12: 14805842-14805842 on Build GRCh38
rs562961859 Chr.12: 14806193-14806193 on Build GRCh38
rs767051517 Chr.12: 14806139-14806139 on Build GRCh38
rs762791212 Chr.12: 14806095-14806095 on Build GRCh38
rs540670046 Chr.12: 14805927-14805927 on Build GRCh38
rs755400649 Chr.12: 14806148-14806148 on Build GRCh38
rs779977624 Chr.12: 14806187-14806187 on Build GRCh38
rs777497549 Chr.12: 14806030-14806030 on Build GRCh38
rs376938990 Chr.12: 14806086-14806086 on Build GRCh38
rs769702138 Chr.12: 14806169-14806169 on Build GRCh38
rs767616071 Chr.12: 14806131-14806131 on Build GRCh38
rs759286011 Chr.12: 14805970-14805970 on Build GRCh38
rs775970639 Chr.12: 14805896-14805896 on Build GRCh38
rs924087338 Chr.12: 14806022-14806022 on Build GRCh38
rs760359895 Chr.12: 14806109-14806111 on Build GRCh38
rs370727838 Chr.12: 14806005-14806005 on Build GRCh38
rs761060859 Chr.12: 14806048-14806050 on Build GRCh38
rs947998742 Chr.12: 14806049-14806049 on Build GRCh38
rs745783395 Chr.12: 14806072-14806072 on Build GRCh38
rs36108893 Chr.12: 14806130-14806131 on Build GRCh38
rs543703519 Chr.12: 14805926-14805926 on Build GRCh38
rs769758771 Chr.12: 14806081-14806081 on Build GRCh38
rs774668127 Chr.12: 14806096-14806096 on Build GRCh38
rs927264482 Chr.12: 14806073-14806073 on Build GRCh38
rs558247382 Chr.12: 14806236-14806236 on Build GRCh38
rs57839882 Chr.12: 14805952-14805952 on Build GRCh38
rs867109281 Chr.12: 14805831-14805831 on Build GRCh38
rs756685601 Chr.12: 14806138-14806138 on Build GRCh38
rs75082633 Chr.12: 14805918-14805918 on Build GRCh38
rs532916993 Chr.12: 14805951-14805951 on Build GRCh38
rs1007103786 Chr.12: 14805975-14805975 on Build GRCh38
rs34678546 Chr.12: 14806239-14806240 on Build GRCh38
rs199648611 Chr.12: 14806026-14806026 on Build GRCh38
rs893790479 Chr.12: 14805795-14805795 on Build GRCh38
rs746435652 Chr.12: 14806037-14806037 on Build GRCh38
rs555649645 Chr.12: 14805914-14805914 on Build GRCh38
rs1045083473 Chr.12: 14806060-14806060 on Build GRCh38
rs372237546 Chr.12: 14806189-14806189 on Build GRCh38
rs780538890 Chr.12: 14806016-14806016 on Build GRCh38
rs776633171 Chr.12: 14805964-14805964 on Build GRCh38
rs941050122 Chr.12: 14805829-14805829 on Build GRCh38
rs776383564 Chr.12: 14806053-14806053 on Build GRCh38
rs1050849792 Chr.12: 14806170-14806170 on Build GRCh38
rs61732281 Chr.12: 14806155-14806155 on Build GRCh38
rs768521827 Chr.12: 14806198-14806198 on Build GRCh38
rs1038103200 Chr.12: 14805905-14805905 on Build GRCh38
rs896929868 Chr.12: 14805930-14805930 on Build GRCh38
rs373027449 Chr.12: 14806152-14806152 on Build GRCh38
rs563407389 Chr.12: 14805935-14805935 on Build GRCh38
rs558097753 Chr.12: 14805788-14805788 on Build GRCh38
rs878901658 Chr.12: 14805903-14805903 on Build GRCh38
rs773412526 Chr.12: 14806227-14806227 on Build GRCh38
rs770503888 Chr.12: 14806041-14806041 on Build GRCh38
rs749737279 Chr.12: 14806023-14806023 on Build GRCh38
rs567614549 Chr.12: 14806092-14806092 on Build GRCh38
rs772254777 Chr.12: 14806218-14806218 on Build GRCh38
rs938582808 Chr.12: 14806120-14806120 on Build GRCh38

View Full Product Details