Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14806313-14806843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
NM_175874.3 XM_005253322.4 XM_017018871.1 XM_017018872.1 XM_017018873.1 XM_017018874.1 NM_001013698.2 XM_017019312.1

Gene Symbol
SMCO3
Entrez Gene ID
440087
Gene Name
single-pass membrane protein with coiled-coil domains 3
Gene Aliases
C12orf69
UniGene
Hs.220931
Gene Chromosome Location
Chr.12: 14804650-14816761 on Build GRCh38
Transcripts
NM_175874.3 XM_005253322.4 XM_017018871.1 XM_017018872.1 XM_017018873.1 XM_017018874.1 NM_001013698.2 XM_017019312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198023 c.311A>T p.E104V Chr.12: 14806370-14806370 on Build GRCh38
6227938 c.68A>C p.Q23P Chr.12: 14806613-14806613 on Build GRCh38
140684 c.227T>C p.I76T Chr.12: 14806454-14806454 on Build GRCh38
6315590 c.320C>A p.T107K Chr.12: 14806361-14806361 on Build GRCh38
6486731 c.43C>T p.R15W Chr.12: 14806638-14806638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918532740 Chr.12: 14806798-14806798 on Build GRCh38
rs781434492 Chr.12: 14806638-14806638 on Build GRCh38
rs755814181 Chr.12: 14806399-14806399 on Build GRCh38
rs757535647 Chr.12: 14806564-14806564 on Build GRCh38
rs546938311 Chr.12: 14806695-14806695 on Build GRCh38
rs11609202 Chr.12: 14806536-14806536 on Build GRCh38
rs750104770 Chr.12: 14806396-14806396 on Build GRCh38
rs774846552 Chr.12: 14806681-14806681 on Build GRCh38
rs536305929 Chr.12: 14806696-14806696 on Build GRCh38
rs965681953 Chr.12: 14806343-14806343 on Build GRCh38
rs750630915 Chr.12: 14806639-14806639 on Build GRCh38
rs146732271 Chr.12: 14806687-14806687 on Build GRCh38
rs769220421 Chr.12: 14806595-14806595 on Build GRCh38
rs776842204 Chr.12: 14806447-14806447 on Build GRCh38
rs186323170 Chr.12: 14806350-14806350 on Build GRCh38
rs2241221 Chr.12: 14806457-14806457 on Build GRCh38
rs918434163 Chr.12: 14806632-14806632 on Build GRCh38
rs773575920 Chr.12: 14806341-14806341 on Build GRCh38
rs905191646 Chr.12: 14806607-14806607 on Build GRCh38
rs1004491239 Chr.12: 14806376-14806376 on Build GRCh38
rs1009969754 Chr.12: 14806481-14806481 on Build GRCh38
rs201776106 Chr.12: 14806640-14806640 on Build GRCh38
rs765185528 Chr.12: 14806535-14806535 on Build GRCh38
rs774686155 Chr.12: 14806600-14806600 on Build GRCh38
rs764421551 Chr.12: 14806478-14806478 on Build GRCh38
rs944849370 Chr.12: 14806465-14806465 on Build GRCh38
rs370155859 Chr.12: 14806690-14806690 on Build GRCh38
rs776182743 Chr.12: 14806612-14806612 on Build GRCh38
rs552576725 Chr.12: 14806433-14806433 on Build GRCh38
rs747208866 Chr.12: 14806439-14806439 on Build GRCh38
rs777676311 Chr.12: 14806551-14806551 on Build GRCh38
rs201011813 Chr.12: 14806656-14806656 on Build GRCh38
rs920907729 Chr.12: 14806435-14806435 on Build GRCh38
rs751342043 Chr.12: 14806636-14806636 on Build GRCh38
rs778090071 Chr.12: 14806434-14806434 on Build GRCh38
rs199916742 Chr.12: 14806520-14806520 on Build GRCh38
rs762179150 Chr.12: 14806608-14806608 on Build GRCh38
rs139378466 Chr.12: 14806358-14806358 on Build GRCh38
rs770276935 Chr.12: 14806460-14806460 on Build GRCh38
rs530617756 Chr.12: 14806574-14806574 on Build GRCh38
rs575449284 Chr.12: 14806428-14806428 on Build GRCh38
rs561150713 Chr.12: 14806647-14806647 on Build GRCh38
rs201197590 Chr.12: 14806637-14806637 on Build GRCh38
rs74068129 Chr.12: 14806686-14806686 on Build GRCh38
rs758456011 Chr.12: 14806537-14806537 on Build GRCh38
rs867713866 Chr.12: 14806766-14806766 on Build GRCh38
rs1015803317 Chr.12: 14806732-14806732 on Build GRCh38
rs761656761 Chr.12: 14806573-14806573 on Build GRCh38
rs771315633 Chr.12: 14806443-14806443 on Build GRCh38
rs781283957 Chr.12: 14806568-14806568 on Build GRCh38
rs754930763 Chr.12: 14806419-14806419 on Build GRCh38
rs754454798 Chr.12: 14806533-14806533 on Build GRCh38
rs267603396 Chr.12: 14806499-14806499 on Build GRCh38
rs373282630 Chr.12: 14806679-14806679 on Build GRCh38
rs375037271 Chr.12: 14806375-14806375 on Build GRCh38
rs969450833 Chr.12: 14806584-14806584 on Build GRCh38
rs753356358 Chr.12: 14806414-14806414 on Build GRCh38
rs752289435 Chr.12: 14806622-14806622 on Build GRCh38
rs778986129 Chr.12: 14806805-14806805 on Build GRCh38
rs774739394 Chr.12: 14806485-14806485 on Build GRCh38
rs980830564 Chr.12: 14806734-14806734 on Build GRCh38
rs533370487 Chr.12: 14806807-14806807 on Build GRCh38
rs769794820 Chr.12: 14806581-14806581 on Build GRCh38
rs369538893 Chr.12: 14806609-14806609 on Build GRCh38
rs766494478 Chr.12: 14806619-14806619 on Build GRCh38
rs749523465 Chr.12: 14806692-14806692 on Build GRCh38
rs202090335 Chr.12: 14806623-14806623 on Build GRCh38
rs1022456077 Chr.12: 14806691-14806691 on Build GRCh38
rs759041348 Chr.12: 14806617-14806617 on Build GRCh38
rs960958437 Chr.12: 14806754-14806754 on Build GRCh38
rs775864955 Chr.12: 14806470-14806470 on Build GRCh38
rs753092756 Chr.12: 14806371-14806371 on Build GRCh38
rs369044986 Chr.12: 14806411-14806411 on Build GRCh38
rs749729584 Chr.12: 14806591-14806591 on Build GRCh38
rs760799839 Chr.12: 14806345-14806345 on Build GRCh38
rs771154364 Chr.12: 14806346-14806346 on Build GRCh38
rs778733013 Chr.12: 14806426-14806426 on Build GRCh38
rs375028145 Chr.12: 14806475-14806475 on Build GRCh38
rs773677201 Chr.12: 14806663-14806663 on Build GRCh38
rs892718603 Chr.12: 14806688-14806688 on Build GRCh38
rs369381649 Chr.12: 14806676-14806676 on Build GRCh38
rs559452173 Chr.12: 14806712-14806712 on Build GRCh38
rs750301749 Chr.12: 14806721-14806721 on Build GRCh38
rs764180671 Chr.12: 14806635-14806635 on Build GRCh38
rs746863504 Chr.12: 14806555-14806555 on Build GRCh38
rs953691062 Chr.12: 14806787-14806787 on Build GRCh38
rs776568427 Chr.12: 14806509-14806510 on Build GRCh38
rs767087930 Chr.12: 14806393-14806393 on Build GRCh38
rs769093901 Chr.12: 14806642-14806642 on Build GRCh38
rs757440130 Chr.12: 14806739-14806739 on Build GRCh38
rs776014062 Chr.12: 14806582-14806582 on Build GRCh38

View Full Product Details