Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14840754-14841258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART4
Entrez Gene ID
420
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
Gene Aliases
ARTC4, CD297, DO, DOK1
UniGene
Hs.591158
Gene Chromosome Location
Chr.12: 14829311-14843479 on Build GRCh38
Transcripts
NM_021071.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849808 c.413G>C p.R138T Chr.12: 14840885-14840885 on Build GRCh38
256350 c.273G>T p.K91N Chr.12: 14841025-14841025 on Build GRCh38
937574 c.265G>T p.A89S Chr.12: 14841033-14841033 on Build GRCh38
274056 c.475T>G p.L159V Chr.12: 14840823-14840823 on Build GRCh38
937575 c.171C>T p.F57F Chr.12: 14841127-14841127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762396780 Chr.12: 14840813-14840813 on Build GRCh38
rs113022131 Chr.12: 14840816-14840816 on Build GRCh38
rs187129757 Chr.12: 14841172-14841172 on Build GRCh38
rs144350521 Chr.12: 14840941-14840941 on Build GRCh38
rs574273608 Chr.12: 14840860-14840860 on Build GRCh38
rs773617232 Chr.12: 14840932-14840932 on Build GRCh38
rs754839745 Chr.12: 14841079-14841079 on Build GRCh38
rs745633535 Chr.12: 14840862-14840862 on Build GRCh38
rs534501549 Chr.12: 14841133-14841133 on Build GRCh38
rs1861698 Chr.12: 14840920-14840920 on Build GRCh38
rs751556309 Chr.12: 14841201-14841201 on Build GRCh38
rs760979592 Chr.12: 14840826-14840826 on Build GRCh38
rs201915266 Chr.12: 14841127-14841127 on Build GRCh38
rs142165195 Chr.12: 14840946-14840946 on Build GRCh38
rs748187879 Chr.12: 14840990-14840990 on Build GRCh38
rs753699386 Chr.12: 14840963-14840963 on Build GRCh38
rs757126077 Chr.12: 14840859-14840859 on Build GRCh38
rs750017545 Chr.12: 14841059-14841059 on Build GRCh38
rs768259592 Chr.12: 14841039-14841039 on Build GRCh38
rs974606146 Chr.12: 14841229-14841229 on Build GRCh38
rs141907337 Chr.12: 14841010-14841010 on Build GRCh38
rs587777831 Chr.12: 14841155-14841155 on Build GRCh38
rs28362797 Chr.12: 14840975-14840975 on Build GRCh38
rs963550699 Chr.12: 14841204-14841204 on Build GRCh38
rs751664806 Chr.12: 14840991-14840992 on Build GRCh38
rs756117992 Chr.12: 14840959-14840959 on Build GRCh38
rs758495352 Chr.12: 14841180-14841180 on Build GRCh38
rs146389191 Chr.12: 14840930-14840931 on Build GRCh38
rs201524568 Chr.12: 14840833-14840833 on Build GRCh38
rs569342021 Chr.12: 14841077-14841077 on Build GRCh38
rs752932674 Chr.12: 14841095-14841095 on Build GRCh38
rs912879393 Chr.12: 14840991-14840991 on Build GRCh38
rs551070552 Chr.12: 14841042-14841042 on Build GRCh38
rs28362799 Chr.12: 14840893-14840893 on Build GRCh38
rs781300086 Chr.12: 14841205-14841205 on Build GRCh38
rs904706147 Chr.12: 14841052-14841052 on Build GRCh38
rs772253839 Chr.12: 14840995-14840995 on Build GRCh38
rs775874295 Chr.12: 14841132-14841132 on Build GRCh38
rs774612409 Chr.12: 14840792-14840792 on Build GRCh38
rs560106322 Chr.12: 14840904-14840904 on Build GRCh38
rs140754622 Chr.12: 14841134-14841134 on Build GRCh38
rs199993800 Chr.12: 14841126-14841126 on Build GRCh38
rs150640567 Chr.12: 14841113-14841113 on Build GRCh38
rs779947895 Chr.12: 14840887-14840887 on Build GRCh38
rs757382784 Chr.12: 14841203-14841203 on Build GRCh38
rs777697516 Chr.12: 14841108-14841108 on Build GRCh38
rs764385453 Chr.12: 14841181-14841181 on Build GRCh38
rs587777832 Chr.12: 14840948-14840955 on Build GRCh38
rs759583311 Chr.12: 14841170-14841170 on Build GRCh38
rs752593220 Chr.12: 14841174-14841174 on Build GRCh38
rs748244898 Chr.12: 14840915-14840915 on Build GRCh38
rs868222097 Chr.12: 14841058-14841058 on Build GRCh38
rs766344506 Chr.12: 14841068-14841068 on Build GRCh38
rs747130006 Chr.12: 14841112-14841112 on Build GRCh38
rs201635777 Chr.12: 14840926-14840926 on Build GRCh38
rs771530070 Chr.12: 14841015-14841015 on Build GRCh38
rs758710680 Chr.12: 14841104-14841104 on Build GRCh38
rs200270921 Chr.12: 14841028-14841028 on Build GRCh38
rs77336576 Chr.12: 14840853-14840853 on Build GRCh38
rs559028739 Chr.12: 14841139-14841139 on Build GRCh38
rs1036425458 Chr.12: 14840960-14840961 on Build GRCh38
rs182981694 Chr.12: 14841081-14841081 on Build GRCh38
rs777946228 Chr.12: 14840998-14840998 on Build GRCh38
rs781739863 Chr.12: 14841125-14841125 on Build GRCh38
rs376049820 Chr.12: 14841138-14841138 on Build GRCh38
rs937472500 Chr.12: 14841060-14841060 on Build GRCh38
rs761290035 Chr.12: 14840936-14840936 on Build GRCh38
rs373370107 Chr.12: 14840815-14840815 on Build GRCh38
rs201613255 Chr.12: 14840843-14840843 on Build GRCh38
rs1025626373 Chr.12: 14841123-14841123 on Build GRCh38
rs779308025 Chr.12: 14840986-14840986 on Build GRCh38
rs759901596 Chr.12: 14841030-14841030 on Build GRCh38
rs1009210488 Chr.12: 14841124-14841124 on Build GRCh38
rs28362798 Chr.12: 14840948-14840948 on Build GRCh38
rs140760247 Chr.12: 14840791-14840791 on Build GRCh38
rs771165427 Chr.12: 14840940-14840940 on Build GRCh38
rs766177871 Chr.12: 14841161-14841161 on Build GRCh38
rs774516023 Chr.12: 14840910-14840910 on Build GRCh38
rs755882503 Chr.12: 14840876-14840876 on Build GRCh38
rs150107956 Chr.12: 14840828-14840828 on Build GRCh38
rs201266552 Chr.12: 14840961-14840961 on Build GRCh38
rs138414095 Chr.12: 14840844-14840844 on Build GRCh38
rs760471370 Chr.12: 14841064-14841064 on Build GRCh38
rs56340844 Chr.12: 14840856-14840856 on Build GRCh38
rs926189320 Chr.12: 14840907-14840907 on Build GRCh38
rs761504024 Chr.12: 14841051-14841051 on Build GRCh38
rs776684513 Chr.12: 14841168-14841168 on Build GRCh38
rs775603761 Chr.12: 14840786-14840786 on Build GRCh38
rs758090906 Chr.12: 14840841-14840841 on Build GRCh38
rs778333849 Chr.12: 14840954-14840954 on Build GRCh38

View Full Product Details