Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 18738797-18739301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA3
Entrez Gene ID
93661
Gene Name
capping actin protein of muscle Z-line alpha subunit 3
Gene Aliases
CAPPA3, Gsg3, HEL-S-86
UniGene
Hs.131288
Gene Chromosome Location
Chr.12: 18738111-18739188 on Build GRCh38
Transcripts
NM_033328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692950 c.897A>G p.I299M Chr.12: 18739165-18739165 on Build GRCh38
348621 c.627G>A p.L209L Chr.12: 18738895-18738895 on Build GRCh38
3458932 c.778C>T p.L260F Chr.12: 18739046-18739046 on Build GRCh38
6620043 c.896T>C p.I299T Chr.12: 18739164-18739164 on Build GRCh38
4912605 c.862A>G p.I288V Chr.12: 18739130-18739130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748631856 Chr.12: 18739100-18739100 on Build GRCh38
rs760616985 Chr.12: 18739020-18739020 on Build GRCh38
rs762265968 Chr.12: 18739024-18739024 on Build GRCh38
rs548812304 Chr.12: 18739014-18739014 on Build GRCh38
rs144355988 Chr.12: 18739068-18739068 on Build GRCh38
rs573954336 Chr.12: 18739222-18739222 on Build GRCh38
rs753594924 Chr.12: 18738930-18738930 on Build GRCh38
rs762434385 Chr.12: 18739127-18739127 on Build GRCh38
rs902648501 Chr.12: 18738854-18738854 on Build GRCh38
rs751069055 Chr.12: 18739152-18739152 on Build GRCh38
rs867785228 Chr.12: 18738827-18738827 on Build GRCh38
rs753268001 Chr.12: 18738825-18738825 on Build GRCh38
rs149089246 Chr.12: 18738831-18738831 on Build GRCh38
rs776565339 Chr.12: 18739078-18739078 on Build GRCh38
rs773063949 Chr.12: 18739013-18739013 on Build GRCh38
rs909076245 Chr.12: 18739002-18739002 on Build GRCh38
rs775598852 Chr.12: 18739007-18739007 on Build GRCh38
rs769970340 Chr.12: 18738889-18738889 on Build GRCh38
rs1043008066 Chr.12: 18739101-18739101 on Build GRCh38
rs757152109 Chr.12: 18739056-18739056 on Build GRCh38
rs780418160 Chr.12: 18739192-18739192 on Build GRCh38
rs146803009 Chr.12: 18739124-18739124 on Build GRCh38
rs1006602063 Chr.12: 18738928-18738928 on Build GRCh38
rs34013843 Chr.12: 18739045-18739045 on Build GRCh38
rs775442096 Chr.12: 18738891-18738891 on Build GRCh38
rs757174551 Chr.12: 18739158-18739158 on Build GRCh38
rs768925797 Chr.12: 18739092-18739092 on Build GRCh38
rs755787082 Chr.12: 18738822-18738822 on Build GRCh38
rs374397815 Chr.12: 18738918-18738918 on Build GRCh38
rs368077520 Chr.12: 18738823-18738823 on Build GRCh38
rs746788497 Chr.12: 18738951-18738951 on Build GRCh38
rs15547 Chr.12: 18739037-18739037 on Build GRCh38
rs779069350 Chr.12: 18739209-18739209 on Build GRCh38
rs760973279 Chr.12: 18739121-18739121 on Build GRCh38
rs749578410 Chr.12: 18739201-18739204 on Build GRCh38
rs139395998 Chr.12: 18739042-18739042 on Build GRCh38
rs982225881 Chr.12: 18739010-18739010 on Build GRCh38
rs542967315 Chr.12: 18739266-18739266 on Build GRCh38
rs143378750 Chr.12: 18738970-18738970 on Build GRCh38
rs767377385 Chr.12: 18739160-18739160 on Build GRCh38
rs901404760 Chr.12: 18739230-18739230 on Build GRCh38
rs148240048 Chr.12: 18738910-18738910 on Build GRCh38
rs1029794528 Chr.12: 18738915-18738915 on Build GRCh38
rs963233488 Chr.12: 18739023-18739023 on Build GRCh38
rs148768083 Chr.12: 18739096-18739096 on Build GRCh38
rs777482385 Chr.12: 18738856-18738856 on Build GRCh38
rs746412073 Chr.12: 18739129-18739129 on Build GRCh38
rs143115929 Chr.12: 18738841-18738841 on Build GRCh38
rs918445686 Chr.12: 18738995-18738995 on Build GRCh38
rs781255101 Chr.12: 18738986-18738986 on Build GRCh38
rs146957132 Chr.12: 18738947-18738947 on Build GRCh38
rs997115304 Chr.12: 18738966-18738966 on Build GRCh38
rs1021106187 Chr.12: 18738848-18738848 on Build GRCh38
rs137913550 Chr.12: 18738950-18738950 on Build GRCh38
rs780970849 Chr.12: 18739059-18739059 on Build GRCh38
rs773589348 Chr.12: 18739114-18739114 on Build GRCh38
rs367620204 Chr.12: 18738925-18738925 on Build GRCh38
rs750209976 Chr.12: 18739179-18739179 on Build GRCh38
rs565732016 Chr.12: 18739029-18739029 on Build GRCh38
rs200098261 Chr.12: 18739038-18739038 on Build GRCh38
rs536940718 Chr.12: 18739122-18739122 on Build GRCh38
rs141221149 Chr.12: 18738924-18738924 on Build GRCh38
rs147540838 Chr.12: 18738987-18738987 on Build GRCh38
rs764011869 Chr.12: 18739046-18739046 on Build GRCh38
rs370457183 Chr.12: 18738826-18738826 on Build GRCh38
rs775037231 Chr.12: 18739017-18739017 on Build GRCh38
rs751553411 Chr.12: 18739055-18739055 on Build GRCh38
rs749347585 Chr.12: 18739085-18739085 on Build GRCh38
rs962348037 Chr.12: 18739268-18739268 on Build GRCh38
rs61740731 Chr.12: 18738833-18738833 on Build GRCh38
rs759527168 Chr.12: 18738873-18738873 on Build GRCh38
rs149635089 Chr.12: 18739061-18739061 on Build GRCh38
rs375738945 Chr.12: 18738944-18738944 on Build GRCh38
rs569664267 Chr.12: 18738941-18738941 on Build GRCh38
rs754432502 Chr.12: 18738932-18738932 on Build GRCh38
rs375023759 Chr.12: 18739207-18739207 on Build GRCh38
rs998380330 Chr.12: 18739248-18739248 on Build GRCh38
rs769909361 Chr.12: 18738997-18738997 on Build GRCh38
rs184416750 Chr.12: 18739221-18739221 on Build GRCh38
rs143745111 Chr.12: 18739106-18739106 on Build GRCh38
rs369139076 Chr.12: 18739208-18739208 on Build GRCh38
rs756310359 Chr.12: 18739066-18739066 on Build GRCh38
rs746840009 Chr.12: 18738860-18738860 on Build GRCh38
rs751922545 Chr.12: 18738920-18738920 on Build GRCh38
rs755900713 Chr.12: 18739191-18739191 on Build GRCh38
rs972617460 Chr.12: 18738981-18738981 on Build GRCh38
rs776949051 Chr.12: 18738862-18738862 on Build GRCh38

View Full Product Details