Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24811884-24812385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT1
Entrez Gene ID
586
Gene Name
branched chain amino acid transaminase 1
Gene Aliases
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18
UniGene
Hs.438993
Gene Chromosome Location
Chr.12: 24810024-24949459 on Build GRCh38
Transcripts
NM_001178091.1 NM_001178092.1 NM_001178093.1 NM_001178094.1 NM_005504.6 XM_017019768.1

SNP Detail

SNP ID Chromosome Location
rs964292197 Chr.12: 24811981-24811981 on Build GRCh38
rs61429607 Chr.12: 24812175-24812175 on Build GRCh38
rs35804569 Chr.12: 24812109-24812110 on Build GRCh38
rs908590913 Chr.12: 24812082-24812082 on Build GRCh38
rs746749813 Chr.12: 24811944-24811944 on Build GRCh38
rs1041710158 Chr.12: 24812155-24812155 on Build GRCh38
rs967579104 Chr.12: 24811929-24811929 on Build GRCh38
rs149830240 Chr.12: 24812188-24812188 on Build GRCh38
rs1038355592 Chr.12: 24812110-24812110 on Build GRCh38
rs1011295959 Chr.12: 24812253-24812253 on Build GRCh38
rs558616078 Chr.12: 24812331-24812331 on Build GRCh38
rs964569110 Chr.12: 24811984-24811984 on Build GRCh38
rs35585041 Chr.12: 24812154-24812155 on Build GRCh38
rs79282831 Chr.12: 24811927-24811927 on Build GRCh38
rs1021356613 Chr.12: 24812296-24812296 on Build GRCh38
rs1014575801 Chr.12: 24812321-24812322 on Build GRCh38
rs548424682 Chr.12: 24811953-24811953 on Build GRCh38
rs1030294094 Chr.12: 24812303-24812303 on Build GRCh38
rs933163472 Chr.12: 24812037-24812037 on Build GRCh38
rs974417533 Chr.12: 24812344-24812344 on Build GRCh38
rs920411479 Chr.12: 24812092-24812092 on Build GRCh38
rs767787260 Chr.12: 24812135-24812135 on Build GRCh38
rs769792177 Chr.12: 24812055-24812055 on Build GRCh38
rs899988521 Chr.12: 24812291-24812291 on Build GRCh38
rs36085546 Chr.12: 24812319-24812320 on Build GRCh38
rs577011508 Chr.12: 24812354-24812354 on Build GRCh38
rs907653559 Chr.12: 24812058-24812058 on Build GRCh38
rs534836829 Chr.12: 24812070-24812070 on Build GRCh38
rs955794442 Chr.12: 24812241-24812241 on Build GRCh38
rs955993740 Chr.12: 24811942-24811942 on Build GRCh38
rs397824972 Chr.12: 24812324-24812325 on Build GRCh38
rs563256845 Chr.12: 24812019-24812019 on Build GRCh38
rs563439556 Chr.12: 24812003-24812003 on Build GRCh38
rs780090775 Chr.12: 24812100-24812100 on Build GRCh38
rs1000353527 Chr.12: 24811941-24811941 on Build GRCh38
rs955644449 Chr.12: 24812310-24812310 on Build GRCh38
rs186347875 Chr.12: 24812111-24812111 on Build GRCh38
rs914259066 Chr.12: 24811967-24811967 on Build GRCh38
rs954487963 Chr.12: 24811990-24811990 on Build GRCh38
rs535671279 Chr.12: 24812154-24812154 on Build GRCh38
rs35186964 Chr.12: 24812013-24812014 on Build GRCh38
rs770700724 Chr.12: 24811961-24811961 on Build GRCh38
rs1033153805 Chr.12: 24812202-24812202 on Build GRCh38
rs761954292 Chr.12: 24812112-24812112 on Build GRCh38
rs772038139 Chr.12: 24811999-24812001 on Build GRCh38
rs1055695278 Chr.12: 24812121-24812121 on Build GRCh38
rs1032262331 Chr.12: 24812228-24812228 on Build GRCh38
rs546867523 Chr.12: 24812085-24812085 on Build GRCh38
rs759369378 Chr.12: 24812009-24812009 on Build GRCh38
rs964326806 Chr.12: 24812332-24812332 on Build GRCh38
rs528769253 Chr.12: 24812348-24812348 on Build GRCh38
rs557601277 Chr.12: 24812160-24812160 on Build GRCh38
rs776450677 Chr.12: 24811979-24811979 on Build GRCh38
rs199939066 Chr.12: 24812319-24812319 on Build GRCh38
rs373375247 Chr.12: 24811978-24811978 on Build GRCh38
rs923211758 Chr.12: 24812032-24812032 on Build GRCh38
rs921634988 Chr.12: 24811987-24811987 on Build GRCh38
rs397802152 Chr.12: 24812325-24812326 on Build GRCh38
rs570588709 Chr.12: 24812027-24812027 on Build GRCh38
rs997059304 Chr.12: 24812298-24812298 on Build GRCh38
rs530483177 Chr.12: 24812023-24812023 on Build GRCh38
rs907483451 Chr.12: 24812189-24812189 on Build GRCh38
rs895818044 Chr.12: 24812131-24812131 on Build GRCh38
rs1003011315 Chr.12: 24812224-24812224 on Build GRCh38
rs987583768 Chr.12: 24812316-24812317 on Build GRCh38
rs1037559595 Chr.12: 24812090-24812090 on Build GRCh38

View Full Product Details