Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24815313-24815813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT1
Entrez Gene ID
586
Gene Name
branched chain amino acid transaminase 1
Gene Aliases
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18
UniGene
Hs.438993
Gene Chromosome Location
Chr.12: 24810024-24949459 on Build GRCh38
Transcripts
NM_001178091.1 NM_001178092.1 NM_001178093.1 NM_001178094.1 NM_005504.6 XM_017019768.1

SNP Detail

SNP ID Chromosome Location
rs1043776876 Chr.12: 24815640-24815640 on Build GRCh38
rs184680605 Chr.12: 24815456-24815456 on Build GRCh38
rs571620828 Chr.12: 24815708-24815708 on Build GRCh38
rs529600333 Chr.12: 24815618-24815618 on Build GRCh38
rs896630191 Chr.12: 24815574-24815574 on Build GRCh38
rs879069603 Chr.12: 24815378-24815380 on Build GRCh38
rs549194793 Chr.12: 24815600-24815600 on Build GRCh38
rs76576230 Chr.12: 24815555-24815555 on Build GRCh38
rs997888240 Chr.12: 24815483-24815483 on Build GRCh38
rs912395349 Chr.12: 24815398-24815398 on Build GRCh38
rs1000451600 Chr.12: 24815617-24815617 on Build GRCh38
rs888005820 Chr.12: 24815521-24815521 on Build GRCh38
rs985765828 Chr.12: 24815631-24815631 on Build GRCh38
rs371369654 Chr.12: 24815373-24815373 on Build GRCh38
rs143612272 Chr.12: 24815634-24815634 on Build GRCh38
rs1025297729 Chr.12: 24815534-24815534 on Build GRCh38
rs7315828 Chr.12: 24815614-24815614 on Build GRCh38
rs565128614 Chr.12: 24815762-24815762 on Build GRCh38
rs551707368 Chr.12: 24815666-24815666 on Build GRCh38
rs919307673 Chr.12: 24815778-24815778 on Build GRCh38
rs1049099321 Chr.12: 24815515-24815515 on Build GRCh38
rs567545790 Chr.12: 24815613-24815613 on Build GRCh38
rs1019551042 Chr.12: 24815726-24815726 on Build GRCh38
rs1005279169 Chr.12: 24815548-24815551 on Build GRCh38
rs951141128 Chr.12: 24815536-24815536 on Build GRCh38
rs762371170 Chr.12: 24815506-24815506 on Build GRCh38
rs878863719 Chr.12: 24815589-24815589 on Build GRCh38
rs531908375 Chr.12: 24815359-24815359 on Build GRCh38
rs117477136 Chr.12: 24815567-24815567 on Build GRCh38
rs965584219 Chr.12: 24815758-24815758 on Build GRCh38
rs1040840664 Chr.12: 24815453-24815453 on Build GRCh38
rs953956248 Chr.12: 24815620-24815620 on Build GRCh38
rs981105839 Chr.12: 24815565-24815565 on Build GRCh38
rs1040595708 Chr.12: 24815406-24815408 on Build GRCh38
rs953964963 Chr.12: 24815357-24815357 on Build GRCh38
rs910716002 Chr.12: 24815712-24815712 on Build GRCh38
rs1008226051 Chr.12: 24815403-24815403 on Build GRCh38
rs988129430 Chr.12: 24815612-24815612 on Build GRCh38
rs879495783 Chr.12: 24815457-24815457 on Build GRCh38
rs749947021 Chr.12: 24815585-24815585 on Build GRCh38
rs925170683 Chr.12: 24815347-24815347 on Build GRCh38
rs150885323 Chr.12: 24815592-24815592 on Build GRCh38
rs969210884 Chr.12: 24815556-24815556 on Build GRCh38
rs943913234 Chr.12: 24815401-24815401 on Build GRCh38
rs532776262 Chr.12: 24815539-24815539 on Build GRCh38

View Full Product Details