Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24816644-24817146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT1
Entrez Gene ID
586
Gene Name
branched chain amino acid transaminase 1
Gene Aliases
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18
UniGene
Hs.438993
Gene Chromosome Location
Chr.12: 24810024-24949459 on Build GRCh38
Transcripts
NM_001178091.1 NM_001178092.1 NM_001178093.1 NM_001178094.1 NM_005504.6 XM_017019768.1

SNP Detail

SNP ID Chromosome Location
rs777576778 Chr.12: 24816817-24816817 on Build GRCh38
rs896230019 Chr.12: 24816761-24816761 on Build GRCh38
rs7134619 Chr.12: 24816788-24816788 on Build GRCh38
rs114241843 Chr.12: 24817081-24817081 on Build GRCh38
rs997610163 Chr.12: 24816838-24816838 on Build GRCh38
rs757162221 Chr.12: 24816729-24816729 on Build GRCh38
rs779196638 Chr.12: 24817050-24817050 on Build GRCh38
rs942258981 Chr.12: 24816723-24816723 on Build GRCh38
rs894292812 Chr.12: 24816925-24816925 on Build GRCh38
rs904831973 Chr.12: 24816748-24816748 on Build GRCh38
rs7306168 Chr.12: 24816994-24816994 on Build GRCh38
rs536255582 Chr.12: 24817091-24817091 on Build GRCh38
rs958254723 Chr.12: 24817031-24817031 on Build GRCh38
rs1038052587 Chr.12: 24816741-24816741 on Build GRCh38
rs371311626 Chr.12: 24816827-24816827 on Build GRCh38
rs7134871 Chr.12: 24816875-24816875 on Build GRCh38
rs557301021 Chr.12: 24816824-24816824 on Build GRCh38
rs1056148779 Chr.12: 24816915-24816915 on Build GRCh38
rs922379287 Chr.12: 24816826-24816826 on Build GRCh38
rs1046766298 Chr.12: 24816874-24816874 on Build GRCh38
rs147746575 Chr.12: 24817046-24817046 on Build GRCh38
rs937706166 Chr.12: 24816976-24816976 on Build GRCh38
rs972966157 Chr.12: 24816865-24816865 on Build GRCh38
rs189236479 Chr.12: 24816758-24816758 on Build GRCh38
rs1030979198 Chr.12: 24816995-24816995 on Build GRCh38
rs780855538 Chr.12: 24816765-24816765 on Build GRCh38
rs531498813 Chr.12: 24816934-24816934 on Build GRCh38
rs558791430 Chr.12: 24816697-24816697 on Build GRCh38
rs75092198 Chr.12: 24816669-24816669 on Build GRCh38
rs1025501734 Chr.12: 24816689-24816689 on Build GRCh38
rs372335636 Chr.12: 24816966-24816966 on Build GRCh38
rs983644878 Chr.12: 24816877-24816877 on Build GRCh38
rs929427055 Chr.12: 24816694-24816694 on Build GRCh38
rs959694540 Chr.12: 24816681-24816681 on Build GRCh38
rs958285568 Chr.12: 24816703-24816703 on Build GRCh38
rs1012316914 Chr.12: 24816687-24816687 on Build GRCh38
rs929728242 Chr.12: 24817115-24817115 on Build GRCh38
rs926316049 Chr.12: 24816945-24816945 on Build GRCh38
rs749478994 Chr.12: 24817038-24817038 on Build GRCh38
rs532481594 Chr.12: 24817016-24817016 on Build GRCh38
rs115193920 Chr.12: 24817095-24817095 on Build GRCh38
rs965034683 Chr.12: 24816834-24816834 on Build GRCh38
rs377561726 Chr.12: 24816933-24816933 on Build GRCh38
rs1042217787 Chr.12: 24817042-24817042 on Build GRCh38
rs111658289 Chr.12: 24816696-24816696 on Build GRCh38
rs7305794 Chr.12: 24816693-24816693 on Build GRCh38
rs12366411 Chr.12: 24816879-24816879 on Build GRCh38
rs989729518 Chr.12: 24817041-24817041 on Build GRCh38
rs796092696 Chr.12: 24816952-24816952 on Build GRCh38
rs937777859 Chr.12: 24816752-24816752 on Build GRCh38
rs7134734 Chr.12: 24816804-24816804 on Build GRCh38
rs1025051861 Chr.12: 24816941-24816941 on Build GRCh38

View Full Product Details