Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24996018-24996515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf77
Entrez Gene ID
196415
Gene Name
chromosome 12 open reading frame 77
Gene Aliases
-
UniGene
Hs.434453
Gene Chromosome Location
Chr.12: 24993424-24997673 on Build GRCh38
Transcripts
NM_001101339.2 XM_017018966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4793252 c.165A>G p.T55T Chr.12: 24996049-24996049 on Build GRCh38
4751424 c.109G>T p.A37S Chr.12: 24996234-24996234 on Build GRCh38
6614274 c.36C>T p.S12S Chr.12: 24996307-24996307 on Build GRCh38
4878413 c.117G>A p.L39L Chr.12: 24996226-24996226 on Build GRCh38
4845347 c.100G>C p.D34H Chr.12: 24996243-24996243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886666324 Chr.12: 24996136-24996136 on Build GRCh38
rs775591584 Chr.12: 24996080-24996080 on Build GRCh38
rs536602912 Chr.12: 24996417-24996417 on Build GRCh38
rs546518395 Chr.12: 24996387-24996387 on Build GRCh38
rs888746650 Chr.12: 24996484-24996484 on Build GRCh38
rs756199015 Chr.12: 24996107-24996107 on Build GRCh38
rs763994402 Chr.12: 24996126-24996126 on Build GRCh38
rs190975931 Chr.12: 24996487-24996487 on Build GRCh38
rs755406146 Chr.12: 24996164-24996164 on Build GRCh38
rs546291465 Chr.12: 24996230-24996230 on Build GRCh38
rs754370101 Chr.12: 24996163-24996163 on Build GRCh38
rs762944541 Chr.12: 24996084-24996084 on Build GRCh38
rs899333542 Chr.12: 24996205-24996205 on Build GRCh38
rs73297350 Chr.12: 24996423-24996423 on Build GRCh38
rs35648127 Chr.12: 24996271-24996272 on Build GRCh38
rs769614515 Chr.12: 24996306-24996306 on Build GRCh38
rs940899851 Chr.12: 24996422-24996422 on Build GRCh38
rs575844935 Chr.12: 24996043-24996043 on Build GRCh38
rs779769904 Chr.12: 24996287-24996287 on Build GRCh38
rs753193234 Chr.12: 24996245-24996245 on Build GRCh38
rs372039444 Chr.12: 24996217-24996217 on Build GRCh38
rs373725485 Chr.12: 24996218-24996218 on Build GRCh38
rs1009998550 Chr.12: 24996106-24996106 on Build GRCh38
rs550987750 Chr.12: 24996383-24996383 on Build GRCh38
rs764605788 Chr.12: 24996371-24996371 on Build GRCh38
rs569672109 Chr.12: 24996392-24996392 on Build GRCh38
rs148580600 Chr.12: 24996141-24996141 on Build GRCh38
rs755899786 Chr.12: 24996381-24996381 on Build GRCh38
rs572817968 Chr.12: 24996481-24996481 on Build GRCh38
rs375447046 Chr.12: 24996058-24996058 on Build GRCh38
rs199556919 Chr.12: 24996335-24996335 on Build GRCh38
rs371145607 Chr.12: 24996054-24996054 on Build GRCh38
rs753890518 Chr.12: 24996234-24996234 on Build GRCh38
rs748065726 Chr.12: 24996334-24996334 on Build GRCh38
rs758911717 Chr.12: 24996368-24996368 on Build GRCh38
rs774168932 Chr.12: 24996317-24996317 on Build GRCh38
rs781383542 Chr.12: 24996041-24996041 on Build GRCh38
rs531373660 Chr.12: 24996361-24996361 on Build GRCh38
rs563751189 Chr.12: 24996074-24996074 on Build GRCh38
rs367610113 Chr.12: 24996246-24996246 on Build GRCh38
rs760935631 Chr.12: 24996347-24996347 on Build GRCh38
rs776769156 Chr.12: 24996360-24996360 on Build GRCh38
rs376946621 Chr.12: 24996044-24996044 on Build GRCh38
rs117451073 Chr.12: 24996425-24996425 on Build GRCh38
rs760843588 Chr.12: 24996232-24996232 on Build GRCh38
rs895948741 Chr.12: 24996105-24996105 on Build GRCh38
rs922061731 Chr.12: 24996039-24996040 on Build GRCh38
rs775110721 Chr.12: 24996216-24996216 on Build GRCh38
rs201330932 Chr.12: 24996215-24996215 on Build GRCh38
rs755807440 Chr.12: 24996281-24996281 on Build GRCh38
rs375630600 Chr.12: 24996214-24996214 on Build GRCh38
rs751961743 Chr.12: 24996376-24996376 on Build GRCh38
rs904282410 Chr.12: 24996454-24996454 on Build GRCh38
rs767628637 Chr.12: 24996097-24996097 on Build GRCh38
rs542420093 Chr.12: 24996103-24996103 on Build GRCh38
rs1041872849 Chr.12: 24996426-24996426 on Build GRCh38
rs753176120 Chr.12: 24996362-24996362 on Build GRCh38
rs745556326 Chr.12: 24996280-24996280 on Build GRCh38
rs774268091 Chr.12: 24996220-24996220 on Build GRCh38
rs542127798 Chr.12: 24996068-24996068 on Build GRCh38
rs999922540 Chr.12: 24996469-24996469 on Build GRCh38
rs574818895 Chr.12: 24996413-24996413 on Build GRCh38
rs772959527 Chr.12: 24996346-24996346 on Build GRCh38
rs370857814 Chr.12: 24996149-24996149 on Build GRCh38
rs763990265 Chr.12: 24996088-24996088 on Build GRCh38
rs142535528 Chr.12: 24996353-24996353 on Build GRCh38
rs779295463 Chr.12: 24996239-24996239 on Build GRCh38
rs780725313 Chr.12: 24996196-24996196 on Build GRCh38
rs772417083 Chr.12: 24996187-24996187 on Build GRCh38
rs763954284 Chr.12: 24996331-24996331 on Build GRCh38
rs573549489 Chr.12: 24996075-24996075 on Build GRCh38
rs748459368 Chr.12: 24996182-24996182 on Build GRCh38
rs748801008 Chr.12: 24996312-24996312 on Build GRCh38
rs762151731 Chr.12: 24996092-24996092 on Build GRCh38
rs866962830 Chr.12: 24996428-24996428 on Build GRCh38
rs528682778 Chr.12: 24996233-24996233 on Build GRCh38
rs768144020 Chr.12: 24996313-24996313 on Build GRCh38
rs761517221 Chr.12: 24996222-24996222 on Build GRCh38
rs921999907 Chr.12: 24996401-24996401 on Build GRCh38
rs748173841 Chr.12: 24996184-24996185 on Build GRCh38
rs377651931 Chr.12: 24996062-24996062 on Build GRCh38
rs954809481 Chr.12: 24996404-24996404 on Build GRCh38
rs564380856 Chr.12: 24996231-24996231 on Build GRCh38
rs779104687 Chr.12: 24996061-24996061 on Build GRCh38
rs749732818 Chr.12: 24996056-24996056 on Build GRCh38
rs751290889 Chr.12: 24996089-24996089 on Build GRCh38

View Full Product Details