Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26938280-26938769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
UniGene
Hs.505077
Gene Chromosome Location
Chr.12: 26905171-26938326 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 NM_001171887.1 NM_001171888.1 NM_015633.2

Gene Symbol
FGFR1OP2
Entrez Gene ID
26127
Gene Name
FGFR1 oncogene partner 2
Gene Aliases
HSPC123-like, WIT3.0
UniGene
Hs.591162
Gene Chromosome Location
Chr.12: 26938372-26966648 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 NM_001171887.1 NM_001171888.1 NM_015633.2

SNP Detail

SNP ID Chromosome Location
rs566627997 Chr.12: 26938652-26938652 on Build GRCh38
rs752383348 Chr.12: 26938553-26938553 on Build GRCh38
rs917605493 Chr.12: 26938389-26938389 on Build GRCh38
rs891217118 Chr.12: 26938529-26938529 on Build GRCh38
rs897844610 Chr.12: 26938517-26938517 on Build GRCh38
rs551476872 Chr.12: 26938684-26938684 on Build GRCh38
rs577915596 Chr.12: 26938512-26938512 on Build GRCh38
rs926906085 Chr.12: 26938312-26938312 on Build GRCh38
rs151037189 Chr.12: 26938399-26938399 on Build GRCh38
rs953160788 Chr.12: 26938740-26938740 on Build GRCh38
rs1034333582 Chr.12: 26938558-26938558 on Build GRCh38
rs765789665 Chr.12: 26938336-26938336 on Build GRCh38
rs999858339 Chr.12: 26938719-26938719 on Build GRCh38
rs959186421 Chr.12: 26938373-26938373 on Build GRCh38
rs184910886 Chr.12: 26938576-26938576 on Build GRCh38
rs953545629 Chr.12: 26938595-26938595 on Build GRCh38
rs189020728 Chr.12: 26938522-26938522 on Build GRCh38
rs554331699 Chr.12: 26938493-26938494 on Build GRCh38
rs902688978 Chr.12: 26938708-26938708 on Build GRCh38
rs140569759 Chr.12: 26938499-26938500 on Build GRCh38
rs1032705129 Chr.12: 26938732-26938732 on Build GRCh38
rs529893255 Chr.12: 26938346-26938346 on Build GRCh38
rs905300110 Chr.12: 26938661-26938661 on Build GRCh38
rs930675340 Chr.12: 26938521-26938521 on Build GRCh38
rs890429771 Chr.12: 26938379-26938379 on Build GRCh38
rs992315317 Chr.12: 26938377-26938377 on Build GRCh38
rs992925889 Chr.12: 26938440-26938440 on Build GRCh38
rs1036446296 Chr.12: 26938406-26938406 on Build GRCh38
rs924906618 Chr.12: 26938516-26938516 on Build GRCh38
rs967505969 Chr.12: 26938458-26938458 on Build GRCh38
rs145065811 Chr.12: 26938504-26938506 on Build GRCh38
rs764012786 Chr.12: 26938308-26938308 on Build GRCh38
rs533037462 Chr.12: 26938359-26938359 on Build GRCh38
rs565098350 Chr.12: 26938513-26938513 on Build GRCh38
rs913010613 Chr.12: 26938325-26938325 on Build GRCh38
rs895370867 Chr.12: 26938572-26938572 on Build GRCh38
rs12314170 Chr.12: 26938309-26938309 on Build GRCh38
rs543844611 Chr.12: 26938647-26938647 on Build GRCh38
rs963262647 Chr.12: 26938352-26938352 on Build GRCh38
rs966441717 Chr.12: 26938591-26938592 on Build GRCh38
rs759359507 Chr.12: 26938370-26938370 on Build GRCh38
rs912170510 Chr.12: 26938503-26938503 on Build GRCh38
rs1015865981 Chr.12: 26938343-26938343 on Build GRCh38
rs975065175 Chr.12: 26938518-26938518 on Build GRCh38
rs566411428 Chr.12: 26938703-26938703 on Build GRCh38
rs978449482 Chr.12: 26938584-26938584 on Build GRCh38
rs546412993 Chr.12: 26938433-26938433 on Build GRCh38
rs192816167 Chr.12: 26938415-26938415 on Build GRCh38
rs902850673 Chr.12: 26938717-26938717 on Build GRCh38
rs1042007846 Chr.12: 26938398-26938398 on Build GRCh38
rs994936176 Chr.12: 26938422-26938422 on Build GRCh38
rs932161616 Chr.12: 26938493-26938493 on Build GRCh38
rs1050630879 Chr.12: 26938495-26938495 on Build GRCh38
rs1014182982 Chr.12: 26938564-26938564 on Build GRCh38
rs1032758976 Chr.12: 26938593-26938593 on Build GRCh38
rs1034250901 Chr.12: 26938549-26938549 on Build GRCh38
rs757477275 Chr.12: 26938615-26938615 on Build GRCh38
rs568010032 Chr.12: 26938737-26938737 on Build GRCh38
rs986728587 Chr.12: 26938610-26938610 on Build GRCh38
rs974718580 Chr.12: 26938639-26938639 on Build GRCh38
rs931319035 Chr.12: 26938672-26938673 on Build GRCh38
rs977730652 Chr.12: 26938515-26938515 on Build GRCh38
rs1032495923 Chr.12: 26938750-26938750 on Build GRCh38
rs1037066888 Chr.12: 26938548-26938548 on Build GRCh38
rs386761374 Chr.12: 26938522-26938524 on Build GRCh38
rs978501927 Chr.12: 26938471-26938471 on Build GRCh38
rs926139534 Chr.12: 26938353-26938353 on Build GRCh38
rs970636316 Chr.12: 26938499-26938500 on Build GRCh38
rs932214586 Chr.12: 26938361-26938361 on Build GRCh38
rs926085757 Chr.12: 26938478-26938478 on Build GRCh38
rs919217167 Chr.12: 26938643-26938643 on Build GRCh38
rs527924814 Chr.12: 26938707-26938707 on Build GRCh38
rs556596224 Chr.12: 26938490-26938490 on Build GRCh38
rs188445449 Chr.12: 26938648-26938648 on Build GRCh38
rs940124183 Chr.12: 26938661-26938662 on Build GRCh38
rs11542284 Chr.12: 26938550-26938550 on Build GRCh38
rs567946644 Chr.12: 26938434-26938434 on Build GRCh38
rs1029705262 Chr.12: 26938333-26938333 on Build GRCh38
rs7979780 Chr.12: 26938611-26938611 on Build GRCh38
rs959786886 Chr.12: 26938492-26938492 on Build GRCh38
rs895748534 Chr.12: 26938562-26938562 on Build GRCh38
rs959981468 Chr.12: 26938315-26938315 on Build GRCh38
rs767795075 Chr.12: 26938640-26938640 on Build GRCh38
rs1020210767 Chr.12: 26938706-26938706 on Build GRCh38
rs796662949 Chr.12: 26938596-26938596 on Build GRCh38
rs549578529 Chr.12: 26938720-26938720 on Build GRCh38
rs945029034 Chr.12: 26938390-26938390 on Build GRCh38
rs936545960 Chr.12: 26938411-26938411 on Build GRCh38
rs540022980 Chr.12: 26938552-26938552 on Build GRCh38
rs534362605 Chr.12: 26938424-26938424 on Build GRCh38
rs996150630 Chr.12: 26938466-26938466 on Build GRCh38
rs573007702 Chr.12: 26938541-26938541 on Build GRCh38
rs761565893 Chr.12: 26938397-26938397 on Build GRCh38
rs895390307 Chr.12: 26938678-26938678 on Build GRCh38
rs944705853 Chr.12: 26938504-26938504 on Build GRCh38
rs185242591 Chr.12: 26938460-26938460 on Build GRCh38
rs532745692 Chr.12: 26938673-26938673 on Build GRCh38
rs61923399 Chr.12: 26938342-26938342 on Build GRCh38
rs930663949 Chr.12: 26938650-26938650 on Build GRCh38
rs371369108 Chr.12: 26938423-26938423 on Build GRCh38
rs750005491 Chr.12: 26938497-26938497 on Build GRCh38
rs115084512 Chr.12: 26938524-26938524 on Build GRCh38
rs944924638 Chr.12: 26938614-26938614 on Build GRCh38
rs1020349517 Chr.12: 26938565-26938565 on Build GRCh38
rs1001418001 Chr.12: 26938426-26938426 on Build GRCh38
rs992581348 Chr.12: 26938319-26938319 on Build GRCh38
rs180898686 Chr.12: 26938668-26938668 on Build GRCh38
rs1014550679 Chr.12: 26938690-26938690 on Build GRCh38
rs1004232583 Chr.12: 26938443-26938443 on Build GRCh38
rs766931833 Chr.12: 26938448-26938448 on Build GRCh38
rs1011872698 Chr.12: 26938514-26938514 on Build GRCh38

View Full Product Details