Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31078161-31078665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX11
Entrez Gene ID
1663
Gene Name
DEAD/H-box helicase 11
Gene Aliases
CHL1, CHLR1, KRG2, WABS
UniGene
Hs.443960
Gene Chromosome Location
Chr.12: 31073574-31104799 on Build GRCh38
Transcripts
NM_001257144.1 NM_001257145.1 NM_004399.2 NM_030653.3 NM_152438.1 XM_005253331.3 XM_005253333.3 XM_006719050.3 XM_011520592.2 XM_011520595.2 XM_011520597.2 XM_017018918.1 XM_017018919.1 XM_017018920.1 XM_017018921.1 XM_017018922.1 XM_017018923.1 XM_017018924.1 XM_017018925.1 XM_017018926.1 XM_017018927.1 XM_017018928.1 XM_017018929.1 XM_017018930.1 XM_017018931.1 XM_017018932.1 XM_017018933.1 XM_017018934.1 XM_017018938.1 XM_017018939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5529630 c.45T>A p.F15L Chr.12: 31078438-31078438 on Build GRCh38
6393930 c.49T>C p.F17L Chr.12: 31078442-31078442 on Build GRCh38
6393929 c.49T>C p.F17L Chr.12: 31078442-31078442 on Build GRCh38
5081270 c.144+5G>T p.? Chr.12: 31078542-31078542 on Build GRCh38
6393931 c.49T>C p.F17L Chr.12: 31078442-31078442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781518039 Chr.12: 31078586-31078586 on Build GRCh38
rs1811284 Chr.12: 31078224-31078224 on Build GRCh38
rs749873885 Chr.12: 31078506-31078506 on Build GRCh38
rs773162826 Chr.12: 31078365-31078365 on Build GRCh38
rs748184802 Chr.12: 31078264-31078264 on Build GRCh38
rs751149868 Chr.12: 31078302-31078302 on Build GRCh38
rs905053319 Chr.12: 31078526-31078526 on Build GRCh38
rs933412668 Chr.12: 31078385-31078385 on Build GRCh38
rs972056074 Chr.12: 31078351-31078351 on Build GRCh38
rs756744607 Chr.12: 31078305-31078305 on Build GRCh38
rs747032595 Chr.12: 31078575-31078575 on Build GRCh38
rs954788318 Chr.12: 31078624-31078624 on Build GRCh38
rs773350144 Chr.12: 31078419-31078420 on Build GRCh38
rs77842499 Chr.12: 31078204-31078204 on Build GRCh38
rs199851675 Chr.12: 31078487-31078487 on Build GRCh38
rs562931663 Chr.12: 31078363-31078363 on Build GRCh38
rs1016571030 Chr.12: 31078604-31078604 on Build GRCh38
rs758819549 Chr.12: 31078550-31078550 on Build GRCh38
rs879208105 Chr.12: 31078549-31078549 on Build GRCh38
rs527700858 Chr.12: 31078419-31078419 on Build GRCh38
rs145236589 Chr.12: 31078483-31078483 on Build GRCh38
rs780512953 Chr.12: 31078336-31078336 on Build GRCh38
rs749110936 Chr.12: 31078587-31078587 on Build GRCh38
rs773427749 Chr.12: 31078279-31078279 on Build GRCh38
rs878879524 Chr.12: 31078358-31078358 on Build GRCh38
rs769128850 Chr.12: 31078233-31078233 on Build GRCh38
rs4031358 Chr.12: 31078233-31078235 on Build GRCh38
rs368329103 Chr.12: 31078486-31078486 on Build GRCh38
rs759255266 Chr.12: 31078282-31078282 on Build GRCh38
rs767078889 Chr.12: 31078321-31078321 on Build GRCh38
rs750154502 Chr.12: 31078323-31078323 on Build GRCh38
rs7299908 Chr.12: 31078296-31078296 on Build GRCh38
rs765147088 Chr.12: 31078292-31078292 on Build GRCh38
rs768609131 Chr.12: 31078362-31078362 on Build GRCh38
rs376585013 Chr.12: 31078441-31078441 on Build GRCh38
rs538124837 Chr.12: 31078614-31078614 on Build GRCh38
rs903543463 Chr.12: 31078190-31078190 on Build GRCh38
rs765974869 Chr.12: 31078529-31078529 on Build GRCh38
rs2553159 Chr.12: 31078293-31078293 on Build GRCh38
rs755275094 Chr.12: 31078535-31078535 on Build GRCh38
rs150002506 Chr.12: 31078432-31078432 on Build GRCh38
rs771036752 Chr.12: 31078401-31078401 on Build GRCh38
rs551585416 Chr.12: 31078417-31078417 on Build GRCh38
rs771219222 Chr.12: 31078578-31078578 on Build GRCh38
rs779399256 Chr.12: 31078542-31078542 on Build GRCh38
rs749620384 Chr.12: 31078346-31078346 on Build GRCh38
rs779342078 Chr.12: 31078352-31078352 on Build GRCh38
rs147639058 Chr.12: 31078276-31078276 on Build GRCh38
rs765168834 Chr.12: 31078414-31078417 on Build GRCh38
rs1013387447 Chr.12: 31078217-31078217 on Build GRCh38
rs1026305145 Chr.12: 31078620-31078627 on Build GRCh38
rs892241192 Chr.12: 31078209-31078209 on Build GRCh38
rs762113306 Chr.12: 31078369-31078371 on Build GRCh38
rs755509483 Chr.12: 31078350-31078350 on Build GRCh38
rs746777307 Chr.12: 31078253-31078253 on Build GRCh38
rs755704784 Chr.12: 31078334-31078334 on Build GRCh38
rs963266976 Chr.12: 31078607-31078607 on Build GRCh38
rs566914559 Chr.12: 31078418-31078418 on Build GRCh38
rs1046226646 Chr.12: 31078488-31078488 on Build GRCh38
rs533511750 Chr.12: 31078416-31078416 on Build GRCh38
rs776950438 Chr.12: 31078366-31078367 on Build GRCh38
rs774360942 Chr.12: 31078485-31078485 on Build GRCh38
rs868073588 Chr.12: 31078449-31078449 on Build GRCh38
rs760415148 Chr.12: 31078514-31078514 on Build GRCh38
rs747314230 Chr.12: 31078370-31078370 on Build GRCh38
rs542177518 Chr.12: 31078218-31078218 on Build GRCh38
rs777958495 Chr.12: 31078559-31078559 on Build GRCh38
rs756431001 Chr.12: 31078202-31078202 on Build GRCh38
rs753315620 Chr.12: 31078544-31078544 on Build GRCh38
rs562066003 Chr.12: 31078230-31078230 on Build GRCh38
rs774453873 Chr.12: 31078238-31078238 on Build GRCh38

View Full Product Details