Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48153290-48153800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB8
Entrez Gene ID
140461
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
PP14212
UniGene
Hs.432699
Gene Chromosome Location
Chr.12: 48147788-48157595 on Build GRCh38
Transcripts
NM_001319296.1 NM_001319297.1 NM_001319298.1 NM_001319299.1 NM_001319300.1 NM_001319301.1 NM_024095.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4428783 c.87C>G p.I29M Chr.12: 48153410-48153410 on Build GRCh38
5927665 c.128G>A p.G43E Chr.12: 48153369-48153369 on Build GRCh38
1492951 c.97C>T p.P33S Chr.12: 48153400-48153400 on Build GRCh38
2005769 c.21T>C p.Y7Y Chr.12: 48153476-48153476 on Build GRCh38
6225361 c.89G>A p.R30H Chr.12: 48153408-48153408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913655184 Chr.12: 48153322-48153322 on Build GRCh38
rs920523619 Chr.12: 48153750-48153750 on Build GRCh38
rs746217590 Chr.12: 48153319-48153319 on Build GRCh38
rs368523694 Chr.12: 48153458-48153458 on Build GRCh38
rs573253511 Chr.12: 48153714-48153714 on Build GRCh38
rs756849001 Chr.12: 48153537-48153539 on Build GRCh38
rs11168428 Chr.12: 48153345-48153345 on Build GRCh38
rs771619467 Chr.12: 48153337-48153337 on Build GRCh38
rs531211168 Chr.12: 48153338-48153338 on Build GRCh38
rs369323738 Chr.12: 48153604-48153604 on Build GRCh38
rs775543638 Chr.12: 48153445-48153445 on Build GRCh38
rs780675708 Chr.12: 48153326-48153326 on Build GRCh38
rs1050123493 Chr.12: 48153739-48153739 on Build GRCh38
rs766782408 Chr.12: 48153516-48153516 on Build GRCh38
rs778875437 Chr.12: 48153506-48153506 on Build GRCh38
rs147288343 Chr.12: 48153462-48153462 on Build GRCh38
rs201422942 Chr.12: 48153370-48153370 on Build GRCh38
rs76905550 Chr.12: 48153440-48153440 on Build GRCh38
rs746741245 Chr.12: 48153740-48153740 on Build GRCh38
rs754252876 Chr.12: 48153519-48153519 on Build GRCh38
rs868615106 Chr.12: 48153559-48153559 on Build GRCh38
rs975030517 Chr.12: 48153720-48153720 on Build GRCh38
rs950118438 Chr.12: 48153777-48153777 on Build GRCh38
rs931869052 Chr.12: 48153686-48153686 on Build GRCh38
rs779542622 Chr.12: 48153399-48153399 on Build GRCh38
rs370749362 Chr.12: 48153351-48153351 on Build GRCh38
rs771662561 Chr.12: 48153427-48153427 on Build GRCh38
rs567644656 Chr.12: 48153355-48153355 on Build GRCh38
rs901720108 Chr.12: 48153341-48153341 on Build GRCh38
rs774665656 Chr.12: 48153712-48153712 on Build GRCh38
rs753282616 Chr.12: 48153530-48153531 on Build GRCh38
rs868063048 Chr.12: 48153417-48153417 on Build GRCh38
rs943195263 Chr.12: 48153699-48153699 on Build GRCh38
rs373639181 Chr.12: 48153441-48153441 on Build GRCh38
rs747598837 Chr.12: 48153426-48153426 on Build GRCh38
rs188638799 Chr.12: 48153468-48153468 on Build GRCh38
rs371610181 Chr.12: 48153430-48153430 on Build GRCh38
rs1021075966 Chr.12: 48153591-48153591 on Build GRCh38
rs548295200 Chr.12: 48153769-48153769 on Build GRCh38
rs887556193 Chr.12: 48153350-48153350 on Build GRCh38
rs139466759 Chr.12: 48153558-48153558 on Build GRCh38
rs61558457 Chr.12: 48153334-48153334 on Build GRCh38
rs769059257 Chr.12: 48153564-48153564 on Build GRCh38
rs530266306 Chr.12: 48153389-48153389 on Build GRCh38
rs930592905 Chr.12: 48153756-48153756 on Build GRCh38
rs1040578923 Chr.12: 48153605-48153605 on Build GRCh38
rs537342888 Chr.12: 48153530-48153530 on Build GRCh38
rs936110727 Chr.12: 48153549-48153549 on Build GRCh38
rs751583580 Chr.12: 48153356-48153356 on Build GRCh38
rs372798083 Chr.12: 48153748-48153748 on Build GRCh38
rs879101873 Chr.12: 48153465-48153465 on Build GRCh38
rs760297793 Chr.12: 48153389-48153390 on Build GRCh38
rs989159823 Chr.12: 48153574-48153574 on Build GRCh38
rs755429850 Chr.12: 48153520-48153520 on Build GRCh38
rs753291307 Chr.12: 48153548-48153548 on Build GRCh38
rs1014326424 Chr.12: 48153451-48153451 on Build GRCh38
rs936053856 Chr.12: 48153313-48153313 on Build GRCh38
rs376414157 Chr.12: 48153396-48153396 on Build GRCh38
rs113300976 Chr.12: 48153331-48153331 on Build GRCh38
rs565893897 Chr.12: 48153436-48153436 on Build GRCh38
rs547070872 Chr.12: 48153435-48153435 on Build GRCh38
rs759494741 Chr.12: 48153352-48153352 on Build GRCh38
rs376121340 Chr.12: 48153346-48153346 on Build GRCh38
rs763774928 Chr.12: 48153441-48153444 on Build GRCh38
rs11537785 Chr.12: 48153404-48153404 on Build GRCh38
rs987244264 Chr.12: 48153659-48153659 on Build GRCh38
rs767608278 Chr.12: 48153365-48153365 on Build GRCh38
rs1017141591 Chr.12: 48153388-48153388 on Build GRCh38
rs374413605 Chr.12: 48153391-48153391 on Build GRCh38
rs779266644 Chr.12: 48153542-48153542 on Build GRCh38
rs957902896 Chr.12: 48153572-48153572 on Build GRCh38
rs199904430 Chr.12: 48153487-48153487 on Build GRCh38
rs757411635 Chr.12: 48153359-48153359 on Build GRCh38
rs759440534 Chr.12: 48153439-48153439 on Build GRCh38
rs192857522 Chr.12: 48153320-48153320 on Build GRCh38
rs911706336 Chr.12: 48153690-48153690 on Build GRCh38
rs369642519 Chr.12: 48153332-48153332 on Build GRCh38
rs192726327 Chr.12: 48153771-48153771 on Build GRCh38
rs987298081 Chr.12: 48153333-48153333 on Build GRCh38

View Full Product Details