Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48472580-48473079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32D
Entrez Gene ID
23519
Gene Name
acidic nuclear phosphoprotein 32 family member D
Gene Aliases
PP32R2
UniGene
Hs.532658
Gene Chromosome Location
Chr.12: 48472665-48473060 on Build GRCh38
Transcripts
NM_012404.2 XM_011537896.2 XM_017018796.1

Gene Symbol
C12orf54
Entrez Gene ID
121273
Gene Name
chromosome 12 open reading frame 54
Gene Aliases
HSD-29, HSD-30
UniGene
Hs.98202
Gene Chromosome Location
Chr.12: 48413154-48496514 on Build GRCh38
Transcripts
NM_012404.2 XM_011537896.2 XM_017018796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986949 c.237C>T p.G79G Chr.12: 48472901-48472901 on Build GRCh38
4042235 c.341A>G p.N114S Chr.12: 48473005-48473005 on Build GRCh38
939823 c.34C>T p.R12W Chr.12: 48472698-48472698 on Build GRCh38
3812124 c.44C>T p.T15M Chr.12: 48472708-48472708 on Build GRCh38
939825 c.297A>C p.K99N Chr.12: 48472961-48472961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769079974 Chr.12: 48472909-48472909 on Build GRCh38
rs890426022 Chr.12: 48472996-48472996 on Build GRCh38
rs756960518 Chr.12: 48472640-48472640 on Build GRCh38
rs757763351 Chr.12: 48472995-48472995 on Build GRCh38
rs570410317 Chr.12: 48472673-48472673 on Build GRCh38
rs150593500 Chr.12: 48472706-48472706 on Build GRCh38
rs770289422 Chr.12: 48472908-48472908 on Build GRCh38
rs780433474 Chr.12: 48472846-48472847 on Build GRCh38
rs754324242 Chr.12: 48472618-48472618 on Build GRCh38
rs141807268 Chr.12: 48472698-48472698 on Build GRCh38
rs747041373 Chr.12: 48472785-48472785 on Build GRCh38
rs199783781 Chr.12: 48472764-48472764 on Build GRCh38
rs113589703 Chr.12: 48473044-48473044 on Build GRCh38
rs777415345 Chr.12: 48472637-48472637 on Build GRCh38
rs754851394 Chr.12: 48472780-48472780 on Build GRCh38
rs866550642 Chr.12: 48472721-48472721 on Build GRCh38
rs371961147 Chr.12: 48472816-48472816 on Build GRCh38
rs7966826 Chr.12: 48473009-48473009 on Build GRCh38
rs756248711 Chr.12: 48472974-48472974 on Build GRCh38
rs776059285 Chr.12: 48473005-48473005 on Build GRCh38
rs911428382 Chr.12: 48472897-48472897 on Build GRCh38
rs748563124 Chr.12: 48472991-48472991 on Build GRCh38
rs750090213 Chr.12: 48472771-48472771 on Build GRCh38
rs755224166 Chr.12: 48472987-48472987 on Build GRCh38
rs769470726 Chr.12: 48472991-48472992 on Build GRCh38
rs753531819 Chr.12: 48472831-48472831 on Build GRCh38
rs201684707 Chr.12: 48472862-48472862 on Build GRCh38
rs1052311498 Chr.12: 48472743-48472743 on Build GRCh38
rs762353947 Chr.12: 48472905-48472905 on Build GRCh38
rs752603319 Chr.12: 48472942-48472942 on Build GRCh38
rs142042575 Chr.12: 48473006-48473006 on Build GRCh38
rs945394900 Chr.12: 48472954-48472954 on Build GRCh38
rs761713730 Chr.12: 48473028-48473028 on Build GRCh38
rs201047451 Chr.12: 48472812-48472812 on Build GRCh38
rs138630151 Chr.12: 48472758-48472758 on Build GRCh38
rs748652681 Chr.12: 48472920-48472920 on Build GRCh38
rs747139165 Chr.12: 48472718-48472718 on Build GRCh38
rs778198916 Chr.12: 48473056-48473056 on Build GRCh38
rs376682025 Chr.12: 48472658-48472658 on Build GRCh38
rs755908495 Chr.12: 48472709-48472709 on Build GRCh38
rs751519124 Chr.12: 48473021-48473021 on Build GRCh38
rs778935683 Chr.12: 48472782-48472782 on Build GRCh38
rs549832830 Chr.12: 48473048-48473048 on Build GRCh38
rs139598237 Chr.12: 48472824-48472824 on Build GRCh38
rs781503751 Chr.12: 48472790-48472790 on Build GRCh38
rs375275376 Chr.12: 48472924-48472924 on Build GRCh38
rs749711435 Chr.12: 48472803-48472803 on Build GRCh38
rs145125600 Chr.12: 48472886-48472886 on Build GRCh38
rs771232119 Chr.12: 48472724-48472724 on Build GRCh38
rs376422835 Chr.12: 48472699-48472699 on Build GRCh38
rs780345556 Chr.12: 48472888-48472888 on Build GRCh38
rs746441828 Chr.12: 48472998-48472998 on Build GRCh38
rs753147438 Chr.12: 48472686-48472686 on Build GRCh38
rs775401523 Chr.12: 48473012-48473012 on Build GRCh38
rs764174334 Chr.12: 48472968-48472968 on Build GRCh38
rs747396482 Chr.12: 48472970-48472970 on Build GRCh38
rs559434498 Chr.12: 48472684-48472684 on Build GRCh38
rs750180206 Chr.12: 48472708-48472708 on Build GRCh38
rs374647516 Chr.12: 48472639-48472639 on Build GRCh38
rs762862538 Chr.12: 48473015-48473015 on Build GRCh38
rs777020204 Chr.12: 48472742-48472742 on Build GRCh38
rs758770989 Chr.12: 48473052-48473052 on Build GRCh38
rs368938365 Chr.12: 48472629-48472629 on Build GRCh38
rs771323740 Chr.12: 48472672-48472672 on Build GRCh38
rs748130555 Chr.12: 48472728-48472728 on Build GRCh38
rs570431628 Chr.12: 48472607-48472607 on Build GRCh38
rs374246487 Chr.12: 48472753-48472753 on Build GRCh38
rs145165709 Chr.12: 48472939-48472939 on Build GRCh38
rs753906042 Chr.12: 48473034-48473034 on Build GRCh38
rs747036685 Chr.12: 48472608-48472608 on Build GRCh38
rs780187865 Chr.12: 48473033-48473033 on Build GRCh38
rs765013681 Chr.12: 48472834-48472834 on Build GRCh38
rs980459310 Chr.12: 48472835-48472835 on Build GRCh38
rs374421529 Chr.12: 48472666-48472666 on Build GRCh38
rs767002083 Chr.12: 48472823-48472823 on Build GRCh38
rs149836219 Chr.12: 48473051-48473051 on Build GRCh38
rs772503300 Chr.12: 48472823-48472827 on Build GRCh38
rs147147368 Chr.12: 48472932-48472932 on Build GRCh38
rs149297885 Chr.12: 48472772-48472772 on Build GRCh38
rs934283979 Chr.12: 48472868-48472868 on Build GRCh38
rs570494201 Chr.12: 48473041-48473041 on Build GRCh38
rs768994706 Chr.12: 48472808-48472808 on Build GRCh38
rs752199492 Chr.12: 48472633-48472633 on Build GRCh38
rs745983472 Chr.12: 48472879-48472879 on Build GRCh38
rs770483490 Chr.12: 48473001-48473001 on Build GRCh38
rs199826854 Chr.12: 48472828-48472828 on Build GRCh38
rs770065599 Chr.12: 48472752-48472752 on Build GRCh38
rs989390704 Chr.12: 48472908-48472908 on Build GRCh38
rs776182487 Chr.12: 48472754-48472754 on Build GRCh38
rs762950778 Chr.12: 48472962-48472962 on Build GRCh38
rs760092788 Chr.12: 48472628-48472628 on Build GRCh38
rs749574437 Chr.12: 48472907-48472907 on Build GRCh38
rs756188573 Chr.12: 48473019-48473019 on Build GRCh38
rs774755802 Chr.12: 48472911-48472911 on Build GRCh38
rs772690730 Chr.12: 48472927-48472927 on Build GRCh38
rs374148537 Chr.12: 48472657-48472657 on Build GRCh38
rs527368156 Chr.12: 48472985-48472985 on Build GRCh38
rs750420888 Chr.12: 48472970-48472970 on Build GRCh38
rs759908744 Chr.12: 48472746-48472746 on Build GRCh38
rs935731011 Chr.12: 48472606-48472606 on Build GRCh38
rs745701997 Chr.12: 48472643-48472643 on Build GRCh38
rs754837794 Chr.12: 48472833-48472833 on Build GRCh38
rs765108512 Chr.12: 48472940-48472940 on Build GRCh38
rs752691227 Chr.12: 48472836-48472836 on Build GRCh38
rs1043033308 Chr.12: 48472630-48472630 on Build GRCh38
rs759998457 Chr.12: 48472681-48472681 on Build GRCh38
rs779269398 Chr.12: 48472900-48472900 on Build GRCh38
rs7956679 Chr.12: 48472802-48472802 on Build GRCh38
rs770454318 Chr.12: 48472933-48472933 on Build GRCh38
rs760217570 Chr.12: 48472826-48472826 on Build GRCh38
rs926434534 Chr.12: 48472667-48472667 on Build GRCh38
rs778286922 Chr.12: 48472997-48472997 on Build GRCh38
rs780959216 Chr.12: 48472641-48472641 on Build GRCh38
rs749170529 Chr.12: 48472712-48472712 on Build GRCh38
rs138194545 Chr.12: 48472692-48472692 on Build GRCh38
rs1026562901 Chr.12: 48472797-48472797 on Build GRCh38
rs377221100 Chr.12: 48472646-48472646 on Build GRCh38
rs770119437 Chr.12: 48472799-48472799 on Build GRCh38
rs758465937 Chr.12: 48472845-48472845 on Build GRCh38
rs764082472 Chr.12: 48473016-48473016 on Build GRCh38
rs780082324 Chr.12: 48472981-48472981 on Build GRCh38
rs188770785 Chr.12: 48472715-48472715 on Build GRCh38
rs367801459 Chr.12: 48472776-48472776 on Build GRCh38
rs746071349 Chr.12: 48472794-48472794 on Build GRCh38
rs776346690 Chr.12: 48472936-48472936 on Build GRCh38
rs777687157 Chr.12: 48472716-48472716 on Build GRCh38
rs773400972 Chr.12: 48472660-48472660 on Build GRCh38
rs150406578 Chr.12: 48472650-48472650 on Build GRCh38

View Full Product Details