Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48486089-48486588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf54
Entrez Gene ID
121273
Gene Name
chromosome 12 open reading frame 54
Gene Aliases
HSD-29, HSD-30
UniGene
Hs.98202
Gene Chromosome Location
Chr.12: 48413154-48496514 on Build GRCh38
Transcripts
NM_152319.3 XM_005268636.3 XM_011537896.2 XM_017018796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350470 c.66-10T>G p.? Chr.12: 48486168-48486168 on Build GRCh38
7351551 c.96+1G>C p.? Chr.12: 48486209-48486209 on Build GRCh38
6285937 c.66-4G>C p.? Chr.12: 48486174-48486174 on Build GRCh38
3792626 c.87G>C p.M29I Chr.12: 48486199-48486199 on Build GRCh38
6376213 c.96G>T p.Q32H Chr.12: 48486208-48486208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559027208 Chr.12: 48486431-48486431 on Build GRCh38
rs770803237 Chr.12: 48486214-48486214 on Build GRCh38
rs561269522 Chr.12: 48486160-48486160 on Build GRCh38
rs1055105144 Chr.12: 48486119-48486119 on Build GRCh38
rs1005545997 Chr.12: 48486564-48486564 on Build GRCh38
rs370921531 Chr.12: 48486163-48486163 on Build GRCh38
rs1014898219 Chr.12: 48486435-48486435 on Build GRCh38
rs1032527371 Chr.12: 48486488-48486488 on Build GRCh38
rs900572610 Chr.12: 48486493-48486493 on Build GRCh38
rs897749130 Chr.12: 48486438-48486438 on Build GRCh38
rs200634485 Chr.12: 48486138-48486138 on Build GRCh38
rs777479964 Chr.12: 48486459-48486460 on Build GRCh38
rs372584517 Chr.12: 48486228-48486228 on Build GRCh38
rs11168599 Chr.12: 48486168-48486168 on Build GRCh38
rs927803810 Chr.12: 48486351-48486351 on Build GRCh38
rs2068293 Chr.12: 48486307-48486307 on Build GRCh38
rs974990887 Chr.12: 48486180-48486180 on Build GRCh38
rs774547736 Chr.12: 48486238-48486238 on Build GRCh38
rs528496193 Chr.12: 48486247-48486247 on Build GRCh38
rs374989249 Chr.12: 48486186-48486186 on Build GRCh38
rs763040985 Chr.12: 48486146-48486146 on Build GRCh38
rs758141471 Chr.12: 48486191-48486191 on Build GRCh38
rs946239403 Chr.12: 48486129-48486129 on Build GRCh38
rs571431402 Chr.12: 48486261-48486261 on Build GRCh38
rs772385179 Chr.12: 48486260-48486260 on Build GRCh38
rs866115755 Chr.12: 48486117-48486117 on Build GRCh38
rs4620769 Chr.12: 48486125-48486125 on Build GRCh38
rs1024849640 Chr.12: 48486453-48486453 on Build GRCh38
rs539326852 Chr.12: 48486426-48486426 on Build GRCh38
rs550936506 Chr.12: 48486376-48486376 on Build GRCh38
rs952335082 Chr.12: 48486544-48486544 on Build GRCh38
rs12579547 Chr.12: 48486387-48486387 on Build GRCh38
rs992001240 Chr.12: 48486246-48486246 on Build GRCh38
rs932101447 Chr.12: 48486414-48486414 on Build GRCh38
rs969167230 Chr.12: 48486267-48486267 on Build GRCh38
rs988223301 Chr.12: 48486536-48486536 on Build GRCh38
rs768630882 Chr.12: 48486229-48486229 on Build GRCh38
rs565332707 Chr.12: 48486495-48486495 on Build GRCh38
rs544504612 Chr.12: 48486507-48486507 on Build GRCh38
rs937844330 Chr.12: 48486372-48486372 on Build GRCh38
rs1045313121 Chr.12: 48486344-48486344 on Build GRCh38
rs748780743 Chr.12: 48486494-48486494 on Build GRCh38
rs905481459 Chr.12: 48486353-48486353 on Build GRCh38
rs754452949 Chr.12: 48486183-48486183 on Build GRCh38
rs902504055 Chr.12: 48486416-48486416 on Build GRCh38
rs566776403 Chr.12: 48486422-48486422 on Build GRCh38
rs767016841 Chr.12: 48486178-48486178 on Build GRCh38
rs545676144 Chr.12: 48486296-48486297 on Build GRCh38
rs777685814 Chr.12: 48486136-48486136 on Build GRCh38
rs770891213 Chr.12: 48486142-48486142 on Build GRCh38
rs925002713 Chr.12: 48486318-48486318 on Build GRCh38
rs368212280 Chr.12: 48486474-48486474 on Build GRCh38
rs762482326 Chr.12: 48486164-48486167 on Build GRCh38
rs768881712 Chr.12: 48486149-48486149 on Build GRCh38
rs559650289 Chr.12: 48486195-48486195 on Build GRCh38
rs887806947 Chr.12: 48486423-48486423 on Build GRCh38
rs567051517 Chr.12: 48486330-48486330 on Build GRCh38
rs1013506641 Chr.12: 48486440-48486440 on Build GRCh38
rs761909752 Chr.12: 48486256-48486256 on Build GRCh38
rs980572487 Chr.12: 48486484-48486484 on Build GRCh38
rs775522769 Chr.12: 48486143-48486143 on Build GRCh38
rs187824566 Chr.12: 48486199-48486199 on Build GRCh38
rs367940723 Chr.12: 48486222-48486222 on Build GRCh38
rs575503148 Chr.12: 48486296-48486296 on Build GRCh38
rs141753072 Chr.12: 48486207-48486207 on Build GRCh38
rs1040520239 Chr.12: 48486402-48486402 on Build GRCh38
rs753361083 Chr.12: 48486181-48486181 on Build GRCh38
rs913528526 Chr.12: 48486539-48486539 on Build GRCh38
rs1042430501 Chr.12: 48486413-48486413 on Build GRCh38
rs946585958 Chr.12: 48486391-48486391 on Build GRCh38
rs756372552 Chr.12: 48486289-48486289 on Build GRCh38
rs373446482 Chr.12: 48486154-48486154 on Build GRCh38
rs145409227 Chr.12: 48486316-48486316 on Build GRCh38

View Full Product Details